Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HQSTLAPLL AVHKDEGSDP RRETPPGADP REYCTSDRDI VEVVRTEYVY TRPPPWSDTL PTIHVVTPTY SRPVQKAELT RMANTLLHVP NLHWLVVEDA PRRTPLTARL LRDTGLNYTH LHVETPRNYK LRGDARDPRI PRGTMQRNLA LRWLRETFPR NSSQPGVVYF ADDDNTYSLE LFEEMRSTRR VSVWPVAFVG GLRYEAPRVN GAGKVVGWKT VFDPHRPFAI DMAGFAVNLR LILQRSQAYF KLRGVKGGYQ ESSLLRELVT LNDLEPKAAN CTKILVWHTR TEKPVLVNEG KKGFTDPSVE I (25-334 aa encoded by BC010466)
CD57, also known as HNK-1 and Leu-7, is an oligosaccharide antigenic determinant present on a variety of polypeptides, lipids, and chondroitin sulphate proteoglycans. Its function is poorly understood. CD57 is expressed on a subset of natural killer (NK) cells and NK T cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-1,3-glucuronyltransferase 1; Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1; GlcAT-P; GlcUAT-P; Glucuronosyltransferase P; glucuronosyltransferase-P; HNK-1; Leu-7; LEU7 antigen; UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase
Gene Aliases: 0710007K08Rik; AI846286; B3GAT1; CD57; GlcAT-P; GLCATP; GLCUATP; HNK1; LEU7; NK-1; NK1
UniProt ID: (Human) Q9P2W7, (Mouse) Q9CW73
Entrez Gene ID: (Human) 27087, (Mouse) 76898
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support