Product References
Long-term culture with lipopolysaccharide induces dose-dependent cytostatic and cytotoxic effects in THP-1 monocytes.
Toxicology in vitro : an international journal published in association with BIBRA
Mytych J,Romerowicz-Misielak M,Koziorowski M
MA110259 was used in western blot to report the effects of long-term lipopolysaccharide exposure on monocytes
Tue Aug 01 00:00:00 UTC 2017
Cross-reactive multifunctional CD4+ T cell responses against Salmonella enterica serovars Typhi, Paratyphi A and Paratyphi B in humans following immunization with live oral typhoid vaccine Ty21a.
Clinical immunology (Orlando, Fla.)
Wahid R,Fresnay S,Levine MM,Sztein MB
MA110259 was used in flow cytometry to determine the S. Typhi, S. Paratyphi A, and S. Paratyphi B cross-reactive CD4 positive T cell responses elicited by immunization with Ty21a
Thu Dec 01 00:00:00 UTC 2016
Mimicking the tumour microenvironment of chronic lymphocytic leukaemia in vitro critically depends on the type of B-cell receptor stimulation.
British journal of cancer
Rombout A,Lust S,Offner F,Naessens E,Verhasselt B,Philippé J
MA110259 was used in flow cytometry to develop a robust in vitro model to study chronic lymphocytic leukemia B cell receptor ligation
Tue Mar 15 00:00:00 UTC 2016
Strain-specific Plasmodium falciparum multifunctional CD4(+) T cell cytokine expression in Malian children immunized with the FMP2.1/AS02A vaccine candidate.
Vaccine
Graves SF,Kouriba B,Diarra I,Daou M,Niangaly A,Coulibaly D,Keita Y,Laurens MB,Berry AA,Vekemans J,Ripley Ballou W,Lanar DE,Dutta S,Gray Heppner D,Soisson L,Diggs CL,Thera MA,Doumbo OK,Plowe CV,Sztein MB,Lyke KE
MA110259 was used in flow cytometry to characterize the immune responses of children immunized with Plasmodium falciparum apical membrane antigen 1
Tue May 17 00:00:00 UTC 2016
IL-12-secreting CD19-targeted cord blood-derived T cells for the immunotherapy of B-cell acute lymphoblastic leukemia.
Leukemia
Pegram HJ,Purdon TJ,van Leeuwen DG,Curran KJ,Giralt SA,Barker JN,Brentjens RJ
MA1-10259 was used in flow cytometry to developed a strategy to expand umbilical cord blood T cells and test their effects after transplantation
Sun Feb 01 00:00:00 UTC 2015
Differential Immune Profiles in Two Pandemic Influenza A(H1N1)pdm09 Virus Waves at Pandemic Epicenter.
Archives of medical research
Arriaga-Pizano L,Ferat-Osorio E,Rodríguez-Abrego G,Mancilla-Herrera I,Domínguez-Cerezo E,Valero-Pacheco N,Pérez-Toledo M,Lozano-Patiño F,Laredo-Sánchez F,Malagón-Rangel J,Nellen-Hummel H,González-Bonilla C,Arteaga-Troncoso G,Cérbulo-Vázquez A,Pastelin-Palacios R,Klenerman P,Isibasi A,López-Macías C
MA1-10259 was used in flow cytometry to analyze the difference in immune profiles at the pandemic epicenter in two pandemic Influenza A (H1N1) pdm09 virus waves
Sun Nov 01 00:00:00 UTC 2015
HIV-1 Nef and Vpu Interfere with L-Selectin (CD62L) Cell Surface Expression To Inhibit Adhesion and Signaling in Infected CD4+ T Lymphocytes.
Journal of virology
Vassena L,Giuliani E,Koppensteiner H,Bolduan S,Schindler M,Doria M
MA1-10259 was used in immunohistochemistry and western blot to study dysregulation of CD62L expression during HIV infection
Fri May 01 00:00:00 UTC 2015
Monitoring of antigen-specific CD8 T cells in patients with type 1 diabetes treated with antiCD3 monoclonal antibodies.
Clinical immunology (Orlando, Fla.)
Cernea S,Herold KC
MA1-10259 was used in flow cytometry to investigate how anti-CD3 monoclonal antibodies alter human immune responses in type 1 diabetes
Mon Feb 01 00:00:00 UTC 2010
Nicotine downregulates the l-selectin system that mediates cytotrophoblast emigration from cell columns and attachment to the uterine wall.
Reproductive toxicology (Elmsford, N.Y.)
Zdravkovic T,Genbacev O,Prakobphol A,Cvetkovic M,Schanz A,McMaster M,Fisher SJ
MA1-10259 was used in immunohistochemistry - paraffin section to elucidate how nicotine impairs the development of cell columns
Sat Jul 01 00:00:00 UTC 2006
A role for the L-selectin adhesion system in mediating cytotrophoblast emigration from the placenta.
Developmental biology
Prakobphol A,Genbacev O,Gormley M,Kapidzic M,Fisher SJ
MA1-10259 was used in immunohistochemistry - frozen section to propose an autocrine role for the cytotrophoblast L-selectin adhesion system in forming and maintaining cell columns during the early stages of placental development
Sun Oct 01 00:00:00 UTC 2006
Contemporaneous fluctuations in T cell responses to persistent herpes virus infections.
European journal of immunology
Crough T,Burrows JM,Fazou C,Walker S,Davenport MP,Khanna R
MA1-10259 was used in flow cytometry to show that T cell expansion and contractions in persistent viral infections is not limited to the acute phase of infection, but continues in latent infection
Sat Jan 01 00:00:00 UTC 2005
Differential expression of human granzymes A, B, and K in natural killer cells and during CD8+ T cell differentiation in peripheral blood.
European journal of immunology
Bratke K,Kuepper M,Bade B,Virchow JC,Luttmann W
MA1-10259 was used in flow cytometry to use flow cytometry to study granzyme A, B, and K expression in human peripheral blood lymphocytes
Thu Sep 01 00:00:00 UTC 2005
Phenotypic and functional differences between human saphenous vein (HSVEC) and umbilical vein (HUVEC) endothelial cells.
Atherosclerosis
Tan PH,Chan C,Xue SA,Dong R,Ananthesayanan B,Manunta M,Kerouedan C,Cheshire NJ,Wolfe JH,Haskard DO,Taylor KM,George AJ
MA1-10259 was used in flow cytometry to compare human saphenous vein endothelial cells with human umbilical vein endothelial cells
Thu Apr 01 00:00:00 UTC 2004
Role of L-selectin in the vascular homing of peripheral blood-derived endothelial progenitor cells.
Journal of immunology (Baltimore, Md. : 1950)
Biancone L,Cantaluppi V,Duò D,Deregibus MC,Torre C,Camussi G
MA1-10259 was used in flow cytometry to report that leukocyte adhesion is involved in the vascular homing of endothelial progenitor cells
Fri Oct 15 00:00:00 UTC 2004
Proinflammatory phenotype of endothelial cells after coculture with biomaterial-treated blood cells.
Journal of biomedical materials research. Part A
Lester EA,Babensee JE
MA1-10259 was used in flow cytometry to study interactions among endothelial cells, blood, and biomaterials
Sat Mar 01 00:00:00 UTC 2003
Characterization of multiple CD34+ cell populations in cord blood.
Journal of hematotherapy & stem cell research
Greco NJ,Lee WR,Kurtz J,Seetharaman S,Moroff G
MA1-10259 was used in flow cytometry to characterize CD34+ umbilical cord blood cells
Tue Apr 01 00:00:00 UTC 2003
Role for granulocyte colony-stimulating factor in the generation of human T regulatory type 1 cells.
Blood
Rutella S,Pierelli L,Bonanno G,Sica S,Ameglio F,Capoluongo E,Mariotti A,Scambia G,d'Onofrio G,Leone G
MA1-10259 was used in flow cytometry to measure interleukin-10 and transforming growth factor-beta1 produced by T cells from healthy volunteer donors treated with recombinant human G-CSF.
Tue Oct 01 00:00:00 UTC 2002
Tropism of varicella-zoster virus for human tonsillar CD4(+) T lymphocytes that express activation, memory, and skin homing markers.
Journal of virology
Ku CC,Padilla JA,Grose C,Butcher EC,Arvin AM
MA1-10259 was used in flow cytometry to report that Varicella-zoster virus infects human tonsillar CD4(+) T cells.
Fri Nov 01 00:00:00 UTC 2002
Expression and coreceptor activity of STRL33/Bonzo on primary peripheral blood lymphocytes.
Blood
Sharron M,Pöhlmann S,Price K,Lolis E,Tsang M,Kirchhoff F,Doms RW,Lee B
MA1-10259 was used in flow cytometry to characterize the localization and function of STRL33/Bonzo.
Sat Jul 01 00:00:00 UTC 2000
Relationship between selectin-mediated rolling of hematopoietic stem and progenitor cells and progression in hematopoietic development.
Blood
Greenberg AW,Kerr WG,Hammer DA
MA1-10259 was used in blocking assay to identify adhesion molecules and study mechanisms that regulate hematopoietic stem and progenitor cell homing to the bone marrow.
Sat Jan 15 00:00:00 UTC 2000
Quantification of CD4, CCR5, and CXCR4 levels on lymphocyte subsets, dendritic cells, and differentially conditioned monocyte-derived macrophages.
Proceedings of the National Academy of Sciences of the United States of America
Lee B,Sharron M,Montaner LJ,Weissman D,Doms RW
MA1-10259 was used in flow cytometry to determine the number of CD4, CCR5, and CXCR4 antibody-binding sites on various T cells and macrophages.
Tue Apr 27 00:00:00 UTC 1999