Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
FIGURE: 1 / 1
The human CD62L is a 74-95 kDa glycoprotein member of the selectin family of adhesion receptors. L-Selectin is comprised of an aminoterminal C-type lectin binding domain, an epidermal growth factor-like domain, two short consensus repeat (SCR) sequences homologous to those found in complement binding proteins, a short spacer region, a transmembrane region and a short cytoplasmic region. Human CD62L (L-Selectin) is constitutively expressed on all classes of leukocytes including lymphocytes (except a substantial population of memory T cells), monocytes and polymorphonuclear cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD62 antigen-like family member L; CD62L; hLHRc; L-selectin; LAM-1; LECAM1; Leu-8; Leukocyte adhesion molecule 1; Leukocyte-endothelial cell adhesion molecule 1; Ly-22; Lymph node homing receptor; Lymphocyte antigen 22; Lymphocyte surface MEL-14 antigen
Gene Aliases: AI528707; CD62L; L-selectin; LECAM-1; Lnhr; Ly-22; Ly-m22; Ly22; Lyam-1; Lyam1; Sell
UniProt ID: (Mouse) P18337
Entrez Gene ID: (Mouse) 20343
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support