Product References
Mesenchymal stem cell-derived exosomes can alleviate GVHD and preserve the GVL effect in allogeneic stem cell transplantation animal models.
Frontiers in immunology
Jiang Y,Zhao J,Wang M,Huang F,Li J,Liu R,Wan J,Hao S
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Fri Dec 22 00:00:00 UTC 2023
Plexin B1 controls Treg numbers, limits allergic airway inflammation, and regulates mucins.
Frontiers in immunology
Chapoval SP,Gao H,Fanaroff R,Keegan AD
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Tue Feb 20 00:00:00 UTC 2024
CD138 promotes the accumulation and activation of autoreactive T cells in autoimmune MRL/lpr mice.
Experimental and therapeutic medicine
Xie T,Liu X,Li P
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Fri Dec 01 00:00:00 UTC 2023
CD138 promotes the accumulation and activation of autoreactive T cells in autoimmune MRL/lpr mice.
Experimental and therapeutic medicine
Xie T,Liu X,Li P
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Fri Dec 01 00:00:00 UTC 2023
HSF1 promotes CD69+ Treg differentiation to inhibit colitis progression.
Theranostics
Yu L,Zhou B,Zhu Y,Li L,Zhong Y,Zhu L,Wang H,Chen H,Xu J,Guo T,Feng L,Wang X,Cai Z,Wang J,Jin H
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Tue Apr 18 00:00:00 UTC 2023
Modulating CD40 and integrin signaling in the proinflammatory nexus using a 15-amino-acid peptide, KGYY15.
The Journal of biological chemistry
Vaitaitis GM,Wagner DH
17-0691-82 was used in Flow cytometry/Cell sorting to demonstrate interactions between CD40 and integrin CD11a/CD18, which complicates the understanding of the inflammatory nexus and how to prevent autoinflammation.
Mon May 01 00:00:00 UTC 2023
TRAF6 controls T cell homeostasis by maintaining the equilibrium of MALT1 scaffolding and protease functions.
Frontiers in immunology
O'Neill TJ,Gewies A,Seeholzer T,Krappmann D
17-0691-82 was used in Flow Cytometry to demonstrate that TRAF6 deletion or MALT1 paracaspase inactivation are highly interdependent in causing the distinct immune pathologies.
Mon Feb 13 00:00:00 UTC 2023
Novel Bruton's tyrosine kinase inhibitor TAS5315 suppresses the progression of inflammation and joint destruction in rodent collagen-induced arthritis.
PloS one
Akasaka D,Iguchi S,Kaneko R,Yoshiga Y,Kajiwara D,Nakachi Y,Noma N,Tanaka K,Shimizu A,Hosoi F
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Mon Feb 27 00:00:00 UTC 2023
TCR repertoire landscape reveals macrophage-mediated clone deletion in endotoxin tolerance.
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Zhao J,Jia L,Tao Y,Zhao X,Yang J,Lu Y,Yan Y,Mao L,Hu L,Lu J,Guo M,Chen C,Zhou Y,Wen Z,He Z,Xu L
17-0691-82 was used in Flow cytometry/Cell sorting to suggest that the clone selection in thymus in Endotoxin tolerance may confer protection against microbial sepsis.
Wed Mar 01 00:00:00 UTC 2023
Mesenchymal stromal cells equipped by IFNα empower T cells with potent anti-tumor immunity.
Oncogene
Zhang T,Wang Y,Li Q,Lin L,Xu C,Xue Y,Hu M,Shi Y,Wang Y
17-0691-82 was used in Flow Cytometry to conclude that IFNα-MSCs represent a novel tumor immunotherapy strategy, especially when combined with PD-L1 blockade.
Tue Mar 01 00:00:00 UTC 2022
Intranasal Immunization With a c-di-GMP-Adjuvanted Acellular Pertussis Vaccine Provides Superior Immunity Against Bordetella pertussis in a Mouse Model.
Frontiers in immunology
Jiang W,Wang X,Su Y,Cai L,Li J,Liang J,Gu Q,Sun M,Shi L
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Tue May 03 00:00:00 UTC 2022
Meningeal lymphatic drainage promotes T cell responses against Toxoplasma gondii but is dispensable for parasite control in the brain.
eLife
Kovacs MA,Cowan MN,Babcock IW,Sibley LA,Still K,Batista SJ,Labuzan SA,Sethi I,Harris TH
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Wed Dec 21 00:00:00 UTC 2022
An autonomous TCR signal-sensing switch influences CD4/CD8 lineage choice in mice.
Communications biology
Basu J,Zha J,Nicolas E,Coulton M,Czyzewicz P,Hua X,Ge L,Kappes DJ
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Fri Jan 21 00:00:00 UTC 2022
CP-25 exerts a protective effect against ConA-induced hepatitis via regulating inflammation and immune response.
Frontiers in pharmacology
Li N,Wu JJ,Qi M,Wang ZY,Zhang SN,Li XQ,Chen TT,Wang MF,Zhang LL,Wei W,Sun WY
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Sat Dec 03 00:00:00 UTC 2022
A novel costimulatory molecule gene-modified leukemia cell-derived exosome-targeted CD4+ T cell vaccine efficiently enhances anti-leukemia immunity.
Frontiers in immunology
Li J,Huang F,Jiang Y,Zhao J,Wan J,Hao S
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Tue Dec 06 00:00:00 UTC 2022
ADAM12 is a costimulatory molecule that determines Th1 cell fate and mediates tissue inflammation.
Cellular & molecular immunology
Liu Y,Bockermann R,Hadi M,Safari I,Carrion B,Kveiborg M,Issazadeh-Navikas S
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Sun Aug 01 00:00:00 UTC 2021
Uncovering a novel role of PLCβ4 in selectively mediating TCR signaling in CD8+ but not CD4+ T cells.
The Journal of experimental medicine
Sasai M,Ma JS,Okamoto M,Nishino K,Nagaoka H,Takashima E,Pradipta A,Lee Y,Kosako H,Suh PG,Yamamoto M
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Mon Jul 05 00:00:00 UTC 2021
Fasudil prevents liver fibrosis via activating natural killer cells and suppressing hepatic stellate cells.
World journal of gastroenterology
Han QJ,Mu YL,Zhao HJ,Zhao RR,Guo QJ,Su YH,Zhang J
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Mon Jun 28 00:00:00 UTC 2021
TRPM8 channel augments T-cell activation and proliferation.
Cell biology international
Acharya TK,Tiwari A,Majhi RK,Goswami C
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2021
Cell-permeable transgelin-2 as a potent therapeutic for dendritic cell-based cancer immunotherapy.
Journal of hematology & oncology
Kim HR,Park JS,Park JH,Yasmin F,Kim CH,Oh SK,Chung IJ,Jun CD
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Wed Mar 17 00:00:00 UTC 2021
Suppression of mucosal Th17 memory responses by acellular pertussis vaccines enhances nasal Bordetella pertussis carriage.
NPJ vaccines
Dubois V,Chatagnon J,Thiriard A,Bauderlique-Le Roy H,Debrie AS,Coutte L,Locht C
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Fri Jan 08 00:00:00 UTC 2021
STING Activated Tumor-Intrinsic Type I Interferon Signaling Promotes CXCR3 Dependent Antitumor Immunity in Pancreatic Cancer.
Cellular and molecular gastroenterology and hepatology
Vonderhaar EP,Barnekow NS,McAllister D,McOlash L,Eid MA,Riese MJ,Tarakanova VL,Johnson BD,Dwinell MB
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Mon Jan 24 00:00:00 UTC 2022
GPR120 induces regulatory dendritic cells by inhibiting HK2-dependent glycolysis to alleviate fulminant hepatic failure.
Cell death & disease
Yu H,Yang W,Huang J,Miao X,Wang B,Ren X,Gu Y,Wang Q,Ding X,Guo X,Qian F,Zhang Y,Xu H,Zheng L,Jin M
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Thu Dec 16 00:00:00 UTC 2021
Robust induction of TRMs by combinatorial nanoshells confers cross-strain sterilizing immunity against lethal influenza viruses.
Molecular therapy. Methods & clinical development
Lin PH,Liang CY,Yao BY,Chen HW,Pan CF,Wu LL,Lin YH,Hsu YS,Liu YH,Chen PJ,Hu CJ,Yang HC
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Fri Jun 11 00:00:00 UTC 2021
SPARC regulation of PMN clearance protects from pristane-induced lupus and rheumatoid arthritis.
iScience
Sangaletti S,Botti L,Gulino A,Lecis D,Bassani B,Portararo P,Milani M,Cancila V,De Cecco L,Dugo M,Tripodo C,Colombo MP
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Fri Jun 25 00:00:00 UTC 2021
Hybrid Gene Origination Creates Human-Virus Chimeric Proteins during Infection.
Cell
Ho JSY,Angel M,Ma Y,Sloan E,Wang G,Martinez-Romero C,Alenquer M,Roudko V,Chung L,Zheng S,Chang M,Fstkchyan Y,Clohisey S,Dinan AM,Gibbs J,Gifford R,Shen R,Gu Q,Irigoyen N,Campisi L,Huang C,Zhao N,Jones JD,van Knippenberg I,Zhu Z,Moshkina N,Meyer L,Noel J,Peralta Z,Rezelj V,Kaake R,Rosenberg B,Wang B,Wei J,Paessler S,Wise HM,Johnson J,Vannini A,Amorim MJ,Baillie JK,Miraldi ER,Benner C,Brierley I,Digard P,Łuksza M,Firth AE,Krogan N,Greenbaum BD,MacLeod MK,van Bakel H,Garcìa-Sastre A,Yewdell JW,Hutchinson E,Marazzi I
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Thu Jun 25 00:00:00 UTC 2020
Microglia Require CD4 T Cells to Complete the Fetal-to-Adult Transition.
Cell
Pasciuto E,Burton OT,Roca CP,Lagou V,Rajan WD,Theys T,Mancuso R,Tito RY,Kouser L,Callaerts-Vegh Z,de la Fuente AG,Prezzemolo T,Mascali LG,Brajic A,Whyte CE,Yshii L,Martinez-Muriana A,Naughton M,Young A,Moudra A,Lemaitre P,Poovathingal S,Raes J,De Strooper B,Fitzgerald DC,Dooley J,Liston A
17-0691 was used in Flow cytometry/Cell sorting to investigate the function of brain-resident CD4 T cells.
Thu Aug 06 00:00:00 UTC 2020
A Hyper-IgM Syndrome Mutation in Activation-Induced Cytidine Deaminase Disrupts G-Quadruplex Binding and Genome-wide Chromatin Localization.
Immunity
Yewdell WT,Kim Y,Chowdhury P,Lau CM,Smolkin RM,Belcheva KT,Fernandez KC,Cols M,Yen WF,Vaidyanathan B,Angeletti D,McDermott AB,Yewdell JW,Sun JC,Chaudhuri J
17-0691-82 was used in Flow cytometry/Cell sorting to indicate a crucial role for G4 binding in AID targeting and suggest that AID activity may extend beyond Ig loci to regulate the expression of genes relevant to the physiology and pathology of activated B cells.
Tue Nov 17 00:00:00 UTC 2020
Contact-dependent delivery of IL-2 by dendritic cells to CD4 T cells in the contraction phase promotes their long-term survival.
Protein & cell
Tong D,Zhang L,Ning F,Xu Y,Hu X,Shi Y
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Sat Feb 01 00:00:00 UTC 2020
Podocytes present antigen to activate specific T cell immune responses in inflammatory renal disease.
The Journal of pathology
Li S,Liu Y,He Y,Rong W,Zhang M,Li L,Liu Z,Zen K
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Thu Oct 01 00:00:00 UTC 2020
Increase in liver γδ T cells with concurrent augmentation of IFN-β production during the early stages of a mouse model of acute experimental hepatitis B virus infection.
Experimental and therapeutic medicine
Chang L,Wang L,Ling N,Peng H,Chen M
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Wed Jan 01 00:00:00 UTC 2020
Effects of CD4+ T lymphocytes from ovariectomized mice on bone marrow mesenchymal stem cell proliferation and osteogenic differentiation.
Experimental and therapeutic medicine
Shao BY,Wang L,Yu Y,Chen L,Gan N,Huang WM
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Sun Nov 01 00:00:00 UTC 2020
Potentiating the Antitumor Activity of Cytotoxic T Cells via the Transmembrane Domain of IGSF4 That Increases TCR Avidity.
Frontiers in immunology
Kim HR,Park JS,Fatima Y,Kausar M,Park JH,Jun CD
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Tue Jul 27 00:00:00 UTC 2021
Targeting the orphan nuclear receptor NR2F6 in T cells primes tumors for immune checkpoint therapy.
Cell communication and signaling : CCS
Klepsch V,Pommermayr M,Humer D,Brigo N,Hermann-Kleiter N,Baier G
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Tue Jan 14 00:00:00 UTC 2020
SLAMF6 deficiency augments tumor killing and skews toward an effector phenotype revealing it as a novel T cell checkpoint.
eLife
Hajaj E,Eisenberg G,Klein S,Frankenburg S,Merims S,Ben David I,Eisenhaure T,Henrickson SE,Villani AC,Hacohen N,Abudi N,Abramovich R,Cohen JE,Peretz T,Veillette A,Lotem M
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Tue Mar 03 00:00:00 UTC 2020
Loss of testosterone impairs anti-tumor neutrophil function.
Nature communications
Markman JL,Porritt RA,Wakita D,Lane ME,Martinon D,Noval Rivas M,Luu M,Posadas EM,Crother TR,Arditi M
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Tue Mar 31 00:00:00 UTC 2020
Identification of atypical mitogen-activated protein kinase MAPK4 as a novel regulator in acute lung injury.
Cell & bioscience
Mao L,Zhou Y,Chen L,Hu L,Liu S,Zheng W,Zhao J,Guo M,Chen C,He Z,Xu L
17-0691-82 was used in Flow Cytometry to reveal an unknown role of atypical MAPKs member MAPK4 in the pathology of acute lung injury.
Sun Apr 17 00:00:00 UTC 2022
MIR205HG Is a Long Noncoding RNA that Regulates Growth Hormone and Prolactin Production in the Anterior Pituitary.
Developmental cell
Du Q,Hoover AR,Dozmorov I,Raj P,Khan S,Molina E,Chang TC,de la Morena MT,Cleaver OB,Mendell JT,van Oers NSC
17-0691-80 was used in Flow Cytometry to show growth hormone and prolactin production in the anterior pituitary is regulated by the long non-coding RNA MIR205HG.
Mon May 20 00:00:00 UTC 2019
CSF1R antagonism limits local restimulation of antiviral CD8+ T cells during viral encephalitis.
Journal of neuroinflammation
Funk KE,Klein RS
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Thu Jan 31 00:00:00 UTC 2019
TCR and Inflammatory Signals Tune Human MAIT Cells to Exert Specific Tissue Repair and Effector Functions.
Cell reports
Leng T,Akther HD,Hackstein CP,Powell K,King T,Friedrich M,Christoforidou Z,McCuaig S,Neyazi M,Arancibia-Cárcamo CV,Hagel J,Powrie F,Peres RS,Millar V,Ebner D,Lamichhane R,Ussher J,Hinks TSC,Marchi E,Willberg C,Klenerman P
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Tue Sep 17 00:00:00 UTC 2019
Enhanced germinal center reaction by targeting vaccine antigen to major histocompatibility complex class II molecules.
NPJ vaccines
Andersen TK,Huszthy PC,Gopalakrishnan RP,Jacobsen JT,Fauskanger M,Tveita AA,Grødeland G,Bogen B
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Thu Oct 05 00:00:00 UTC 2023
Noc4L-Mediated Ribosome Biogenesis Controls Activation of Regulatory and Conventional T Cells.
Cell reports
Zhu X,Zhang W,Guo J,Zhang X,Li L,Wang T,Yan J,Zhang F,Hou B,Gao N,Gao GF,Zhou X
17-0691 was used in Flow cytometry/Cell sorting to identify the control of Treg activation and immune tolerance maintenance is due to Noc4L mediated ribosome biogenesis.
Tue Apr 23 00:00:00 UTC 2019
Initiation of Antiviral B Cell Immunity Relies on Innate Signals from Spatially Positioned NKT Cells.
Cell
Gaya M,Barral P,Burbage M,Aggarwal S,Montaner B,Warren Navia A,Aid M,Tsui C,Maldonado P,Nair U,Ghneim K,Fallon PG,Sekaly RP,Barouch DH,Shalek AK,Bruckbauer A,Strid J,Batista FD
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Thu Jan 25 00:00:00 UTC 2018
A Pck1-directed glycogen metabolic program regulates formation and maintenance of memory CD8+ T cells.
Nature cell biology
Ma R,Ji T,Zhang H,Dong W,Chen X,Xu P,Chen D,Liang X,Yin X,Liu Y,Ma J,Tang K,Zhang Y,Peng Y,Lu J,Zhang Y,Qin X,Cao X,Wan Y,Huang B
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Mon Jan 01 00:00:00 UTC 2018
Intranasal delivery of adjuvant-free peptide nanofibers elicits resident CD8+ T cell responses.
Journal of controlled release : official journal of the Controlled Release Society
Si Y,Wen Y,Kelly SH,Chong AS,Collier JH
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Sat Jul 28 00:00:00 UTC 2018
MicroRNA-126 deficiency enhanced the activation and function of CD4+ T cells by elevating IRS-1 pathway.
Clinical and experimental immunology
Chu F,Hu Y,Zhou Y,Guo M,Lu J,Zheng W,Xu H,Zhao J,Xu L
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Thu Feb 01 00:00:00 UTC 2018
Oncofetal gene SALL4 reactivation by hepatitis B virus counteracts miR-200c in PD-L1-induced T cell exhaustion.
Nature communications
Sun C,Lan P,Han Q,Huang M,Zhang Z,Xu G,Song J,Wang J,Wei H,Zhang J,Sun R,Zhang C,Tian Z
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Wed Mar 28 00:00:00 UTC 2018
MAIT cells contribute to protection against lethal influenza infection in vivo.
Nature communications
van Wilgenburg B,Loh L,Chen Z,Pediongco TJ,Wang H,Shi M,Zhao Z,Koutsakos M,Nüssing S,Sant S,Wang Z,D'Souza C,Jia X,Almeida CF,Kostenko L,Eckle SBG,Meehan BS,Kallies A,Godfrey DI,Reading PC,Corbett AJ,McCluskey J,Klenerman P,Kedzierska K,Hinks TSC
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Fri Nov 09 00:00:00 UTC 2018
Homoharringtonine induced immune alteration for an Efficient Anti-tumor Response in Mouse Models of Non-small Cell Lung Adenocarcinoma Expressing Kras Mutation.
Scientific reports
Weng TY,Wu HF,Li CY,Hung YH,Chang YW,Chen YL,Hsu HP,Chen YH,Wang CY,Chang JY,Lai MD
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Tue May 29 00:00:00 UTC 2018
Development of a screening strategy for new modulators of T cell receptor signaling and T cell activation.
Scientific reports
Chen EW,Brzostek J,Gascoigne NRJ,Rybakin V
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Tue Jul 03 00:00:00 UTC 2018
A method for manual and automated multiplex RNAscope in situ hybridization and immunocytochemistry on cytospin samples.
PloS one
Chan S,Filézac de L'Etang A,Rangell L,Caplazi P,Lowe JB,Romeo V
17-0691 was used in Flow cytometry/Cell sorting to develop a method for manual and automated multiplex RNAscope in situ hybridisation and immunocytochemistry on cytospin samples.
Mon Apr 22 00:00:00 UTC 2019
The Cyclopeptide Astin C Specifically Inhibits the Innate Immune CDN Sensor STING.
Cell reports
Li S,Hong Z,Wang Z,Li F,Mei J,Huang L,Lou X,Zhao S,Song L,Chen W,Wang Q,Liu H,Cai Y,Yu H,Xu H,Zeng G,Wang Q,Zhu J,Liu X,Tan N,Wang C
17-0691 was used in Flow cytometry/Cell sorting to characterise a STING-specific small-molecular inhibitor that may be applied for potentially manipulating the STING-mediated clinical diseases.
Tue Dec 18 00:00:00 UTC 2018
Adipose tissue B2 cells promote insulin resistance through leukotriene LTB4/LTB4R1 signaling.
The Journal of clinical investigation
Ying W,Wollam J,Ofrecio JM,Bandyopadhyay G,El Ouarrat D,Lee YS,Oh DY,Li P,Osborn O,Olefsky JM
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2017
A type 2 cytokine axis for thymus emigration.
The Journal of experimental medicine
White AJ,Baik S,Parnell SM,Holland AM,Brombacher F,Jenkinson WE,Anderson G
17-0691 was used in Flow cytometry/Cell sorting to show the thymic microenvironment expresses the type 2 IL-4R complex and is functionally responsive to its known ligands, IL-4 and IL-13.
Mon Aug 07 00:00:00 UTC 2017
IL-36 Signaling Facilitates Activation of the NLRP3 Inflammasome and IL-23/IL-17 Axis in Renal Inflammation and Fibrosis.
Journal of the American Society of Nephrology : JASN
Chi HH,Hua KF,Lin YC,Chu CL,Hsieh CY,Hsu YJ,Ka SM,Tsai YL,Liu FC,Chen A
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Sat Jul 01 00:00:00 UTC 2017
FHL2 Regulates Natural Killer Cell Development and Activation during Streptococcus pneumoniae Infection.
Frontiers in immunology
Baranek T,Morello E,Valayer A,Aimar RF,Bréa D,Henry C,Besnard AG,Dalloneau E,Guillon A,Dequin PF,Narni-Mancinelli E,Vivier E,Laurent F,Wei Y,Paget C,Si-Tahar M
17-0691 was used in Flow cytometry/Cell sorting to determine the expression and role of FHL2 in natural killer cell biology.
Wed Nov 20 00:00:00 UTC 2019
Betulin from Hedyotis hedyotidea ameliorates concanavalin A-induced and T cell-mediated autoimmune hepatitis in mice.
Acta pharmacologica Sinica
Zhou YQ,Weng XF,Dou R,Tan XS,Zhang TT,Fang JB,Wu XW
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Wed Feb 01 00:00:00 UTC 2017
14-3-3 Proteins regulate K2P 5.1 surface expression on T lymphocytes.
Traffic (Copenhagen, Denmark)
Fernández-Orth J,Ehling P,Ruck T,Pankratz S,Hofmann MS,Landgraf P,Dieterich DC,Smalla KH,Kähne T,Seebohm G,Budde T,Wiendl H,Bittner S,Meuth SG
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Sun Jan 01 00:00:00 UTC 2017
Selectivity of Human TLR9 for Double CpG Motifs and Implications for the Recognition of Genomic DNA.
Journal of immunology (Baltimore, Md. : 1950)
Pohar J,Yamamoto C,Fukui R,Cajnko MM,Miyake K,Jerala R,Benčina M
17-0691 was used in Flow cytometry/Cell sorting to study the selectivity of human TLR9 for double CpG motifs.
Wed Mar 01 00:00:00 UTC 2017
IRF6 Regulates Alternative Activation by Suppressing PPARγ in Male Murine Macrophages.
Endocrinology
Li C,Ying W,Huang Z,Brehm T,Morin A,Vella AT,Zhou B
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Fri Sep 01 00:00:00 UTC 2017
Tristetraprolin inhibits macrophage IL-27-induced activation of antitumour cytotoxic T cell responses.
Nature communications
Wang Q,Ning H,Peng H,Wei L,Hou R,Hoft DF,Liu J
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Wed Oct 11 00:00:00 UTC 2017
Serum Gp96 is a chaperone of complement-C3 during graft-versus-host disease.
JCI insight
Seignez A,Joly AL,Chaumonnot K,Hazoumé A,Sanka M,Marcion G,Boudesco C,Hammann A,Seigneuric R,Jégo G,Ducoroy P,Delarue P,Senet P,Castilla-Llorente C,Solary E,Durey MA,Rubio MT,Hermine O,Kohli E,Garrido C
17-0691 was used in Flow cytometry/Cell sorting to identify glycoprotein 96 and complement C3 as partner graft-versus-host disease-induced tissue damage-associated molecular pattern molecules.
Thu Mar 23 00:00:00 UTC 2017
The efficacy of oral and subcutaneous antigen-specific immunotherapy in murine cow's milk- and peanut allergy models.
Clinical and translational allergy
Vonk MM,Wagenaar L,Pieters RHH,Knippels LMJ,Willemsen LEM,Smit JJ,van Esch BCAM,Garssen J
17-0691 was used in Flow cytometry/Cell sorting to compare oral immunotherapy and subcutaneous immunotherapy models for cows milk and peanut allergy.
Fri Apr 08 00:00:00 UTC 2022
MyD88-dependent pro-interleukin-1β induction in dendritic cells exposed to food-grade synthetic amorphous silica.
Particle and fibre toxicology
Winkler HC,Kornprobst J,Wick P,von Moos LM,Trantakis I,Schraner EM,Bathke B,Hochrein H,Suter M,Naegeli H
Fri Jun 23 00:00:00 UTC 2017
Regulatory T cell frequencies and phenotypes following anti-viral vaccination.
PloS one
de Wolf ACMT,van Aalst S,Ludwig IS,Bodinham CL,Lewis DJ,van der Zee R,van Eden W,Broere F
17-0691 was used in Flow cytometry/Cell sorting to determine the impact of anti-viral vaccination on Treg homeostasis.
Fri Sep 22 00:00:00 UTC 2017
MicroRNA-7 Deficiency Ameliorates the Pathologies of Acute Lung Injury through Elevating KLF4.
Frontiers in immunology
Zhao J,Chen C,Guo M,Tao Y,Cui P,Zhou Y,Qin N,Zheng J,Zhang J,Xu L
17-0691 was used in Flow cytometry/Cell sorting to study the role of microRNA-7 in acute lung injury, allowing the development of new therapeutic strategies against inflammatory lung disease.
Wed Mar 16 00:00:00 UTC 2022
Resident T Cells Are Unable To Control Herpes Simplex Virus-1 Activity in the Brain Ependymal Region during Latency.
Journal of immunology (Baltimore, Md. : 1950)
Menendez CM,Jinkins JK,Carr DJ
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Mon Aug 15 00:00:00 UTC 2016
Affinity and dose of TCR engagement yield proportional enhancer and gene activity in CD4+ T cells.
eLife
Allison KA,Sajti E,Collier JG,Gosselin D,Troutman TD,Stone EL,Hedrick SM,Glass CK
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Mon Jul 04 00:00:00 UTC 2016
The HSP90 inhibitor, 17AAG, protects the intestinal stem cell niche and inhibits graft versus host disease development.
Oncogene
Joly AL,Deepti A,Seignez A,Goloudina A,Hebrard S,Schmitt E,Richaud S,Fourmaux E,Hammann A,Collura A,Svrcek M,Jego G,Robinet E,Solary E,Demidov O,Kohli E,Garrido C
17-0691-82 was used in Flow cytometry/Cell sorting to investigate HSP90 inhibitors in graft versus host disease development in intestinal stem cell niches.
Thu Jun 02 00:00:00 UTC 2016
Alternative splicing of MALT1 controls signalling and activation of CD4(+) T cells.
Nature communications
Meininger I,Griesbach RA,Hu D,Gehring T,Seeholzer T,Bertossi A,Kranich J,Oeckinghaus A,Eitelhuber AC,Greczmiel U,Gewies A,Schmidt-Supprian M,Ruland J,Brocker T,Heissmeyer V,Heyd F,Krappmann D
17-0691 was used in Flow cytometry/Cell sorting to study how TCR-induced alternative splicing augments MALT1 scaffolding to enhance downstream signalling and to promote optimal T-cell activation.
Tue Apr 12 00:00:00 UTC 2016
MicroRNA-223 is a crucial mediator of PPARγ-regulated alternative macrophage activation.
The Journal of clinical investigation
Ying W,Tseng A,Chang RC,Morin A,Brehm T,Triff K,Nair V,Zhuang G,Song H,Kanameni S,Wang H,Golding MC,Bazer FW,Chapkin RS,Safe S,Zhou B
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Mon Nov 02 00:00:00 UTC 2015
Single-Dose CpG Immunization Protects Against a Heterosubtypic Challenge and Generates Antigen-Specific Memory T Cells.
Frontiers in immunology
Vogel AJ,Brown DM
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Fri Jul 10 00:00:00 UTC 2015
MFG-E8 Is Critical for Embryonic Stem Cell-Mediated T Cell Immunomodulation.
Stem cell reports
Tan Y,AlKhamees B,Jia D,Li L,Couture JF,Figeys D,Jinushi M,Wang L
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Tue Nov 10 00:00:00 UTC 2015
Sunitinib depletes myeloid-derived suppressor cells and synergizes with a cancer vaccine to enhance antigen-specific immune responses and tumor eradication.
Oncoimmunology
Draghiciu O,Nijman HW,Hoogeboom BN,Meijerhof T,Daemen T
17-0691 was used in Flow cytometry/Cell sorting to demonstrate that sunitinib can create a favourable microenvironment depleted of MDSCs, and act synergistically with a cancer vaccine, resulting in enhanced levels of active tumor-antigen specific CTLs.
Sun Mar 01 00:00:00 UTC 2015
Immunoglobulin E plays an immunoregulatory role in lupus.
The Journal of experimental medicine
Dema B,Charles N,Pellefigues C,Ricks TK,Suzuki R,Jiang C,Scheffel J,Hasni S,Hoffman V,Jablonski M,Sacré K,Gobert D,Papo T,Daugas E,Crampton S,Bolland S,Rivera J
17-0691 was used in Flow cytometry/Cell sorting to explore the effect of IgE deficiency on the inflammatory response in FcγRIIB-deficient mice, which hastens and results in a more aggressive lupuslike disease.
Mon Oct 20 00:00:00 UTC 2014
Peptidylarginine deiminase 4 contributes to tumor necrosis factor α-induced inflammatory arthritis.
Arthritis & rheumatology (Hoboken, N.J.)
Shelef MA,Sokolove J,Lahey LJ,Wagner CA,Sackmann EK,Warner TF,Wang Y,Beebe DJ,Robinson WH,Huttenlocher A
17-0691 was used in Flow cytometry/Cell sorting to investigate whether TNFα and PAD4 may synergistically exacerbate autoantibody production and inflammatory arthritis, showing that PAD4 contributes to TNFα-induced inflammatory arthritis.
Sun Jun 01 00:00:00 UTC 2014
Mechanisms of the immunosuppressive effects of mouse adipose tissue-derived mesenchymal stromal cells on mouse alloreactively stimulated spleen cells.
Experimental and therapeutic medicine
Nagaya R,Mizuno-Kamiya M,Takayama E,Kawaki H,Onoe I,Tanabe T,Nagahara K,Kondoh N
17-0691 was used in Flow cytometry/Cell sorting to demonstrate that adipose tissue-derived mesenchymal stromal cells have significant immune modulatory effects on alloreactively stimulated spleen cells.
Wed Jan 01 00:00:00 UTC 2014
Tim-3 negatively mediates natural killer cell function in LPS-induced endotoxic shock.
PloS one
Hou H,Liu W,Wu S,Lu Y,Peng J,Zhu Y,Lu Y,Wang F,Sun Z
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Mon Jul 06 00:00:00 UTC 2015
Establishment of a murine graft-versus-myeloma model using allogeneic stem cell transplantation.
PloS one
Binsfeld M,Beguin Y,Belle L,Otjacques E,Hannon M,Briquet A,Heusschen R,Drion P,Zilberberg J,Bogen B,Baron F,Caers J
17-0691 was used in Flow cytometry/Cell sorting to investigate the anti-myeloma effects of allo-SCT from B10.D2 mice into MHC-matched myeloma-bearing Balb/cJ mice.
Tue Dec 15 00:00:00 UTC 2015
The thymic medulla is required for Foxp3+ regulatory but not conventional CD4+ thymocyte development.
The Journal of experimental medicine
Cowan JE,Parnell SM,Nakamura K,Caamano JH,Lane PJ,Jenkinson EJ,Jenkinson WE,Anderson G
17-0691 was used in Flow cytometry/Cell sorting to demonstrate a differential requirement for the thymic medulla in relation to CD4 conventional and Foxp3(+) thymocyte lineages.
Mon Apr 08 00:00:00 UTC 2013
Alterations in regulatory T cells induced by specific oligosaccharides improve vaccine responsiveness in mice.
PloS one
Schijf MA,Kerperien J,Bastiaans J,Szklany K,Meerding J,Hofman G,Boon L,van Wijk F,Garssen J,Van't Land B
17-0691 was used in Flow cytometry/Cell sorting to identify factors affecting the quality of the immune response to vaccination.
Tue May 27 00:00:00 UTC 2014
Angiotensin II is a new component involved in splenic T lymphocyte responses during Plasmodium berghei ANKA infection.
PloS one
Silva-Filho JL,Souza MC,Ferreira-Dasilva CT,Silva LS,Costa MF,Padua TA,Henriques Md,Morrot A,Savino W,Caruso-Neves C,Pinheiro AA
17-0691 was used in Flow cytometry/Cell sorting to provide evidence for the potential role of Ang II in regulating the splenic immune response.
Tue Nov 26 00:00:00 UTC 2013
Microparticles bearing encephalitogenic peptides induce T-cell tolerance and ameliorate experimental autoimmune encephalomyelitis.
Nature biotechnology
Getts DR,Martin AJ,McCarthy DP,Terry RL,Hunter ZN,Yap WT,Getts MT,Pleiss M,Luo X,King NJ,Shea LD,Miller SD
17-0691 was used in Flow cytometry/Cell sorting to highlight the potential for using microparticles to target natural apoptotic clearance pathways to inactivate pathogenic T cells in autoimmunity.
Sat Dec 01 00:00:00 UTC 2012
Dynamic in situ cytometry uncovers T cell receptor signaling during immunological synapses and kinapses in vivo.
Immunity
Moreau HD,Lemaître F,Terriac E,Azar G,Piel M,Lennon-Dumenil AM,Bousso P
17-0691 was used in Flow cytometry/Cell sorting to examine how T cells sense TCR ligands of varying affinities in vivo.
Fri Aug 24 00:00:00 UTC 2012
Anti-HIV B Cell lines as candidate vaccine biosensors.
Journal of immunology (Baltimore, Md. : 1950)
Ota T,Doyle-Cooper C,Cooper AB,Huber M,Falkowska E,Doores KJ,Hangartner L,Le K,Sok D,Jardine J,Lifson J,Wu X,Mascola JR,Poignard P,Binley JM,Chakrabarti BK,Schief WR,Wyatt RT,Burton DR,Nemazee D
17-0691 was used in Flow cytometry/Cell sorting to suggest that HIV Env antigen-expressing, infection-competent virions are poorly recognized by high affinity bNAb-expressing cells.
Thu Nov 15 00:00:00 UTC 2012
Commitment to the regulatory T cell lineage requires CARMA1 in the thymus but not in the periphery.
PLoS biology
Barnes MJ,Krebs P,Harris N,Eidenschenk C,Gonzalez-Quintial R,Arnold CN,Crozat K,Sovath S,Moresco EM,Theofilopoulos AN,Beutler B,Hoebe K
17-0691 was used in Flow cytometry/Cell sorting to investigate methods to distinguish between Tregs of thymic or peripheral origin, showing that commitment requires CARMA1 in the thymus but not in the periphery.
Tue Mar 03 00:00:00 UTC 2009
Immunoregulatory role of Jalpha281 T cells in aged mice developing lupus-like nephritis.
European journal of immunology
Sireci G,Russo D,Dieli F,Porcelli SA,Taniguchi M,La Manna MP,Di Liberto D,Scarpa F,Salerno A
17-0691 was used in Flow cytometry/Cell sorting to study renal functions and autoantibody production in aged Ja281 KO mice lacking iNKT cells.
Thu Feb 01 00:00:00 UTC 2007
Modulation of naive CD4+ T-cell responses to an airway antigen during pulmonary mycobacterial infection.
Infection and immunity
Anis MM,Fulton SA,Reba SM,Harding CV,Boom WH
Published figure using CD69 monoclonal antibody (Product # 17-0691-82) in Flow Cytometry
Tue May 01 00:00:00 UTC 2007