Product References
Exosomal CD73 from serum of patients with melanoma suppresses lymphocyte functions and is associated with therapy resistance to anti-PD-1 agents.
Journal for immunotherapy of cancer
Turiello R,Capone M,Morretta E,Monti MC,Madonna G,Azzaro R,Del Gaudio P,Simeone E,Sorrentino A,Ascierto PA,Morello S
11-0739-42 was used in Flow cytometry/Cell sorting to evaluate levels of exosomal CD73 before and early during treatment with anti-PD-1 agents in patients with melanoma and its potential contribution to affect T-cell functions and to influence the clinical outcomes of anti-PD-1 monotherapy.
Tue Mar 01 00:00:00 EST 2022
Exosomal CD73 from serum of patients with melanoma suppresses lymphocyte functions and is associated with therapy resistance to anti-PD-1 agents.
Journal for immunotherapy of cancer
Turiello R,Capone M,Morretta E,Monti MC,Madonna G,Azzaro R,Del Gaudio P,Simeone E,Sorrentino A,Ascierto PA,Morello S
11-0739-42 was used in Flow cytometry/Cell sorting to evaluate levels of exosomal CD73 before and early during treatment with anti-PD-1 agents in patients with melanoma and its potential contribution to affect T-cell functions and to influence the clinical outcomes of anti-PD-1 monotherapy.
Tue Mar 01 00:00:00 EST 2022
Induced Neurodifferentiation of hBM-MSCs through Activation of the ERK/CREB Pathway via Pulsed Electromagnetic Fields and Physical Stimulation Promotes Neurogenesis in Cerebral Ischemic Models.
International journal of molecular sciences
Park HJ,Choi JH,Nam MH,Seo YK
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Fri Jan 21 00:00:00 EST 2022
Reprogramming bone progenitor identity and potency through control of collagen density and oxygen tension.
iScience
Al Hosni R,Bozec L,Roberts SJ,Cheema U
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Fri Apr 15 00:00:00 EDT 2022
Reprogramming bone progenitor identity and potency through control of collagen density and oxygen tension.
iScience
Al Hosni R,Bozec L,Roberts SJ,Cheema U
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Fri Apr 15 00:00:00 EDT 2022
Human placental mesenchymal stem cells regulate inflammation via the NF‑κB signaling pathway.
Experimental and therapeutic medicine
Liu Y,Zhang X,Hu Y,Kang M,Wu Y,Wang Y,Deng C
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2022
hUCMSCs carrying exenatide prevent T1DM by improving intestinal microflora composition and islet tissue damage repair.
Molecular medicine (Cambridge, Mass.)
Wang W,Wang Y,Chi J,Tan X,Hu J,Ma X,Sun X,Che K,Lv W,Wang Y
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Tue Dec 13 00:00:00 EST 2022
VAP-PLGA microspheres (VAP-PLGA) promote adipose-derived stem cells (ADSCs)-induced wound healing in chronic skin ulcers in mice via PI3K/Akt/HIF-1α pathway.
Bioengineered
Jiang W,Zhang J,Zhang X,Fan C,Huang J
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Wed Dec 01 00:00:00 EST 2021
Peptide 11R‑VIVIT promotes fracture healing in osteoporotic rats.
International journal of molecular medicine
Hou C,Wang X,Jiang W,Bian Z,Zhu L,Li M
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Sun Aug 01 00:00:00 EDT 2021
Peptide 11R‑VIVIT promotes fracture healing in osteoporotic rats.
International journal of molecular medicine
Hou C,Wang X,Jiang W,Bian Z,Zhu L,Li M
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Sun Aug 01 00:00:00 EDT 2021
Bcl-xL mutant promotes cartilage differentiation of BMSCs by upregulating TGF-β/BMP expression levels.
Experimental and therapeutic medicine
Xiao K,Yang L,Xie W,Gao X,Huang R,Xie M
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Thu Jul 01 00:00:00 EDT 2021
TRPA1 triggers hyperalgesia and inflammation after tooth bleaching.
Scientific reports
Chen C,Huang X,Zhu W,Ding C,Huang P,Li R
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Tue Aug 31 00:00:00 EDT 2021
In Vitro Anti-cancer Activity of Adipose-Derived Mesenchymal Stem Cells Increased after Infection with Oncolytic Reovirus.
Advanced pharmaceutical bulletin
Babaei A,Bannazadeh Baghi H,Nezhadi A,Jamalpoor Z
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
In vitro differentiation of human embryonic stem cells to hemogenic endothelium and blood progenitors via embryoid body formation.
STAR protocols
Garcia-Alegria E,Potts B,Menegatti S,Kouskoff V
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Fri Mar 19 00:00:00 EDT 2021
Therapeutic potential of human umbilical cord mesenchymal stem cells on aortic atherosclerotic plaque in a high-fat diet rabbit model.
Stem cell research & therapy
Li Y,Shi G,Han Y,Shang H,Li H,Liang W,Zhao W,Bai L,Qin C
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Thu Jul 15 00:00:00 EDT 2021
Glycemic control by umbilical cord-derived mesenchymal stem cells promotes effects of fasting-mimicking diet on type 2 diabetic mice.
Stem cell research & therapy
Zhao N,Gao YF,Bao L,Lei J,An HX,Pu FX,Cheng RP,Chen J,Ni H,Sui BD,Ji FP,Hu CH
11-0739-42 was used in Flow Cytometry to establish a strategy that combined UC-MSC infusion and FMD and was effective in treating T2D, which provided potential approaches for developing novel clinical T2D therapies.
Tue Jul 13 00:00:00 EDT 2021
Down-Regulated Exosomal MicroRNA-221 - 3p Derived From Senescent Mesenchymal Stem Cells Impairs Heart Repair.
Frontiers in cell and developmental biology
Sun L,Zhu W,Zhao P,Zhang J,Lu Y,Zhu Y,Zhao W,Liu Y,Chen Q,Zhang F
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Mon Sep 28 00:00:00 EDT 2020
Differentiation Potential of Early- and Late-Passage Adipose-Derived Mesenchymal Stem Cells Cultured under Hypoxia and Normoxia.
Stem cells international
Zhao AG,Shah K,Freitag J,Cromer B,Sumer H
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Tue Oct 06 00:00:00 EDT 2020
Vasculogenesis from Human Dental Pulp Stem Cells Grown in Matrigel with Fully Defined Serum-Free Culture Media.
Biomedicines
Luzuriaga J,Irurzun J,Irastorza I,Unda F,Ibarretxe G,Pineda JR
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Mon Nov 09 00:00:00 EST 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Fri Apr 03 00:00:00 EDT 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Fri Apr 03 00:00:00 EDT 2020
IGF-1 enhances BMSC viability, migration, and anti-apoptosis in myocardial infarction via secreted frizzled-related protein 2 pathway.
Stem cell research & therapy
Lin M,Liu X,Zheng H,Huang X,Wu Y,Huang A,Zhu H,Hu Y,Mai W,Huang Y
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Thu Jan 09 00:00:00 EST 2020
IGF-1 enhances BMSC viability, migration, and anti-apoptosis in myocardial infarction via secreted frizzled-related protein 2 pathway.
Stem cell research & therapy
Lin M,Liu X,Zheng H,Huang X,Wu Y,Huang A,Zhu H,Hu Y,Mai W,Huang Y
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Thu Jan 09 00:00:00 EST 2020
Chondrogenic Differentiation from Induced Pluripotent Stem Cells Using Non-Viral Minicircle Vectors.
Cells
Rim YA,Nam Y,Park N,Jung H,Lee K,Lee J,Ju JH
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2020
Chondrogenic Differentiation from Induced Pluripotent Stem Cells Using Non-Viral Minicircle Vectors.
Cells
Rim YA,Nam Y,Park N,Jung H,Lee K,Lee J,Ju JH
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Sun Mar 01 00:00:00 EST 2020
Prospective isolation of human fibroadipogenic progenitors with CD73.
Heliyon
Goloviznina NA,Xie N,Dandapat A,Iaizzo PA,Kyba M
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2020
Circulating Exosomes Inhibit B Cell Proliferation and Activity.
Cancers
Schroeder JC,Puntigam L,Hofmann L,Jeske SS,Beccard IJ,Doescher J,Laban S,Hoffmann TK,Brunner C,Theodoraki MN,Schuler PJ
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Wed Jul 29 00:00:00 EDT 2020
Circulating Exosomes Inhibit B Cell Proliferation and Activity.
Cancers
Schroeder JC,Puntigam L,Hofmann L,Jeske SS,Beccard IJ,Doescher J,Laban S,Hoffmann TK,Brunner C,Theodoraki MN,Schuler PJ
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Wed Jul 29 00:00:00 EDT 2020
Impact of Fibronectin Knockout on Proliferation and Differentiation of Human Infrapatellar Fat Pad-Derived Stem Cells.
Frontiers in bioengineering and biotechnology
Wang Y,Fu Y,Yan Z,Zhang XB,Pei M
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Wed Dec 14 00:00:00 EST 2022
Modulation of proliferation and differentiation of gingiva‑derived mesenchymal stem cells by concentrated growth factors: Potential implications in tissue engineering for dental regeneration and repair.
International journal of molecular medicine
Chen X,Chen Y,Hou Y,Song P,Zhou M,Nie M,Liu X
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2019
Modulation of proliferation and differentiation of gingiva‑derived mesenchymal stem cells by concentrated growth factors: Potential implications in tissue engineering for dental regeneration and repair.
International journal of molecular medicine
Chen X,Chen Y,Hou Y,Song P,Zhou M,Nie M,Liu X
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2019
Therapeutic Effects of Human Urine-Derived Stem Cells in a Rat Model of Cisplatin-Induced Acute Kidney Injury In Vivo and In Vitro.
Stem cells international
Sun B,Luo X,Yang C,Liu P,Yang Y,Dong X,Yang Z,Xu J,Zhang Y,Li L
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Thu May 13 00:00:00 EDT 2021
Sodium Selenite Improves The Therapeutic Effect Of BMSCs Via Promoting The Proliferation And Differentiation, Thereby Promoting The Hematopoietic Factors.
OncoTargets and therapy
Yan D,Tang B,Yan L,Zhang L,Miao M,Chen X,Sui G,Zhang Q,Liu D,Wang H
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Tue Apr 12 00:00:00 EDT 2022
High-yield isolation of menstrual blood-derived endometrial stem cells by direct red blood cell lysis treatment.
Biology open
Sun Y,Ren Y,Yang F,He Y,Liang S,Guan L,Cheng F,Liu Y,Lin J
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Thu May 02 00:00:00 EDT 2019
High-yield isolation of menstrual blood-derived endometrial stem cells by direct red blood cell lysis treatment.
Biology open
Sun Y,Ren Y,Yang F,He Y,Liang S,Guan L,Cheng F,Liu Y,Lin J
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Thu May 02 00:00:00 EDT 2019
Better therapeutic potential of bone marrow-derived mesenchymal stem cells compared with chorionic villi-derived mesenchymal stem cells in airway injury model.
Regenerative medicine
Ji S,Wu C,Tong L,Wang L,Zhou J,Chen C,Song Y
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2019
Isolation and characterization of stem cells from differentially degenerated human lumbar zygapophyseal articular cartilage.
Molecular medicine reports
Xiao L,Xu S,Wang X,Jin Z,Wang J,Yang B,Xu H
11-0739 was used in Flow cytometry/Cell sorting to show stem cells with multilineage differentiation potential and clonal properties were identified in human LZAC, and these characteristics were more prominent in mildly degenerated as compared with severely degenerated articular cartilage.
Sat Dec 01 00:00:00 EST 2018
BMP14 induces tenogenic differentiation of bone marrow mesenchymal stem cells in vitro.
Experimental and therapeutic medicine
Wang D,Jiang X,Lu A,Tu M,Huang W,Huang P
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2018
Human umbilical cord-derived mesenchymal stem cells ameliorate the enteropathy of food allergies in mice.
Experimental and therapeutic medicine
Yan N,Xu J,Zhao C,Wu Y,Gao F,Li C,Zhou W,Xiao T,Zhou X,Shao Q,Xia S
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Sat Dec 01 00:00:00 EST 2018
BNIP3/Bcl-2-mediated apoptosis induced by cyclic tensile stretch in human cartilage endplate-derived stem cells.
Experimental and therapeutic medicine
Yuan C,Pu L,He Z,Wang J
11-0739-42 was used in Flow Cytometry to investigate the molecular mechanisms of cyclic stretch-induced apoptosis in human intervertebral disc cartilage endplate-derived stem cells (CESCs).
Mon Jan 01 00:00:00 EST 2018
The influence of chemotherapy on adenosine-producing B cells in patients with head and neck squamous cell carcinoma.
Oncotarget
Ziebart A,Huber U,Jeske S,Laban S,Doescher J,Hoffmann TK,Brunner C,Jackson EK,Schuler PJ
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Fri Jan 19 00:00:00 EST 2018
A high-yield isolation and enrichment strategy for human lung microvascular endothelial cells.
Pulmonary circulation
Gaskill C,Majka SM
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2017
Human umbilical cord mesenchymal stem cells improve the reserve function of perimenopausal ovary via a paracrine mechanism.
Stem cell research & therapy
Li J,Mao Q,He J,She H,Zhang Z,Yin C
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Thu Mar 09 00:00:00 EST 2017
Etanercept-Synthesising Mesenchymal Stem Cells Efficiently Ameliorate Collagen-Induced Arthritis.
Scientific reports
Park N,Rim YA,Jung H,Kim J,Yi H,Kim Y,Jang Y,Jung SM,Lee J,Kwok SK,Park SH,Ju JH
11-0739 was used in Flow cytometry/Cell sorting to develop a novel method to produce synthetic biological drugs from engineered MSCs, and investigate the anti-arthritic effect of these cells in a collagen-induced arthritis mouse model.
Fri Jan 13 00:00:00 EST 2017
Feasibility of placenta-derived mesenchymal stem cells as a tool for studying pregnancy-related disorders.
Scientific reports
Fuchi N,Miura K,Doi H,Li TS,Masuzaki H
11-0739 was used in Flow cytometry/Cell sorting to evaluate the feasibility of using placenta-derived mesenchymal stem cells as a tool to study pregnancy related disorders.
Wed Apr 12 00:00:00 EDT 2017
A Member of the Nuclear Receptor Superfamily, Designated as NR2F2, Supports the Self-Renewal Capacity and Pluripotency of Human Bone Marrow-Derived Mesenchymal Stem Cells.
Stem cells international
Zhu N,Wang H,Wang B,Wei J,Shan W,Feng J,Huang H
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Tue Jan 19 00:00:00 EST 2016
MIF Plays a Key Role in Regulating Tissue-Specific Chondro-Osteogenic Differentiation Fate of Human Cartilage Endplate Stem Cells under Hypoxia.
Stem cell reports
Yao Y,Deng Q,Song W,Zhang H,Li Y,Yang Y,Fan X,Liu M,Shang J,Sun C,Tang Y,Jin X,Liu H,Huang B,Zhou Y
11-0739 was used in Flow cytometry/Cell sorting to show MIF plays a role in regulating the differentiation fate of human cartilage endplate stem cells under hypoxia.
Tue Aug 09 00:00:00 EDT 2016
CRISPR/Cas9-based genetic correction for recessive dystrophic epidermolysis bullosa.
NPJ Regenerative medicine
Webber BR,Osborn MJ,McElroy AN,Twaroski K,Lonetree CL,DeFeo AP,Xia L,Eide C,Lees CJ,McElmurry RT,Riddle MJ,Kim CJ,Patel DD,Blazar BR,Tolar J
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Wed Sep 30 00:00:00 EDT 2020
Potency of umbilical cord blood- and Wharton's jelly-derived mesenchymal stem cells for scarless wound healing.
Scientific reports
Doi H,Kitajima Y,Luo L,Yan C,Tateishi S,Ono Y,Urata Y,Goto S,Mori R,Masuzaki H,Shimokawa I,Hirano A,Li TS
11-0739 was used in Flow cytometry/Cell sorting to determine that umbilical cord blood offers a potential source of MSCs for therapeutic scarless wound healing.
Tue Jan 05 00:00:00 EST 2016
Focal Adhesion Kinase Signaling Mediated the Enhancement of Osteogenesis of Human Mesenchymal Stem Cells Induced by Extracorporeal Shockwave.
Scientific reports
Hu J,Liao H,Ma Z,Chen H,Huang Z,Zhang Y,Yu M,Chen Y,Xu J
11-0739 was used in Flow cytometry/Cell sorting to investigate the underlying mechanisms of osteogenic effects of bone marrow mesenchymal stem cells induced by extracorporeal shockwave.
Thu Feb 11 00:00:00 EST 2016
Human adipose-derived stem cells partially rescue the stroke syndromes by promoting spatial learning and memory in mouse middle cerebral artery occlusion model.
Stem cell research & therapy
Zhou F,Gao S,Wang L,Sun C,Chen L,Yuan P,Zhao H,Yi Y,Qin Y,Dong Z,Cao L,Ren H,Zhu L,Li Q,Lu B,Liang A,Xu GT,Zhu H,Gao Z,Ma J,Xu J,Chen X
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Sat May 09 00:00:00 EDT 2015
Human Gingival Integration-Free iPSCs; a Source for MSC-Like Cells.
International journal of molecular sciences
Umezaki Y,Hashimoto Y,Nishishita N,Kawamata S,Baba S
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Mon Jun 15 00:00:00 EDT 2015
Comparison of human adipose-derived stem cells isolated from subcutaneous, omental, and intrathoracic adipose tissue depots for regenerative applications.
Stem cells translational medicine
Russo V,Yu C,Belliveau P,Hamilton A,Flynn LE
11-0739 was used in Flow cytometry/Cell sorting to determine that depot selection is an important factor to consider when applying ASCs in tissue-specific cell-based regenerative therapies.
Sat Feb 01 00:00:00 EST 2014
Epithelial cell differentiation of human mesenchymal stromal cells in decellularized lung scaffolds.
Tissue engineering. Part A
Mendez JJ,Ghaedi M,Steinbacher D,Niklason LE
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Sun Jun 01 00:00:00 EDT 2014
Hypoxia-cultured human adipose-derived mesenchymal stem cells are non-oncogenic and have enhanced viability, motility, and tropism to brain cancer.
Cell death & disease
Feng Y,Zhu M,Dangelmajer S,Lee YM,Wijesekera O,Castellanos CX,Denduluri A,Chaichana KL,Li Q,Zhang H,Levchenko A,Guerrero-Cazares H,Quiñones-Hinojosa A
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Thu Dec 11 00:00:00 EST 2014
Tumor-derived exosomes promote tumor progression and T-cell dysfunction through the regulation of enriched exosomal microRNAs in human nasopharyngeal carcinoma.
Oncotarget
Ye SB,Li ZL,Luo DH,Huang BJ,Chen YS,Zhang XS,Cui J,Zeng YX,Li J
11-0739 was used in Flow cytometry/Cell sorting to show that tumour-derived exosomes promote tumour progression and T-cell dysfunction through regulation of exosomal microRNAs in human carcinomas.
Wed Jul 30 00:00:00 EDT 2014
Microvesicles derived from human umbilical cord Wharton's jelly mesenchymal stem cells attenuate bladder tumor cell growth in vitro and in vivo.
PloS one
Wu S,Ju GQ,Du T,Zhu YJ,Liu GH
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Tue Oct 22 00:00:00 EDT 2013
Myoblasts derived from normal hESCs and dystrophic hiPSCs efficiently fuse with existing muscle fibers following transplantation.
Molecular therapy : the journal of the American Society of Gene Therapy
Goudenege S,Lebel C,Huot NB,Dufour C,Fujii I,Gekas J,Rousseau J,Tremblay JP
Published figure using CD73 monoclonal antibody (Product # 11-0739-42) in Flow Cytometry
Thu Nov 01 00:00:00 EDT 2012
A novel mechanism involved in the pathogenesis of Graves ophthalmopathy (GO): clathrin is a possible targeting molecule for inhibiting local immune response in the orbit.
The Journal of clinical endocrinology and metabolism
Meyer zu Hörste M,Ströher E,Berchner-Pfannschmidt U,Schmitz-Spanke S,Pink M,Göthert JR,Fischer JW,Gulbins E,Eckstein AK
11-0739 was used in Flow cytometry/Cell sorting to analyse and inhibit signalling pathways in resident orbital fibroblasts that are involved in Graves opthalmopathy.
Tue Nov 01 00:00:00 EDT 2011