Product References
Loss of C3a and C5a receptors promotes adipocyte browning and attenuates diet-induced obesity via activating inosine/A2aR pathway.
Cell reports
Kong LR,Chen XH,Sun Q,Zhang KY,Xu L,Ding L,Zhou YP,Zhang ZB,Lin JR,Gao PJ
25-0731-82 was used in Flow cytometry/Cell sorting to identify C3aR and C5aR as critical switches to control adipocyte browning and energy balance in male mice, via inosine/A2aR pathway.
Tue Feb 28 00:00:00 EST 2023
Antigen- and scaffold-specific antibody responses to protein nanoparticle immunogens.
Cell reports. Medicine
Kraft JC,Pham MN,Shehata L,Brinkkemper M,Boyoglu-Barnum S,Sprouse KR,Walls AC,Cheng S,Murphy M,Pettie D,Ahlrichs M,Sydeman C,Johnson M,Blackstone A,Ellis D,Ravichandran R,Fiala B,Wrenn S,Miranda M,Sliepen K,Brouwer PJM,Antanasijevic A,Veesler D,Ward AB,Kanekiyo M,Pepper M,Sanders RW,King NP
25-0731-82 was used in Flow cytometry/Cell sorting to define relationships between anti-scaffold and antigen-specific antibody responses elicited by protein nanoparticle immunogens.
Tue Oct 18 00:00:00 EDT 2022
Age-related trabecular bone loss is associated with a decline in serum Galectin-1 level.
BMC musculoskeletal disorders
Xu W,Ni C,Wang Y,Zheng G,Zhang J,Xu Y
Published figure using CD73 monoclonal antibody (Product # 25-0731-82) in Flow Cytometry
Tue Apr 27 00:00:00 EDT 2021
Transplantation of bone marrow-derived mesenchymal stem cells with silencing of microRNA-138 relieves pelvic organ prolapse through the FBLN5/IL-1β/elastin pathway.
Aging
Zhao B,Sun Q,Fan Y,Hu X,Li L,Wang J,Cui S
Published figure using CD73 monoclonal antibody (Product # 25-0731-82) in Flow Cytometry
Sat Jan 16 00:00:00 EST 2021
Negative feedback control of neuronal activity by microglia.
Nature
Badimon A,Strasburger HJ,Ayata P,Chen X,Nair A,Ikegami A,Hwang P,Chan AT,Graves SM,Uweru JO,Ledderose C,Kutlu MG,Wheeler MA,Kahan A,Ishikawa M,Wang YC,Loh YE,Jiang JX,Surmeier DJ,Robson SC,Junger WG,Sebra R,Calipari ES,Kenny PJ,Eyo UB,Colonna M,Quintana FJ,Wake H,Gradinaru V,Schaefer A
Published figure using CD73 monoclonal antibody (Product # 25-0731-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2020
Elicitation of Potent Neutralizing Antibody Responses by Designed Protein Nanoparticle Vaccines for SARS-CoV-2.
Cell
Walls AC,Fiala B,Schäfer A,Wrenn S,Pham MN,Murphy M,Tse LV,Shehata L,O'Connor MA,Chen C,Navarro MJ,Miranda MC,Pettie D,Ravichandran R,Kraft JC,Ogohara C,Palser A,Chalk S,Lee EC,Guerriero K,Kepl E,Chow CM,Sydeman C,Hodge EA,Brown B,Fuller JT,Dinnon KH,Gralinski LE,Leist SR,Gully KL,Lewis TB,Guttman M,Chu HY,Lee KK,Fuller DH,Baric RS,Kellam P,Carter L,Pepper M,Sheahan TP,Veesler D,King NP
25-0731 was used in Flow cytometry/Cell sorting to develop a structure-based design of non-particle immunogens to elicit protective antibody responses against SARS-CoV-2 in mice.
Wed Nov 25 00:00:00 EST 2020
A Single Immunization with Nucleoside-Modified mRNA Vaccines Elicits Strong Cellular and Humoral Immune Responses against SARS-CoV-2 in Mice.
Immunity
Laczkó D,Hogan MJ,Toulmin SA,Hicks P,Lederer K,Gaudette BT,Castaño D,Amanat F,Muramatsu H,Oguin TH,Ojha A,Zhang L,Mu Z,Parks R,Manzoni TB,Roper B,Strohmeier S,Tombácz I,Arwood L,Nachbagauer R,Karikó K,Greenhouse J,Pessaint L,Porto M,Putman-Taylor T,Strasbaugh A,Campbell TA,Lin PJC,Tam YK,Sempowski GD,Farzan M,Choe H,Saunders KO,Haynes BF,Andersen H,Eisenlohr LC,Weissman D,Krammer F,Bates P,Allman D,Locci M,Pardi N
25-0731 was used in Flow cytometry/Cell sorting to study the immunogenicity of lipid nanoparticle-encapsulated, nucleoside-modified mRNA vaccines against SARS-CoV-2 in mice.
Tue Oct 13 00:00:00 EDT 2020
Sema3A and HIF1α co-overexpressed iPSC-MSCs/HA scaffold facilitates the repair of calvarial defect in a mouse model.
Journal of cellular physiology
Li J,Wang T,Li C,Wang Z,Wang P,Zheng L
Published figure using CD73 monoclonal antibody (Product # 25-0731-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2020
Elicitation of potent neutralizing antibody responses by designed protein nanoparticle vaccines for SARS-CoV-2.
bioRxiv : the preprint server for biology
Walls AC,Fiala B,Schäfer A,Wrenn S,Pham MN,Murphy M,Tse LV,Shehata L,O'Connor MA,Chen C,Navarro MJ,Miranda MC,Pettie D,Ravichandran R,Kraft JC,Ogohara C,Palser A,Chalk S,Lee EC,Kepl E,Chow CM,Sydeman C,Hodge EA,Brown B,Fuller JT,Dinnon KH,Gralinski LE,Leist SR,Gully KL,Lewis TB,Guttman M,Chu HY,Lee KK,Fuller DH,Baric RS,Kellam P,Carter L,Pepper M,Sheahan TP,Veesler D,King NP
25-0731-82 was used in Flow Cytometry to highlight the utility of robust antigen display platforms for inducing potent neutralizing antibody responses and launch cGMP manufacturing efforts to advance the lead RBD nanoparticle vaccine into the clinic.
Wed Aug 12 00:00:00 EDT 2020
CD73 Promotes Glioblastoma Pathogenesis and Enhances Its Chemoresistance via A2B Adenosine Receptor Signaling.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Yan A,Joachims ML,Thompson LF,Miller AD,Canoll PD,Bynoe MS
Published figure using CD73 monoclonal antibody (Product # 25-0731-82) in Flow Cytometry
Wed May 29 00:00:00 EDT 2019
PDGF is a potent initiator of bone formation in a tissue engineered model of pathological ossification.
Journal of tissue engineering and regenerative medicine
Davies OG,Grover LM,Lewis MP,Liu Y
Published figure using CD73 monoclonal antibody (Product # 25-0731-82) in Flow Cytometry
Mon Jan 01 00:00:00 EST 2018
PDGF is a potent initiator of bone formation in a tissue engineered model of pathological ossification.
Journal of tissue engineering and regenerative medicine
Davies OG,Grover LM,Lewis MP,Liu Y
Published figure using CD73 monoclonal antibody (Product # 25-0731-82) in Flow Cytometry
Mon Jan 01 00:00:00 EST 2018
Calcium-mediated shaping of naive CD4 T-cell phenotype and function.
eLife
Guichard V,Bonilla N,Durand A,Audemard-Verger A,Guilbert T,Martin B,Lucas B,Auffray C
Published figure using CD73 monoclonal antibody (Product # 25-0731-82) in Flow Cytometry
Thu Dec 14 00:00:00 EST 2017
Three distinct developmental pathways for adaptive and two IFN-γ-producing γδ T subsets in adult thymus.
Nature communications
Buus TB,Ødum N,Geisler C,Lauritsen JPH
Published figure using CD73 monoclonal antibody (Product # 25-0731-82) in Flow Cytometry
Mon Dec 04 00:00:00 EST 2017
Co-inhibition of CD73 and A2AR Adenosine Signaling Improves Anti-tumor Immune Responses.
Cancer cell
Young A,Ngiow SF,Barkauskas DS,Sult E,Hay C,Blake SJ,Huang Q,Liu J,Takeda K,Teng MWL,Sachsenmeier K,Smyth MJ
25-0731 was used in Flow cytometry/Cell sorting to identify that co-blockade of the ectonucleotidase that generates adenosine CD73 and the A2A adenosine receptor that mediates adenosine signalling in leuokocytes limits tumour initiation, growth and metastasis.
Mon Sep 12 00:00:00 EDT 2016
Developmental Progression and Interrelationship of Central and Effector Regulatory T Cell Subsets.
Journal of immunology (Baltimore, Md. : 1950)
Toomer KH,Yuan X,Yang J,Dee MJ,Yu A,Malek TR
25-0731 was used in Flow cytometry/Cell sorting to elucidate the developmental progression and interrelationship of central and effector regulatory T cell subsets.
Sun May 01 00:00:00 EDT 2016
Accelerated craniofacial bone regeneration through dense collagen gel scaffolds seeded with dental pulp stem cells.
Scientific reports
Chamieh F,Collignon AM,Coyac BR,Lesieur J,Ribes S,Sadoine J,Llorens A,Nicoletti A,Letourneur D,Colombier ML,Nazhat SN,Bouchard P,Chaussain C,Rochefort GY
Published figure using CD73 monoclonal antibody (Product # 25-0731-82) in Flow Cytometry
Fri Dec 09 00:00:00 EST 2016
Limited clonal relatedness between gut IgA plasma cells and memory B cells after oral immunization.
Nature communications
Bemark M,Hazanov H,Strömberg A,Komban R,Holmqvist J,Köster S,Mattsson J,Sikora P,Mehr R,Lycke NY
Published figure using CD73 monoclonal antibody (Product # 25-0731-82) in Flow Cytometry
Tue Sep 06 00:00:00 EDT 2016
Perinatal tolerance to proinsulin is sufficient to prevent autoimmune diabetes.
JCI insight
Jhala G,Chee J,Trivedi PM,Selck C,Gurzov EN,Graham KL,Thomas HE,Kay TW,Krishnamurthy B
25-0731 was used in Flow cytometry/Cell sorting to offer that therapeutic interventions targeting proinsulin early on in life in high-risk subjects may prevent later diabetes development.
Thu Jul 07 00:00:00 EDT 2016
Adipose-Derived Mesenchymal Stem Cells Prevent Systemic Bone Loss in Collagen-Induced Arthritis.
Journal of immunology (Baltimore, Md. : 1950)
Garimella MG,Kour S,Piprode V,Mittal M,Kumar A,Rani L,Pote ST,Mishra GC,Chattopadhyay N,Wani MR
25-0731 was used in Flow cytometry/Cell sorting to investigate role of mesechymal stem cells in prevention of bone destruction in rheumatoid arthritis.
Tue Dec 01 00:00:00 EST 2015
Vitamin A Impairs the Reprogramming of Tregs into IL-17-Producing Cells during Intestinal Inflammation.
BioMed research international
Tejón G,Manríquez V,De Calisto J,Flores-Santibáñez F,Hidalgo Y,Crisóstomo N,Fernández D,Sauma D,Mora JR,Bono MR,Rosemblatt M
25-0731 was used in Flow cytometry/Cell sorting to study the contribution of different factors to the reprogramming of iTregs into IL-17-producing cells, showing that vitamin A impairs reprogramming during intestinal inflammation.
Fri Sep 02 00:00:00 EDT 2016
Cell-autonomous heparanase modulates self-renewal and migration in bone marrow-derived mesenchymal stem cells.
Journal of biomedical science
Cheng CC,Lee YH,Lin SP,Huangfu WC,Liu IH
Published figure using CD73 monoclonal antibody (Product # 25-0731-82) in Flow Cytometry
Thu Mar 13 00:00:00 EDT 2014
Multipotent stem cells from trabecular meshwork become phagocytic TM cells.
Investigative ophthalmology & visual science
Du Y,Roh DS,Mann MM,Funderburgh ML,Funderburgh JL,Schuman JS
Published figure using CD73 monoclonal antibody (Product # 25-0731-82) in Flow Cytometry
Wed Mar 21 00:00:00 EDT 2012
Adipose-derived stem cells promote lymphangiogenesis in response to VEGF-C stimulation or TGF-β1 inhibition.
Future oncology (London, England)
Yan A,Avraham T,Zampell JC,Haviv YS,Weitman E,Mehrara BJ
Published figure using CD73 monoclonal antibody (Product # 25-0731-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2011