Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Specificity: Reacts with an extracellular epitope of CD79b.
Preparation: HM79-16 was initially generated by purifying CD79b from WEHI231 B cells and used for immunisation of footpads of an Armenian hamster. Hybridoma supernatants were screened for positive reactivity in surface immunofluorescence of WEHI231 and then the specificity to CD79b was tested by immuno-precipitationRecombinant anti-CD79 antibodies were engineered by cloning and sequencing the full-length IG heavy and light chains of an Armenian hamster (Cricetulus migratorius).
CD79b is the B cell antigen receptor Ig beta chain (BCR-Ig beta), which associates with B cell antigen receptor Ig alpha chain to form a functional heterodimer that interacts with membrane immunoglobulin (mIg). The mIgM component binds antigen and the Ig alpha /I g beta heterodimer tranduces signals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell antigen receptor complex-associated protein beta chain; B-cell-specific glycoprotein B29; CD79; CD79b; Ig beta; Ig-beta; Immunoglobulin-associated B29 protein; immunoglobulin-associated beta; RP23-418O11.7
Gene Aliases: B29; Cd79b; Ig-beta; Igb; Igbeta
UniProt ID: (Mouse) P15530
Entrez Gene ID: (Mouse) 15985
Molecular Function: immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support