Product References
Effect of mesenchymal stem cell-derived exosomes on the induction of mouse tolerogenic dendritic cells.
Journal of cellular physiology
Shahir M,Mahmoud Hashemi S,Asadirad A,Varahram M,Kazempour-Dizaji M,Folkerts G,Garssen J,Adcock I,Mortaz E
12-0831 was used in Flow cytometry/Cell sorting to investigate the effects of mesenchymal stem cell-derived exosomes and to understand their role in modulating the mouse tolergenic dendritic cell immune responses.
Thu Oct 01 00:00:00 UTC 2020
Immunoadjuvant Activity of Fucoidans from the Brown Alga Fucus evanescens.
Marine drugs
Kuznetsova TA,Smolina TP,Makarenkova ID,Ivanushko LA,Persiyanova EV,Ermakova SP,Silchenko AS,Zaporozhets TS,Besednova NN,Fedyanina LN,Kryzhanovsky SP
12-0831 was used in Flow cytometry/Cell sorting to comparatively evaluate the effect of fucoidans from the brown alga Fucus evanescens on effector functions of innate and adaptive immunity cells in vitro and in vivo.
Wed Mar 11 00:00:00 UTC 2020
Activated Peyer's patch B cells sample antigen directly from M cells in the subepithelial dome.
Nature communications
Komban RJ,Strömberg A,Biram A,Cervin J,Lebrero-Fernández C,Mabbott N,Yrlid U,Shulman Z,Bemark M,Lycke N
12-0831-82 was used in Flow Cytometry to report a M cell-B cell antigen-specific transporting pathway in PP that is independent of DC. We propose that this antigen transporting pathway has a critical role in gut IgA responses, and should be taken into account when developing mucosal vaccines.
Mon Jun 03 00:00:00 UTC 2019
Activated Peyer's patch B cells sample antigen directly from M cells in the subepithelial dome.
Nature communications
Komban RJ,Strömberg A,Biram A,Cervin J,Lebrero-Fernández C,Mabbott N,Yrlid U,Shulman Z,Bemark M,Lycke N
12-0831-82 was used in Flow Cytometry to report a M cell-B cell antigen-specific transporting pathway in PP that is independent of DC. We propose that this antigen transporting pathway has a critical role in gut IgA responses, and should be taken into account when developing mucosal vaccines.
Mon Jun 03 00:00:00 UTC 2019
Pplase of Dermatophagoides farinae promotes ovalbumin-induced airway allergy by modulating the functions of dendritic cells in a mouse model.
Scientific reports
Wang H,Mo L,Xiao X,An S,Liu X,Ba J,Wu W,Ran P,Yang P,Liu Z
Published figure using CD83 monoclonal antibody (Product # 12-0831-82) in Flow Cytometry
Mon Feb 27 00:00:00 UTC 2017
Pplase of Dermatophagoides farinae promotes ovalbumin-induced airway allergy by modulating the functions of dendritic cells in a mouse model.
Scientific reports
Wang H,Mo L,Xiao X,An S,Liu X,Ba J,Wu W,Ran P,Yang P,Liu Z
Published figure using CD83 monoclonal antibody (Product # 12-0831-82) in Flow Cytometry
Mon Feb 27 00:00:00 UTC 2017
Der f 31, a novel allergen from Dermatophagoides farinae, activates epithelial cells and enhances lung-resident group 2 innate lymphoid cells.
Scientific reports
Wang H,Lin J,Zeng L,Ouyang C,Ran P,Yang P,Liu Z
Published figure using CD83 monoclonal antibody (Product # 12-0831-82) in Flow Cytometry
Thu Aug 17 00:00:00 UTC 2017
Thiol peroxiredoxin, a novel allergen from Bombyx mori, modulates functions of macrophages and dendritic cells.
American journal of translational research
Wang H,Hu W,Liang Z,Zeng L,Li J,Yan H,Yang P,Liu Z,Wang L
12-0831 was used in Flow cytometry/Cell sorting to study how thiol peroxiredoxin, a novel allergen from Bombyx mori, modulates functions of macrophages and dendritic cells.
Thu Oct 01 00:00:00 UTC 2020
Angiotensin converting enzyme inhibitors and angiotensin II receptor antagonist attenuate tumor growth via polarization of neutrophils toward an antitumor phenotype.
Oncoimmunology
Shrestha S,Noh JM,Kim SY,Ham HY,Kim YJ,Yun YJ,Kim MJ,Kwon MS,Song DK,Hong CW
12-0831 was used in Flow cytometry/Cell sorting to investigate how modulation of Ang II pathway attenuates tumour growth via polarisation of neutrophils to an antitumoural phenotype.
Tue Jan 12 00:00:00 UTC 2021
Enhanced Anti-Tumor Efficacy through a Combination of Integrin αvβ6-Targeted Photodynamic Therapy and Immune Checkpoint Inhibition.
Theranostics
Gao L,Zhang C,Gao D,Liu H,Yu X,Lai J,Wang F,Lin J,Liu Z
12-0831 was used in Flow cytometry/Cell sorting to investigate whether integrin αvβ6-targeted photodynamic therapy (PDT) of tumors using a DSAB-HK could trigger the host immune response, showing that DSAB-HK led to inhibited growth of 4T1 tumours and promoted maturation of dendritic cells.
Tue Dec 13 00:00:00 UTC 2016
Distinct roles for MyD88 and Toll-like receptor 2 during Leishmania braziliensis infection in mice.
Infection and immunity
Vargas-Inchaustegui DA,Tai W,Xin L,Hogg AE,Corry DB,Soong L
12-0831 was used in Flow cytometry/Cell sorting to investigate whether TLRs are involved in L. braziliensis-mediated dendritic cell activation, showing distinct roles for MyD88 and TLR2 in infection.
Wed Jul 01 00:00:00 UTC 2009
Nitric oxide is a key component in inflammation-accelerated tumorigenesis.
Cancer research
Hussain SP,He P,Subleski J,Hofseth LJ,Trivers GE,Mechanic L,Hofseth AB,Bernard M,Schwank J,Nguyen G,Mathe E,Djurickovic D,Haines D,Weiss J,Back T,Gruys E,Laubach VE,Wiltrout RH,Harris CC
12-0831 was used in Flow cytometry/Cell sorting to test the hypothesis that an inflammatory microenvironment with an enhanced level of nitric oxide accelerates spontaneous tumour development (tumorigenesis).
Mon Sep 01 00:00:00 UTC 2008
Leishmania braziliensis infection induces dendritic cell activation, ISG15 transcription, and the generation of protective immune responses.
Journal of immunology (Baltimore, Md. : 1950)
Vargas-Inchaustegui DA,Xin L,Soong L
12-0831 was used in Flow cytometry/Cell sorting to investigate whether Leishmania braziliensis infection efficiently triggers innate responses in dendritic cells and the subsequent adaptive immune responses for parasite clearance.
Sun Jun 01 00:00:00 UTC 2008
Down-regulation of dendritic cell signaling pathways by Leishmania amazonensis amastigotes.
Molecular immunology
Xin L,Li K,Soong L
12-0831 was used in Flow cytometry/Cell sorting to suggest that La parasites, especially in their intracellular forms, have evolved unique strategies to actively down-regulate early innate signalling events.
Tue Jul 01 00:00:00 UTC 2008
Role of natural killer cells in modulating dendritic cell responses to Leishmania amazonensis infection.
Infection and immunity
Sanabria MX,Vargas-Inchaustegui DA,Xin L,Soong L
12-0831 was used in Flow cytometry/Cell sorting to suggest differential roles of DC-NK cell cross talk at different stages of Leishmania infection.
Sat Nov 01 00:00:00 UTC 2008
Effects of CXCL10 on dendritic cell and CD4+ T-cell functions during Leishmania amazonensis infection.
Infection and immunity
Vasquez RE,Xin L,Soong L
12-0831 was used in Flow cytometry/Cell sorting to investigate the mechanism underlying enhanced immunity against L. amazonensis infection following CXC10 treatment, showing that treatment has an effect on dendritic cell and CD4+ T-cell functions.
Tue Jan 01 00:00:00 UTC 2008
Role of interleukin-1beta in activating the CD11c(high) CD45RB- dendritic cell subset and priming Leishmania amazonensis-specific CD4+ T cells in vitro and in vivo.
Infection and immunity
Xin L,Li Y,Soong L
12-0831 was used in Flow cytometry/Cell sorting to investigate the mechanism by which the Leishmania amazonensis parasite modulates antigen-presenting cells and disrupts the immune system.
Mon Oct 01 00:00:00 UTC 2007