Product References
A pH-Sensitive Nanoparticle as Reactive Oxygen Species Amplifier to Regulate Tumor Microenvironment and Potentiate Tumor Radiotherapy.
International journal of nanomedicine
Jiang X,Jiang X,Wu D,Xie W,Liu X,Zheng J
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Jan 30 00:00:00 UTC 2024
TLR4 is one of the receptors for Chikungunya virus envelope protein E2 and regulates virus induced pro-inflammatory responses in host macrophages.
Frontiers in immunology
Mahish C,De S,Chatterjee S,Ghosh S,Keshry SS,Mukherjee T,Khamaru S,Tung KS,Subudhi BB,Chattopadhyay S,Chattopadhyay S
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu May 18 00:00:00 UTC 2023
Carvacrol protects mice against LPS-induced sepsis and attenuates inflammatory response in macrophages by modulating the ERK1/2 pathway.
Scientific reports
Yan C,Kuang W,Jin L,Wang R,Niu L,Xie C,Ding J,Liao Y,Wang L,Wan H,Ma G
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Aug 07 00:00:00 UTC 2023
TREM2 expression promotes liver and peritoneal M2 macrophage polarization in mice infected with Schistosoma japonicum.
Journal of cellular and molecular medicine
Zhu D,Huang M,Shen P,Zhang B,Chen G,Chen J,Duan L,Duan Y
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Aug 01 00:00:00 UTC 2023
A platinum@polymer-catechol nanobraker enables radio-immunotherapy for crippling melanoma tumorigenesis, angiogenesis, and radioresistance.
Bioactive materials
Li W,Yan J,Tian H,Li B,Wang G,Sang W,Zhang Z,Zhang X,Dai Y
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sat Apr 01 00:00:00 UTC 2023
BushenHuoxue decoction suppresses M1 macrophage polarization and prevents LPS induced inflammatory bone loss by activating AMPK pathway.
Heliyon
Chen S,Tao L,Zhu F,Wang Z,Zhuang Q,Li Y,Yang Y,Feng C,Shi H,Shi J,Zhu L,Xiao L,Geng D,Wang Z
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon May 01 00:00:00 UTC 2023
CD8α+ dendritic cells potentiate antitumor and immune activities against murine ovarian cancers.
Scientific reports
Lee SW,Lee H,Lee KW,Kim MJ,Kang SW,Lee YJ,Kim H,Kim YM
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Jan 03 00:00:00 UTC 2023
Acinetobacter baumannii outer membrane protein A induces autophagy in bone marrow-derived dendritic cells involving the PI3K/mTOR pathway.
Immunity, inflammation and disease
Tan H,Cao L
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sat Apr 01 00:00:00 UTC 2023
Guidelines for mouse and human DC generation.
European journal of immunology
Lutz MB,Ali S,Audiger C,Autenrieth SE,Berod L,Bigley V,Cyran L,Dalod M,Dörrie J,Dudziak D,Flórez-Grau G,Giusiano L,Godoy GJ,Heuer M,Krug AB,Lehmann CHK,Mayer CT,Naik SH,Scheu S,Schreibelt G,Segura E,Seré K,Sparwasser T,Tel J,Xu H,Zenke M
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Wed Nov 01 00:00:00 UTC 2023
Exosomal Lnc NEAT1 from endothelial cells promote bone regeneration by regulating macrophage polarization via DDX3X/NLRP3 axis.
Journal of nanobiotechnology
Chen Y,Wu Y,Guo L,Yuan S,Sun J,Zhao K,Wang J,An R
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Mar 20 00:00:00 UTC 2023
Role of low-intensity pulsed ultrasound in regulating macrophage polarization to accelerate tendon-bone interface repair.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Xu Z,Li S,Wan L,Hu J,Lu H,Zhang T
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon May 01 00:00:00 UTC 2023
BMP-2 functional polypeptides relieve osteolysis via bi-regulating bone formation and resorption coupled with macrophage polarization.
NPJ Regenerative medicine
Wang J,Xue Y,Wang Y,Liu C,Hu S,Zhao H,Gu Q,Yang H,Huang L,Zhou X,Shi Q
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu Feb 09 00:00:00 UTC 2023
Oxygen-carrying nanoplatform to reprogram tumor immunosuppressive microenvironment and enhance photothermal-immunotherapy.
Materials today. Bio
Huang J,Leng X,Jiang T,Xu L,Zheng J,Fang M,Wang J,Wang Z,Zhang L
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sat Apr 01 00:00:00 UTC 2023
Nanoplateletsomes restrain metastatic tumor formation through decoy and active targeting in a preclinical mouse model.
Acta pharmaceutica Sinica. B
Zhang L,Zhu Y,Wei X,Chen X,Li Y,Zhu Y,Xia J,Huang Y,Huang Y,Wang J,Pang Z
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2022
Enhanced uptake and anti-maturation effect of celastrol-loaded mannosylated liposomes on dendritic cells for psoriasis treatment.
Acta pharmaceutica Sinica. B
Xi L,Lin Z,Qiu F,Chen S,Li P,Chen X,Wang Z,Zheng Y
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sat Jan 01 00:00:00 UTC 2022
Enhancing arginase 2 expression using target site blockers as a strategy to modulate macrophage phenotype.
Molecular therapy. Nucleic acids
De Santi C,Nally FK,Afzal R,Duffy CP,Fitzsimons S,Annett SL,Robson T,Dowling JK,Cryan SA,McCoy CE
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Sep 13 00:00:00 UTC 2022
Oncolytic Zika virus promotes intratumoral T cell infiltration and improves immunotherapy efficacy in glioblastoma.
Molecular therapy oncolytics
Chen L,Zhou C,Chen Q,Shang J,Liu Z,Guo Y,Li C,Wang H,Ye Q,Li X,Zu S,Li F,Xia Q,Zhou T,Li A,Wang C,Chen Y,Wu A,Qin C,Man J
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu Mar 17 00:00:00 UTC 2022
G9a promotes inflammation in Streptococcus pneumoniae induced pneumonia mice by stimulating M1 macrophage polarization and H3K9me2 methylation in FOXP1 promoter region.
Annals of translational medicine
Li Y,Li G,Zhang L,Li Y,Zhao Z
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sun May 01 00:00:00 UTC 2022
Intranasal Immunization With a c-di-GMP-Adjuvanted Acellular Pertussis Vaccine Provides Superior Immunity Against Bordetella pertussis in a Mouse Model.
Frontiers in immunology
Jiang W,Wang X,Su Y,Cai L,Li J,Liang J,Gu Q,Sun M,Shi L
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue May 03 00:00:00 UTC 2022
Meningeal lymphatic drainage promotes T cell responses against Toxoplasma gondii but is dispensable for parasite control in the brain.
eLife
Kovacs MA,Cowan MN,Babcock IW,Sibley LA,Still K,Batista SJ,Labuzan SA,Sethi I,Harris TH
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Wed Dec 21 00:00:00 UTC 2022
MicroRNA-382 Promotes M2-Like Macrophage via the SIRP-α/STAT3 Signaling Pathway in Aristolochic Acid-Induced Renal Fibrosis.
Frontiers in immunology
Wang X,Jia P,Ren T,Zou Z,Xu S,Zhang Y,Shi Y,Bao S,Li Y,Fang Y,Ding X
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Fri May 20 00:00:00 UTC 2022
CD4+ CTLs Act as a Key Effector Population for Allograft Rejection of MSCs in a Donor MHC-II Dependent Manner in Injured Liver.
Aging and disease
Shen S,Li Y,Jin M,Fan D,Pan R,Lin A,Chen Y,Xiang L,Zhao RC,Shao J
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu Dec 01 00:00:00 UTC 2022
Chromomycin A5 induces bona fide immunogenic cell death in melanoma.
Frontiers in immunology
Florêncio KGD,Edson EA,Fernandes KSDS,Luiz JPM,Pinto FDCL,Pessoa ODL,Cunha FQ,Machado-Neto JA,Wilke DV
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Nov 29 00:00:00 UTC 2022
In situ generation of micrometer-sized tumor cell-derived vesicles as autologous cancer vaccines for boosting systemic immune responses.
Nature communications
Guo Y,Wang SZ,Zhang X,Jia HR,Zhu YX,Zhang X,Gao G,Jiang YW,Li C,Chen X,Wu SY,Liu Y,Wu FG
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Nov 01 00:00:00 UTC 2022
Targeting dendritic cells with TLR-2 ligand-coated nanoparticles loaded with Mycobacterium tuberculosis epitope induce antituberculosis immunity.
The Journal of biological chemistry
Kumar Das D,Zafar MA,Nanda S,Singh S,Lamba T,Bashir H,Singh P,Maurya SK,Nadeem S,Sehrawat S,Bhalla V,Agrewala JN
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu Dec 01 00:00:00 UTC 2022
Intracellular osteopontin protects from autoimmunity-driven lymphoma development inhibiting TLR9-MYD88-STAT3 signaling.
Molecular cancer
Rizzello C,Cancila V,Sangaletti S,Botti L,Ratti C,Milani M,Dugo M,Bertoni F,Tripodo C,Chiodoni C,Colombo MP
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Dec 12 00:00:00 UTC 2022
Dental pulp stem cell-derived exosomes suppress M1 macrophage polarization through the ROS-MAPK-NFκB P65 signaling pathway after spinal cord injury.
Journal of nanobiotechnology
Liu C,Hu F,Jiao G,Guo Y,Zhou P,Zhang Y,Zhang Z,Yi J,You Y,Li Z,Wang H,Zhang X
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Wed Feb 02 00:00:00 UTC 2022
Self-assembled traditional Chinese nanomedicine modulating tumor immunosuppressive microenvironment for colorectal cancer immunotherapy.
Theranostics
Mao Q,Min J,Zeng R,Liu H,Li H,Zhang C,Zheng A,Lin J,Liu X,Wu M
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu Sep 29 00:00:00 UTC 2022
Role of circular RNA cdr1as in modulation of macrophage phenotype.
Life sciences
Gonzalez C,Cimini M,Cheng Z,Benedict C,Wang C,Trungcao M,Mallaredy V,Rajan S,Garikipati VNS,Kishore R
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Nov 15 00:00:00 UTC 2022
A bilirubin-derived nanomedicine attenuates the pathological cascade of pulmonary fibrosis.
Biomaterials
Keum H,Kim D,Kim J,Kim TW,Whang CH,Jung W,Jon S
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sun Aug 01 00:00:00 UTC 2021
Loss of p53 in mesenchymal stem cells promotes alteration of bone remodeling through negative regulation of osteoprotegerin.
Cell death and differentiation
Velletri T,Huang Y,Wang Y,Li Q,Hu M,Xie N,Yang Q,Chen X,Chen Q,Shou P,Gan Y,Candi E,Annicchiarico-Petruzzelli M,Agostini M,Yang H,Melino G,Shi Y,Wang Y
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2021
CD11b Deficiency Exacerbates Methicillin-Resistant Staphylococcus aureus-Induced Sepsis by Upregulating Inflammatory Responses of Macrophages.
Immune network
Sim H,Jeong D,Kim HI,Pak S,Thapa B,Kwon HJ,Lee K
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu Apr 01 00:00:00 UTC 2021
Pseudomonas aeruginosa PcrV Enhances the Nitric Oxide-Mediated Tumoricidal Activity of Tumor-Associated Macrophages via a TLR4/PI3K/AKT/mTOR-Glycolysis-Nitric Oxide Circuit.
Frontiers in oncology
Yu H,Bai Y,Qiu J,He X,Xiong J,Dai Q,Wang X,Li Y,Sheng H,Xin R,Jiang L,Li Q,Li D,Zhang H,Zhang L,Chen Q,Peng J,Hu X,Zhang K
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Wed Dec 15 00:00:00 UTC 2021
Prostaglandin E2 serves a dual role in regulating the migration of dendritic cells.
International journal of molecular medicine
Diao G,Huang J,Zheng X,Sun X,Tian M,Han J,Guo J
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Fri Jan 01 00:00:00 UTC 2021
Exosomes derived from inflammatory myoblasts promote M1 polarization and break the balance of myoblast proliferation/differentiation.
World journal of stem cells
Luo ZW,Sun YY,Lin JR,Qi BJ,Chen JW
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Fri Nov 26 00:00:00 UTC 2021
Altered Corneal Epithelial Dendritic Cell Morphology and Phenotype Following Acute Exposure to Hyperosmolar Saline.
Investigative ophthalmology & visual science
Senthil K,Jiao H,Downie LE,Chinnery HR
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Immunocytochemistry
Mon Feb 01 00:00:00 UTC 2021
Myeloid and plasmacytoid dendritic cell combined vaccines loaded with heat-treated tumor cell lysates enhance antitumor activity in murine lung cancer.
Oncology letters
Chen H,Tan J,Li X,Li H,Wu W,Wu Y,Zhang J,Gu L
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2021
Polarization of Type 1 Macrophages Is Associated with the Severity of Viral Encephalitis Caused by Japanese Encephalitis Virus and Dengue Virus.
Cells
Jhan MK,Chen CL,Shen TJ,Tseng PC,Wang YT,Satria RD,Yu CY,Lin CF
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Nov 15 00:00:00 UTC 2021
Antigen presentation between T cells drives Th17 polarization under conditions of limiting antigen.
Cell reports
Boccasavia VL,Bovolenta ER,Villanueva A,Borroto A,Oeste CL,van Santen HM,Prieto C,Alonso-López D,Diaz-Muñoz MD,Batista FD,Alarcón B
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Immunocytochemistry
Tue Mar 16 00:00:00 UTC 2021
Modulation of lactate-lysosome axis in dendritic cells by clotrimazole potentiates antitumor immunity.
Journal for immunotherapy of cancer
Wang Z,Xu F,Hu J,Zhang H,Cui L,Lu W,He W,Wang X,Li M,Zhang H,Xiong W,Xie C,Liu Y,Zhou P,Liu J,Huang P,Qin XF,Xia X
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sat May 01 00:00:00 UTC 2021
A two-adjuvant multiantigen candidate vaccine induces superior protective immune responses against SARS-CoV-2 challenge.
Cell reports
Jiang W,Shi L,Cai L,Wang X,Li J,Li H,Liang J,Gu Q,Ji G,Li J,Liu L,Sun M
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Dec 14 00:00:00 UTC 2021
Theranostic near-infrared-IIb emitting nanoprobes for promoting immunogenic radiotherapy and abscopal effects against cancer metastasis.
Nature communications
Li H,Wang M,Huang B,Zhu SW,Zhou JJ,Chen DR,Cui R,Zhang M,Sun ZJ
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu Dec 09 00:00:00 UTC 2021
Inhibition of TLR4 signaling protects mice from sensory and motor dysfunction in an animal model of autoimmune peripheral neuropathy.
Journal of neuroinflammation
Oladiran O,Shi XQ,Yang M,Fournier S,Zhang J
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Mar 22 00:00:00 UTC 2021
Engineered macrophages as near-infrared light activated drug vectors for chemo-photodynamic therapy of primary and bone metastatic breast cancer.
Nature communications
Huang Y,Guan Z,Dai X,Shen Y,Wei Q,Ren L,Jiang J,Xiao Z,Jiang Y,Liu D,Huang Z,Xu X,Luo Y,Zhao C
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Wed Jul 14 00:00:00 UTC 2021
Adrenomedullin 2 improves bone regeneration in type 1 diabetic rats by restoring imbalanced macrophage polarization and impaired osteogenesis.
Stem cell research & therapy
Wang F,Kong L,Wang W,Shi L,Wang M,Chai Y,Xu J,Kang Q
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu May 13 00:00:00 UTC 2021
Marine-inspired molecular mimicry generates a drug-free, but immunogenic hydrogel adhesive protecting surgical anastomosis.
Bioactive materials
Huang J,Jiang Y,Liu Y,Ren Y,Xu Z,Li Z,Zhao Y,Wu X,Ren J
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Mar 01 00:00:00 UTC 2021
Cytochrome P450 26A1 Modulates the Polarization of Uterine Macrophages During the Peri-Implantation Period.
Frontiers in immunology
Ji WH,Li DD,Wei DP,Gu AQ,Yang Y,Peng JP
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu Feb 10 00:00:00 UTC 2022
C-Reactive Protein Suppresses the Th17 Response Indirectly by Attenuating the Antigen Presentation Ability of Monocyte Derived Dendritic Cells in Experimental Autoimmune Encephalomyelitis.
Frontiers in immunology
Shen ZY,Zheng Y,Pecsok MK,Wang K,Li W,Gong MJ,Wu F,Zhang L
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu Jul 01 00:00:00 UTC 2021
PP2Cδ Controls the Differentiation and Function of Dendritic Cells Through Regulating the NSD2/mTORC2/ACLY Pathway.
Frontiers in immunology
Lv N,Jin S,Liang Z,Wu X,Kang Y,Su L,Dong Y,Wang B,Ma T,Shi L
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Feb 28 00:00:00 UTC 2022
E6‑regulated overproduction of prostaglandin E2 may inhibit migration of dendritic cells in human papillomavirus 16‑positive cervical lesions.
International journal of oncology
Huang J,Diao G,Zhang Q,Chen Y,Han J,Guo J
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Wed Apr 01 00:00:00 UTC 2020
Mesenchymal stem cell-secreted extracellular vesicles carrying TGF-β1 up-regulate miR-132 and promote mouse M2 macrophage polarization.
Journal of cellular and molecular medicine
Wang Y,Han B,Wang Y,Wang C,Zhang H,Xue J,Wang X,Niu T,Niu Z,Chen Y
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sun Nov 01 00:00:00 UTC 2020
Interleukin-9 Inhibits Lung Metastasis of Melanoma through Stimulating Anti-Tumor M1 Macrophages.
Molecules and cells
Park SM,Do-Thi VA,Lee JO,Lee H,Kim YS
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sun May 31 00:00:00 UTC 2020
Effects of combined treatment with PD‑L1 Ig and CD40L mAb on immune tolerance in the CBA/J x DBA/2 mouse model.
Molecular medicine reports
Li G,Yang L,Li D,Zhang J,Du L,Xia L,Liu Y,Hu W
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Wed Apr 01 00:00:00 UTC 2020
Angiotensin II induces RAW264.7 macrophage polarization to the M1‑type through the connexin 43/NF‑κB pathway.
Molecular medicine reports
Wu L,Chen K,Xiao J,Xin J,Zhang L,Li X,Li L,Si J,Wang L,Ma K
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Fri May 01 00:00:00 UTC 2020
Vaccination with early ferroptotic cancer cells induces efficient antitumor immunity.
Journal for immunotherapy of cancer
Efimova I,Catanzaro E,Van der Meeren L,Turubanova VD,Hammad H,Mishchenko TA,Vedunova MV,Fimognari C,Bachert C,Coppieters F,Lefever S,Skirtach AG,Krysko O,Krysko DV
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sun Nov 01 00:00:00 UTC 2020
Intravenous injection of the oncolytic virus M1 awakens antitumor T cells and overcomes resistance to checkpoint blockade.
Cell death & disease
Liu Y,Cai J,Liu W,Lin Y,Guo L,Liu X,Qin Z,Xu C,Zhang Y,Su X,Deng K,Yan G,Liang J
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sat Dec 12 00:00:00 UTC 2020
MiR-103 protects from recurrent spontaneous abortion via inhibiting STAT1 mediated M1 macrophage polarization.
International journal of biological sciences
Zhu X,Liu H,Zhang Z,Wei R,Zhou X,Wang Z,Zhao L,Guo Q,Zhang Y,Chu C,Wang L,Li X
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Fri Nov 12 00:00:00 UTC 2021
Senolytics prevent mt-DNA-induced inflammation and promote the survival of aged organs following transplantation.
Nature communications
Iske J,Seyda M,Heinbokel T,Maenosono R,Minami K,Nian Y,Quante M,Falk CS,Azuma H,Martin F,Passos JF,Niemann CU,Tchkonia T,Kirkland JL,Elkhal A,Tullius SG
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu Aug 27 00:00:00 UTC 2020
Salmonella-Based Therapy Targeting Indoleamine 2,3-Dioxygenase Restructures the Immune Contexture to Improve Checkpoint Blockade Efficacy.
Biomedicines
Ebelt ND,Zuniga E,Marzagalli M,Zamloot V,Blazar BR,Salgia R,Manuel ER
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Wed Dec 16 00:00:00 UTC 2020
Lactobacillus reuteri 5454 and Bifidobacterium animalis ssp. lactis 5764 improve colitis while differentially impacting dendritic cells maturation and antimicrobial responses.
Scientific reports
Hrdý J,Alard J,Couturier-Maillard A,Boulard O,Boutillier D,Delacre M,Lapadatescu C,Cesaro A,Blanc P,Pot B,Ryffel B,Chamaillard M,Grangette C
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Mar 24 00:00:00 UTC 2020
Tregitopes regulate the tolerogenic immune response and decrease the foetal death rate in abortion-prone mouse matings.
Scientific reports
Kedzierska AE,Lorek D,Slawek A,Chelmonska-Soyta A
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Jun 29 00:00:00 UTC 2020
Polyanhydride Nanoparticles Induce Low Inflammatory Dendritic Cell Activation Resulting in CD8+ T Cell Memory and Delayed Tumor Progression.
International journal of nanomedicine
Darling R,Senapati S,Christiansen J,Liu L,Ramer-Tait AE,Narasimhan B,Wannemuehler M
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Dec 07 00:00:00 UTC 2020
Epigenetically modulated FOXM1 suppresses dendritic cell maturation in pancreatic cancer and colon cancer.
Molecular oncology
Zhou Z,Chen H,Xie R,Wang H,Li S,Xu Q,Xu N,Cheng Q,Qian Y,Huang R,Shao Z,Xiang M
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Apr 01 00:00:00 UTC 2019
Immunological Properties of Neural Crest Cells Derived from Human Induced Pluripotent Stem Cells.
Stem cells and development
Fujii S,Yoshida S,Inagaki E,Hatou S,Tsubota K,Takahashi M,Shimmura S,Sugita S
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Jan 01 00:00:00 UTC 2019
CD1d highly expressed on DCs reduces lung tumor burden by enhancing antitumor immunity.
Oncology reports
Li Y,Zhao C,Liu J,Lu Z,Lu M,Gu J,Liu R
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Wed May 01 00:00:00 UTC 2019
cGAS/STING/TBK1/IRF3 Signaling Pathway Activates BMDCs Maturation Following Mycobacterium bovis Infection.
International journal of molecular sciences
Li Q,Liu C,Yue R,El-Ashram S,Wang J,He X,Zhao D,Zhou X,Xu L
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Feb 19 00:00:00 UTC 2019
Arp2/3 complex-driven spatial patterning of the BCR enhances immune synapse formation, BCR signaling and B cell activation.
eLife
Bolger-Munro M,Choi K,Scurll JM,Abraham L,Chappell RS,Sheen D,Dang-Lawson M,Wu X,Priatel JJ,Coombs D,Hammer JA,Gold MR
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Jun 03 00:00:00 UTC 2019
Mutation of an L-Type Calcium Channel Gene Leads to T Lymphocyte Dysfunction.
Frontiers in immunology
Fenninger F,Han J,Stanwood SR,Nohara LL,Arora H,Choi KB,Munro L,Pfeifer CG,Shanina I,Horwitz MS,Jefferies WA
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu Oct 29 00:00:00 UTC 2020
Efficient oral vaccination by bioengineering virus-like particles with protozoan surface proteins.
Nature communications
Serradell MC,Rupil LL,Martino RA,Prucca CG,Carranza PG,Saura A,Fernández EA,Gargantini PR,Tenaglia AH,Petiti JP,Tonelli RR,Reinoso-Vizcaino N,Echenique J,Berod L,Piaggio E,Bellier B,Sparwasser T,Klatzmann D,Luján HD
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Jan 21 00:00:00 UTC 2019
Doxorubicin-polyglycerol-nanodiamond conjugate is a cytostatic agent that evades chemoresistance and reverses cancer-induced immunosuppression in triple-negative breast cancer.
Journal of nanobiotechnology
Yuan SJ,Xu YH,Wang C,An HC,Xu HZ,Li K,Komatsu N,Zhao L,Chen X
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu Oct 17 00:00:00 UTC 2019
Cannabinoid 2 receptor attenuates inflammation during skin wound healing by inhibiting M1 macrophages rather than activating M2 macrophages.
Journal of inflammation (London, England)
Du Y,Ren P,Wang Q,Jiang SK,Zhang M,Li JY,Wang LL,Guan DW
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Immunohistochemistry
Wed Mar 30 00:00:00 UTC 2022
M1-Polarized Macrophages Promote Self-Renewing Phenotype of Hepatic Progenitor Cells with Jagged1-Notch Signalling Involved: Relevance in Primary Sclerosing Cholangitis.
Journal of immunology research
Li H,Sun S,Lei Q,Lei P,Cai X,Wan C,Shen G
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Apr 08 00:00:00 UTC 2019
Ginsenoside Rg3 Mitigates Atherosclerosis Progression in Diabetic apoE-/- Mice by Skewing Macrophages to the M2 Phenotype.
Frontiers in pharmacology
Guo M,Xiao J,Sheng X,Zhang X,Tie Y,Wang L,Zhao L,Ji X
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu Oct 01 00:00:00 UTC 2020
Targeting macrophage and microglia activation with colony stimulating factor 1 receptor inhibitor is an effective strategy to treat injury-triggered neuropathic pain.
Molecular pain
Lee S,Shi XQ,Fan A,West B,Zhang J
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Oct 22 00:00:00 UTC 2018
T cell microvilli constitute immunological synaptosomes that carry messages to antigen-presenting cells.
Nature communications
Kim HR,Mun Y,Lee KS,Park YJ,Park JS,Park JH,Jeon BN,Kim CH,Jun Y,Hyun YM,Kim M,Lee SM,Park CS,Im SH,Jun CD
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Fri Sep 07 00:00:00 UTC 2018
2-carba cyclic phosphatidic acid suppresses inflammation via regulation of microglial polarisation in the stab-wounded mouse cerebral cortex.
Scientific reports
Hashimoto K,Nakashima M,Hamano A,Gotoh M,Ikeshima-Kataoka H,Murakami-Murofushi K,Miyamoto Y
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Immunohistochemistry
Tue Jun 26 00:00:00 UTC 2018
Antibody Tumor Targeting Is Enhanced by CD27 Agonists through Myeloid Recruitment.
Cancer cell
Turaj AH,Hussain K,Cox KL,Rose-Zerilli MJJ,Testa J,Dahal LN,Chan HTC,James S,Field VL,Carter MJ,Kim HJ,West JJ,Thomas LJ,He LZ,Keler T,Johnson PWM,Al-Shamkhani A,Thirdborough SM,Beers SA,Cragg MS,Glennie MJ,Lim SH
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Dec 11 00:00:00 UTC 2017
FcεRI γ-Chain Negatively Modulates Dectin-1 Responses in Dendritic Cells.
Frontiers in immunology
Pan YG,Yu YL,Lin CC,Lanier LL,Chu CL
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Wed Nov 20 00:00:00 UTC 2019
IRF6 Regulates Alternative Activation by Suppressing PPARγ in Male Murine Macrophages.
Endocrinology
Li C,Ying W,Huang Z,Brehm T,Morin A,Vella AT,Zhou B
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Fri Sep 01 00:00:00 UTC 2017
Discrimination of the heterogeneity of bone marrow‑derived dendritic cells.
Molecular medicine reports
Wang J,Dai X,Hsu C,Ming C,He Y,Zhang J,Wei L,Zhou P,Wang CY,Yang J,Gong N
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Wed Nov 01 00:00:00 UTC 2017
Calreticulin is an effective immunologic adjuvant to tumor-associated antigens.
Experimental and therapeutic medicine
Wang J,Gao ZP,Qin S,Liu CB,Zou LL
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sun Oct 01 00:00:00 UTC 2017
Tumor-derived exosomes educate dendritic cells to promote tumor metastasis via HSP72/HSP105-TLR2/TLR4 pathway.
Oncoimmunology
Shen Y,Guo D,Weng L,Wang S,Ma Z,Yang Y,Wang P,Wang J,Cai Z
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Jan 12 00:00:00 UTC 2021
Batf3-dependent CD8α+ Dendritic Cells Aggravates Atherosclerosis via Th1 Cell Induction and Enhanced CCL5 Expression in Plaque Macrophages.
EBioMedicine
Li Y,Liu X,Duan W,Tian H,Zhu G,He H,Yao S,Yi S,Song W,Tang H
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sat Apr 01 00:00:00 UTC 2017
Paeoniflorin Ameliorates Experimental Autoimmune Encephalomyelitis via Inhibition of Dendritic Cell Function and Th17 Cell Differentiation.
Scientific reports
Zhang H,Qi Y,Yuan Y,Cai L,Xu H,Zhang L,Su B,Nie H
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Feb 06 00:00:00 UTC 2017
Repulsive guidance molecule a blockade exerts the immunoregulatory function in DCs stimulated with ABP and LPS.
Human vaccines & immunotherapeutics
Xu X,Gao Y,Zhai Z,Zhang S,Shan F,Feng J
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Aug 02 00:00:00 UTC 2016
Metformin Inhibits Advanced Glycation End Products-Induced Inflammatory Response in Murine Macrophages Partly through AMPK Activation and RAGE/NFκB Pathway Suppression.
Journal of diabetes research
Zhou Z,Tang Y,Jin X,Chen C,Lu Y,Liu L,Shen C
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Jun 06 00:00:00 UTC 2017
CD80 and CD86 knockdown in dendritic cells regulates Th1/Th2 cytokine production in asthmatic mice.
Experimental and therapeutic medicine
Li JG,DU YM,Yan ZD,Yan J,Zhuansun YX,Chen R,Zhang W,Feng SL,Ran PX
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Mar 01 00:00:00 UTC 2016
Temporal Characterization of Microglia/Macrophage Phenotypes in a Mouse Model of Neonatal Hypoxic-Ischemic Brain Injury.
Frontiers in cellular neuroscience
Hellström Erkenstam N,Smith PL,Fleiss B,Nair S,Svedin P,Wang W,Boström M,Gressens P,Hagberg H,Brown KL,Sävman K,Mallard C
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sat Jan 29 00:00:00 UTC 2022
Temporal Characterization of Microglia/Macrophage Phenotypes in a Mouse Model of Neonatal Hypoxic-Ischemic Brain Injury.
Frontiers in cellular neuroscience
Hellström Erkenstam N,Smith PL,Fleiss B,Nair S,Svedin P,Wang W,Boström M,Gressens P,Hagberg H,Brown KL,Sävman K,Mallard C
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sat Jan 29 00:00:00 UTC 2022
Irradiation of necrotic cancer cells, employed for pulsing dendritic cells (DCs), potentiates DC vaccine-induced antitumor immunity against high-grade glioma.
Oncoimmunology
Vandenberk L,Garg AD,Verschuere T,Koks C,Belmans J,Beullens M,Agostinis P,De Vleeschouwer S,Van Gool SW
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2016
D-Alanylation of Teichoic Acids and Loss of Poly-N-Acetyl Glucosamine in Staphylococcus aureus during Exponential Growth Phase Enhance IL-12 Production in Murine Dendritic Cells.
PloS one
Lund LD,Ingmer H,Frøkiær H
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu Jul 14 00:00:00 UTC 2016
Injectable, spontaneously assembling, inorganic scaffolds modulate immune cells in vivo and increase vaccine efficacy.
Nature biotechnology
Kim J,Li WA,Choi Y,Lewin SA,Verbeke CS,Dranoff G,Mooney DJ
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Thu Jan 01 00:00:00 UTC 2015
Towards programming immune tolerance through geometric manipulation of phosphatidylserine.
Biomaterials
Roberts RA,Eitas TK,Byrne JD,Johnson BM,Short PJ,McKinnon KP,Reisdorf S,Luft JC,DeSimone JM,Ting JP
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Dec 01 00:00:00 UTC 2015
Reducing progression of experimental lupus nephritis via inhibition of the B7/CD28 signaling pathway.
Molecular medicine reports
Huang L,Kong Y,Wang J,Sun J,Shi Q,Qiu YH
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2015
Injectable cryogel-based whole-cell cancer vaccines.
Nature communications
Bencherif SA,Warren Sands R,Ali OA,Li WA,Lewin SA,Braschler TM,Shih TY,Verbeke CS,Bhatta D,Dranoff G,Mooney DJ
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Wed Aug 12 00:00:00 UTC 2015
Nizatidine, a small molecular compound, enhances killed H5N1 vaccine cell-mediated responses and protects mice from lethal viral challenge.
Human vaccines & immunotherapeutics
Wang S,Wu B,Xue J,Wang M,Chen R,Wang B
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Mon Jan 05 00:00:00 UTC 2015
BCG vaccine mediated reduction in the MHC-II expression of macrophages and dendritic cells is reversed by activation of Toll-like receptors 7 and 9.
Cellular immunology
Bakhru P,Sirisaengtaksin N,Soudani E,Mukherjee S,Khan A,Jagannath C
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Wed Jan 01 00:00:00 UTC 2014
Lung dendritic cells undergo maturation and polarization towards a T helper type 2-stimulating phenotype in a mouse model of asthma: Role of nerve growth factor.
Experimental and therapeutic medicine
Qin Q,Wang Z,Pan P,Cao Z,Xia Q,Tan H,Hu C
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sat Nov 01 00:00:00 UTC 2014
Alkylglycerols modulate the proliferation and differentiation of non-specific agonist and specific antigen-stimulated splenic lymphocytes.
PloS one
Qian L,Zhang M,Wu S,Zhong Y,Van Tol E,Cai W
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Fri Jul 31 00:00:00 UTC 2015
A new animal model of spontaneous autoimmune peripheral polyneuropathy: implications for Guillain-Barré syndrome.
Acta neuropathologica communications
Yang M,Rainone A,Shi XQ,Fournier S,Zhang J
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Wed Jan 08 00:00:00 UTC 2014
Neutralization of TSLP inhibits airway remodeling in a murine model of allergic asthma induced by chronic exposure to house dust mite.
PloS one
Chen ZG,Zhang TT,Li HT,Chen FH,Zou XL,Ji JZ,Chen H
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Tue Aug 20 00:00:00 UTC 2013
Immunogenicity and protective capacity of a virosomal respiratory syncytial virus vaccine adjuvanted with monophosphoryl lipid A in mice.
PloS one
Kamphuis T,Meijerhof T,Stegmann T,Lederhofer J,Wilschut J,de Haan A
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Wed Sep 19 00:00:00 UTC 2012
Identification of regulatory functions for 4-1BB and 4-1BBL in myelopoiesis and the development of dendritic cells.
Nature immunology
Lee SW,Park Y,So T,Kwon BS,Cheroutre H,Mittler RS,Croft M
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Fri Aug 01 00:00:00 UTC 2008
Region-specific expression of immunoregulatory proteins on microglia in the healthy CNS.
Glia
de Haas AH,Boddeke HW,Biber K
Published figure using CD86 (B7-2) monoclonal antibody (Product # 56-0862-82) in Flow Cytometry
Sun Jun 01 00:00:00 UTC 2008