Product References
Immune response induced by standard and fractional doses of 17DD yellow fever vaccine.
NPJ vaccines
Abdala-Torres T,Campi-Azevedo AC,da Silva-Pereira RA,Dos Santos LI,Henriques PM,Costa-Rocha IA,Otta DA,Peruhype-Magalhães V,Teixeira-Carvalho A,Araújo MSS,Fernandes EG,Sato HK,Fantinato FFST,Domingues CMAS,Kallás EG,Tomiyama HTI,Lemos JAC,Coelho-Dos-Reis JG,de Lima SMB,Schwarcz WD,de Souza Azevedo A,Trindade GF,Ano Bom APD,da Silva AMV,Fernandes CB,Camacho LAB,de Sousa Maia ML,Martins-Filho OA,do Antonelli LRDV
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Fri Mar 08 00:00:00 UTC 2024
Protocol for isolation of tumor-derived extracellular vesicles and functional studies on human T cell subsets.
STAR protocols
Swatler J,Targońska A,Turos-Korgul L,Mosieniak G,Piwocka K
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Fri Jun 21 00:00:00 UTC 2024
BCG immunotherapy promotes tumor-derived T-cell activation through the FLT3/FLT3LG pathway in bladder cancer.
Journal of Cancer
Zhang W,Yu L,Chang Z,Xiong H
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Sat Jan 13 00:00:00 UTC 2024
Mucosal prime-boost immunization with live murine pneumonia virus-vectored SARS-CoV-2 vaccine is protective in macaques.
Nature communications
Kaiser JA,Nelson CE,Liu X,Park HS,Matsuoka Y,Luongo C,Santos C,Ahlers LRH,Herbert R,Moore IN,Wilder-Kofie T,Moore R,Walker A,Yang L,Munir S,Teng IT,Kwong PD,Dowdell K,Nguyen H,Kim J,Cohen JI,Johnson RF,Garza NL,Via LE,Barber DL,Buchholz UJ,Le Nouën C
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Fri Apr 26 00:00:00 UTC 2024
Endosialin-positive tumor-derived pericytes promote tumor progression through impeding the infiltration of CD8+ T cells in clear cell renal cell carcinoma.
Cancer immunology, immunotherapy : CII
Lu T,Zhang J,Lu S,Yang F,Gan L,Wu X,Song H,Liu S,Xu C,Han D,Yang B,Wen W,Qin W,Yang L
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Thu Jun 01 00:00:00 UTC 2023
Soluble NKG2D ligands impair CD8+ T cell antitumor function dependent of NKG2D downregulation in neuroblastoma.
Oncology letters
Zhang Y,Luo F,Dong K
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Sat Jul 01 00:00:00 UTC 2023
Prostate cancer cells synergistically defend against CD8+ T cells by secreting exosomal PD-L1.
Cancer medicine
Li D,Zhou X,Xu W,Chen Y,Mu C,Zhao X,Yang T,Wang G,Wei L,Ma B
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Tue Aug 01 00:00:00 UTC 2023
Rethinking IL-1 Antagonism in Respiratory Viral Infections: A Role for IL-1 Signaling in the Development of Antiviral T Cell Immunity.
International journal of molecular sciences
Van Den Eeckhout B,Ballegeer M,De Clercq J,Burg E,Saelens X,Vandekerckhove L,Gerlo S
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Mon Oct 30 00:00:00 UTC 2023
TCR ligand potency differentially impacts PD-1 inhibitory effects on diverse signaling pathways.
The Journal of experimental medicine
Chan W,Cao YM,Zhao X,Schrom EC,Jia D,Song J,Sibener LV,Dong S,Fernandes RA,Bradfield CJ,Smelkinson M,Kabat J,Hor JL,Altan-Bonnet G,Garcia KC,Germain RN
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Mon Dec 04 00:00:00 UTC 2023
Recurrent visceral leishmaniasis relapses in HIV co-infected patients are characterized by less efficient immune responses and higher parasite load.
iScience
Takele Y,Mulaw T,Adem E,Womersley R,Kaforou M,Franssen SU,Levin M,Taylor GP,Müller I,Cotton JA,Kropf P
25-0088-42 was used in Flow cytometry/Cell sorting to show that the relapse-free survival in patients with recurrent VL/HIV was shorter, that they had higher parasite load, lower weight gain, and lower recovery of all blood cell lineages.
Fri Feb 17 00:00:00 UTC 2023
A Novel Homozygous Stop Mutation in IL23R Causes Mendelian Susceptibility to Mycobacterial Disease.
Journal of clinical immunology
Staels F,Lorenzetti F,De Keukeleere K,Willemsen M,Gerbaux M,Neumann J,Tousseyn T,Pasciuto E,De Munter P,Bossuyt X,Gijsbers R,Liston A,Humblet-Baron S,Schrijvers R
25-0088-42 was used in Flow cytometry/Cell sorting to demonstrate that impaired IL-23 immunity caused by a homozygous R381X mutation in IL23R underlies MSMD, corroborating earlier findings with a homozygous p.C115Y IL23R mutation.
Tue Nov 01 00:00:00 UTC 2022
VLA-4 suppression by senescence signals regulates meningeal immunity and leptomeningeal metastasis.
eLife
Li J,Huang D,Lei B,Huang J,Yang L,Nie M,Su S,Zhao Q,Wang Y
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Fri Dec 09 00:00:00 UTC 2022
PRDM1/BLIMP1 induces cancer immune evasion by modulating the USP22-SPI1-PD-L1 axis in hepatocellular carcinoma cells.
Nature communications
Li Q,Zhang L,You W,Xu J,Dai J,Hua D,Zhang R,Yao F,Zhou S,Huang W,Dai Y,Zhang Y,Baheti T,Qian X,Pu L,Xu J,Xia Y,Zhang C,Tang J,Wang X
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Mon Dec 12 00:00:00 UTC 2022
Screening of an individualized treatment strategy for an advanced gallbladder cancer using patient-derived tumor xenograft and organoid models.
Frontiers in oncology
Tan D,An J,Gong M,Wang H,Li H,Meng H,Zhang C,Zhao Y,Ge X,Shi C
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Tue Jan 03 00:00:00 UTC 2023
Immuno-oncological role of 20S proteasome alpha-subunit 3 in aggravating the progression of esophageal squamous cell carcinoma.
European journal of immunology
Liu J,Shao J,Zhang C,Qin G,Liu J,Li M,Wu P,Zhao X,Zhang Y
25-0088-42 was used in Flow cytometry/Cell sorting to demonstrate the role of PSMA3 in ESCC cancer stemness and the negative regulation of CD8 T-cells infiltration mediated by PSMA3 and provide a potential target for the immuno-oncology effect of PSMA3 in ESCC therapy.
Tue Feb 01 00:00:00 UTC 2022
Immunological factors, but not clinical features, predict visceral leishmaniasis relapse in patients co-infected with HIV.
Cell reports. Medicine
Takele Y,Mulaw T,Adem E,Shaw CJ,Franssen SU,Womersley R,Kaforou M,Taylor GP,Levin M,Müller I,Cotton JA,Kropf P
25-0088-42 was used in Flow Cytometry to identify three immunological markers associated with visceral leishmaniasis relapse in VL/HIV patients.
Tue Jan 18 00:00:00 UTC 2022
Severe T cell hyporeactivity in ventilated COVID-19 patients correlates with prolonged virus persistence and poor outcomes.
Nature communications
Renner K,Schwittay T,Chaabane S,Gottschling J,Müller C,Tiefenböck C,Salewski JN,Winter F,Buchtler S,Balam S,Malfertheiner MV,Lubnow M,Lunz D,Graf B,Hitzenbichler F,Hanses F,Poeck H,Kreutz M,Orsó E,Burkhardt R,Niedermair T,Brochhausen C,Gessner A,Salzberger B,Mack M
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Fri May 21 00:00:00 UTC 2021
Ex vivo expansion of dysfunctional regulatory T lymphocytes restores suppressive function in Parkinson's disease.
NPJ Parkinson's disease
Thome AD,Atassi F,Wang J,Faridar A,Zhao W,Thonhoff JR,Beers DR,Lai EC,Appel SH
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Thu May 13 00:00:00 UTC 2021
Multipeptide stimulated PBMCs generate TEM/TCM for adoptive cell therapy in multiple myeloma.
Oncotarget
Vardam-Kaur T,Pathangey LB,McCormick DJ,Bergsagel PL,Cohen PA,Gendler SJ
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Tue Sep 28 00:00:00 UTC 2021
Human endoglin-CD3 bispecific T cell engager antibody induces anti-tumor effect in vivo.
Theranostics
Zhong L,Shi W,Gan L,Liu X,Huo Y,Wu P,Zhang Z,Wu T,Peng H,Huang Y,Zhao Y,Yuan Y,Deng Z,Tang H
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Wed Jul 28 00:00:00 UTC 2021
Bispecific T-Cell Engaging Antibodies Against MUC16 Demonstrate Efficacy Against Ovarian Cancer in Monotherapy and in Combination With PD-1 and VEGF Inhibition.
Frontiers in immunology
Yeku OO,Rao TD,Laster I,Kononenko A,Purdon TJ,Wang P,Cui Z,Liu H,Brentjens RJ,Spriggs D
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Thu Oct 21 00:00:00 UTC 2021
Preventing ATP Degradation by ASO-Mediated Knockdown of CD39 and CD73 Results in A2aR-Independent Rescue of T Cell Proliferation.
Molecular therapy. Nucleic acids
Festag J,Thelemann T,Schell M,Raith S,Michel S,Jaschinski F,Klar R
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Fri Sep 04 00:00:00 UTC 2020
A Cross-Reactive Small Protein Binding Domain Provides a Model to Study Off-Tumor CAR-T Cell Toxicity.
Molecular therapy oncolytics
Hammill JA,Kwiecien JM,Dvorkin-Gheva A,Lau VWC,Baker C,Wu Y,Bezverbnaya K,Aarts C,Heslen CW,Denisova GF,Derocher H,Milne K,Nelson BH,Bramson JL
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Fri Jun 26 00:00:00 UTC 2020
The TCR Repertoire Reconstitution in Multiple Sclerosis: Comparing One-Shot and Continuous Immunosuppressive Therapies.
Frontiers in immunology
Amoriello R,Greiff V,Aldinucci A,Bonechi E,Carnasciali A,Peruzzi B,Repice AM,Mariottini A,Saccardi R,Mazzanti B,Massacesi L,Ballerini C
25-0088-42 was used in Flow Cytometry to suggest that the majority of public TCRs belonged to virus-associated sequences.
Fri Mar 26 00:00:00 UTC 2021
Immunological Properties of Neural Crest Cells Derived from Human Induced Pluripotent Stem Cells.
Stem cells and development
Fujii S,Yoshida S,Inagaki E,Hatou S,Tsubota K,Takahashi M,Shimmura S,Sugita S
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Tue Jan 01 00:00:00 UTC 2019
Intracellular delivery of mRNA to human primary T cells with microfluidic vortex shedding.
Scientific reports
Jarrell JA,Twite AA,Lau KHWJ,Kashani MN,Lievano AA,Acevedo J,Priest C,Nieva J,Gottlieb D,Pawell RS
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Fri Mar 01 00:00:00 UTC 2019
Niraparib activates interferon signaling and potentiates anti-PD-1 antibody efficacy in tumor models.
Scientific reports
Wang Z,Sun K,Xiao Y,Feng B,Mikule K,Ma X,Feng N,Vellano CP,Federico L,Marszalek JR,Mills GB,Hanke J,Ramaswamy S,Wang J
25-0088-42 was used in Flow Cytometry to investigate the therapeutic potential of combining niraparib with anti-P-1 immune checkpoint inhibitors in preclinical tumour models.
Tue Feb 12 00:00:00 UTC 2019
STAT3 Inhibition Combined with CpG Immunostimulation Activates Antitumor Immunity to Eradicate Genetically Distinct Castration-Resistant Prostate Cancers.
Clinical cancer research : an official journal of the American Association for Cancer Research
Moreira D,Adamus T,Zhao X,Su YL,Zhang Z,White SV,Swiderski P,Lu X,DePinho RA,Pal SK,Kortylewski M
25-0088 was used in Flow cytometry/Cell sorting to show a novel strategy combining STAT3 inhibition with Toll-like Receptor 9 (TLR9) stimulation to unleash immune response against prostate cancers regardless of the genetic background.
Sat Dec 01 00:00:00 UTC 2018
Blocking the recruitment of naive CD4+ T cells reverses immunosuppression in breast cancer.
Cell research
Su S,Liao J,Liu J,Huang D,He C,Chen F,Yang L,Wu W,Chen J,Lin L,Zeng Y,Ouyang N,Cui X,Yao H,Su F,Huang JD,Lieberman J,Liu Q,Song E
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Sat Apr 01 00:00:00 UTC 2017
Two separate effects contribute to regulatory T cell defect in systemic lupus erythematosus patients and their unaffected relatives.
Clinical and experimental immunology
Costa N,Marques O,Godinho SI,Carvalho C,Leal B,Figueiredo AM,Vasconcelos C,Marinho A,Moraes-Fontes MF,Gomes da Costa A,Ponte C,Campanilho-Marques R,Cóias T,Martins AR,Viana JF,Lima M,Martins B,Fesel C
25-0088 was used in Flow cytometry/Cell sorting to determine the origin and role of the SLE Treg phenotype in systemic lupus erythematosus.
Fri Sep 01 00:00:00 UTC 2017
Detection of Retinal Pigment Epithelium-Specific Antibody in iPSC-Derived Retinal Pigment Epithelium Transplantation Models.
Stem cell reports
Sugita S,Makabe K,Fujii S,Iwasaki Y,Kamao H,Shiina T,Ogasawara K,Takahashi M
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Tue Nov 14 00:00:00 UTC 2017
Follicular Regulatory CD8 T Cells Impair the Germinal Center Response in SIV and Ex Vivo HIV Infection.
PLoS pathogens
Miles B,Miller SM,Folkvord JM,Levy DN,Rakasz EG,Skinner PJ,Connick E
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Sat Oct 01 00:00:00 UTC 2016
Human Head and Neck Squamous Cell Carcinoma-Associated Semaphorin 4D Induces Expansion of Myeloid-Derived Suppressor Cells.
Journal of immunology (Baltimore, Md. : 1950)
Younis RH,Han KL,Webb TJ
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2016
Host immunity dictates influenza A(H1N1)pdm09 infection outcome in hematology-oncology patients.
Bone marrow transplantation
Gooskens J,Marijt WA,van Essen EH,Rimmelzwaan GF,Kroes AC
25-0088-42 was used in Flow Cytometry to evaluate the role of innate and adaptive host immune findings in determining virus-specific symptoms and viral clearance among hematology-oncology patients with prolonged viral excretion.
Fri Jan 01 00:00:00 UTC 2016
Host immunity dictates influenza A(H1N1)pdm09 infection outcome in hematology-oncology patients.
Bone marrow transplantation
Gooskens J,Marijt WA,van Essen EH,Rimmelzwaan GF,Kroes AC
25-0088-42 was used in Flow Cytometry to evaluate the role of innate and adaptive host immune findings in determining virus-specific symptoms and viral clearance among hematology-oncology patients with prolonged viral excretion.
Fri Jan 01 00:00:00 UTC 2016
Umbilical Cord-Derived Mesenchymal Stem Cells Suppress Autophagy of T Cells in Patients with Systemic Lupus Erythematosus via Transfer of Mitochondria.
Stem cells international
Chen J,Wang Q,Feng X,Zhang Z,Geng L,Xu T,Wang D,Sun L
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Thu Oct 01 00:00:00 UTC 2020
CD28 family of receptors on T cells in chronic HBV infection: Expression characteristics, clinical significance and correlations with PD-1 blockade.
Molecular medicine reports
Tang ZS,Hao YH,Zhang EJ,Xu CL,Zhou Y,Zheng X,Yang DL
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2016
Lack of T Cell Response to iPSC-Derived Retinal Pigment Epithelial Cells from HLA Homozygous Donors.
Stem cell reports
Sugita S,Iwasaki Y,Makabe K,Kimura T,Futagami T,Suegami S,Takahashi M
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Tue Oct 11 00:00:00 UTC 2016
B cells from African American lupus patients exhibit an activated phenotype.
JCI insight
Menard LC,Habte S,Gonsiorek W,Lee D,Banas D,Holloway DA,Manjarrez-Orduno N,Cunningham M,Stetsko D,Casano F,Kansal S,Davis PM,Carman J,Zhang CK,Abidi F,Furie R,Nadler SG,Suchard SJ
25-0088 was used in Flow cytometry/Cell sorting to investigate immunological differences between African American and European American patients with systemic lupus erythematosus, showing that B cells from African American patients exhibit an activated phenotype.
Thu Jun 16 00:00:00 UTC 2016
Liposomal Glutathione Supplementation Restores TH1 Cytokine Response to Mycobacterium tuberculosis Infection in HIV-Infected Individuals.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research
Ly J,Lagman M,Saing T,Singh MK,Tudela EV,Morris D,Anderson J,Daliva J,Ochoa C,Patel N,Pearce D,Venketaraman V
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Sun Nov 01 00:00:00 UTC 2015
Follicular regulatory T cells impair follicular T helper cells in HIV and SIV infection.
Nature communications
Miles B,Miller SM,Folkvord JM,Kimball A,Chamanian M,Meditz AL,Arends T,McCarter MD,Levy DN,Rakasz EG,Skinner PJ,Connick E
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Tue Oct 20 00:00:00 UTC 2015
RSV-specific airway resident memory CD8+ T cells and differential disease severity after experimental human infection.
Nature communications
Jozwik A,Habibi MS,Paras A,Zhu J,Guvenel A,Dhariwal J,Almond M,Wong EHC,Sykes A,Maybeno M,Del Rosario J,Trujillo-Torralbo MB,Mallia P,Sidney J,Peters B,Kon OM,Sette A,Johnston SL,Openshaw PJ,Chiu C
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Mon Dec 21 00:00:00 UTC 2015
Investigating the causes for decreased levels of glutathione in individuals with type II diabetes.
PloS one
Lagman M,Ly J,Saing T,Kaur Singh M,Vera Tudela E,Morris D,Chi PT,Ochoa C,Sathananthan A,Venketaraman V
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Wed Dec 30 00:00:00 UTC 2015
Accumulation of 4-1BBL+ B cells in the elderly induces the generation of granzyme-B+ CD8+ T cells with potential antitumor activity.
Blood
Lee-Chang C,Bodogai M,Moritoh K,Olkhanud PB,Chan AC,Croft M,Mattison JA,Holst PJ,Gress RE,Ferrucci L,Hakim F,Biragyn A
25-0088 was used in Flow cytometry/Cell sorting to propose that 4BL cells and the 4-1BBL signaling pathway are useful targets for improved effectiveness of natural antitumor defenses and therapeutic immune manipulations in the elderly.
Thu Aug 28 00:00:00 UTC 2014
Immune response to JC virus T antigen in patients with and without colorectal neoplasia.
Gut microbes
Butcher LD,Garcia M,Arnold M,Ueno H,Goel A,Boland CR
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Tue Jul 01 00:00:00 UTC 2014
CD4+CD25(high), CD8+CD28- cells and thyroid autoantibodies in breast cancer patients.
Central-European journal of immunology
Biylgi O,Karagöz B,Türken O,Gültepe M,Özgün A,Tunçel T,Emirzeoğlu L,Çelik S,Müftüoğlu T,Gökhan Kandemir E
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Thu Jul 09 00:00:00 UTC 2015
Ex vivo analysis of resident hepatic pro-inflammatory CD1d-reactive T cells and hepatocyte surface CD1d expression in hepatitis C.
Journal of viral hepatitis
Yanagisawa K,Yue S,van der Vliet HJ,Wang R,Alatrakchi N,Golden-Mason L,Schuppan D,Koziel MJ,Rosen HR,Exley MA
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Thu Aug 01 00:00:00 UTC 2013
Galectin-9 activates and expands human T-helper 1 cells.
PloS one
Gooden MJ,Wiersma VR,Samplonius DF,Gerssen J,van Ginkel RJ,Nijman HW,Hirashima M,Niki T,Eggleton P,Helfrich W,Bremer E
25-0088 was used in Flow cytometry/Cell sorting to demonstrate that galectin-9 activates resting T-cells and promotes their transition to a T-helper 1 cell phenotype.
Fri Jan 10 00:00:00 UTC 2014
Upregulation of programmed death-1 on T cells and programmed death ligand-1 on monocytes in septic shock patients.
Critical care (London, England)
Zhang Y,Li J,Lou J,Zhou Y,Bo L,Zhu J,Zhu K,Wan X,Cai Z,Deng X
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Wed Dec 07 00:00:00 UTC 2011
Purification and direct transformation of epithelial progenitor cells from primary human prostate.
Nature protocols
Goldstein AS,Drake JM,Burnes DL,Finley DS,Zhang H,Reiter RE,Huang J,Witte ON
25-0088 was used in Flow cytometry/Cell sorting to describe a system to directly transform freshly isolated primary human prostate epithelial cells without previous culture or immortalisation.
Sun May 01 00:00:00 UTC 2011
Sepsis-induced human lymphocyte apoptosis and cytokine production in "humanized" mice.
Journal of leukocyte biology
Unsinger J,McDonough JS,Shultz LD,Ferguson TA,Hotchkiss RS
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Sat Aug 01 00:00:00 UTC 2009
B cells and monocytes from patients with active multiple sclerosis exhibit increased surface expression of both HERV-H Env and HERV-W Env, accompanied by increased seroreactivity.
Retrovirology
Brudek T,Christensen T,Aagaard L,Petersen T,Hansen HJ,Møller-Larsen A
Published figure using CD8a monoclonal antibody (Product # 25-0088-42) in Flow Cytometry
Mon Nov 16 00:00:00 UTC 2009