Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 8
The epitope recognized by this antibody is reported to be sensitive to formaldehyde fixation and tissue processing. We recommend the use of acetone fixation for frozen sections. A suggested positive control for immunohistochemical applications is human brain or thymus. For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL.
Mouse anti Human CD90 antibody, clone F15-42-1 recognizes the human CD90 cell surface antigen, a approximately 25 kDa glycoprotein homologous to rat Thy1.
Mouse anti Human CD90 antibody, clone F15-42-1 recognizes the human CD90 cell surface antigen, a approximately 25 kDa glycoprotein homologous to rat Thy1.
CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. CD90 is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. CD90 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain. CD90 can be used as a marker for a variety of stem cells and for the axonal processes of mature neurons. Structural study of CD90 led to the foundation of the Immunoglobulin superfamily, of which it is the smallest member, and led to the first biochemical description and characterization of a vertebrate GPI anchor. Diseases associated with CD90 dysfunction include nasopharyngeal carcinoma and thymoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD90; CDw90; FLJ33325; Thy-1 antigen; Thy-1 membrane glycoprotein; Thy-1 T-cell antigen
Gene Aliases: CD90; CDw90; THY1
UniProt ID: (Human) P04216
Entrez Gene ID: (Human) 7070, (Cynomolgus monkey) 102139037
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support