Product References
Osteogenic potential of apical papilla stem cells mediated by platelet-rich fibrin and low-level laser.
Odontology
Ramírez DG,Inostroza C,Rouabhia M,Rodriguez CA,Gómez LA,Losada M,Muñoz AL
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Mon Apr 01 00:00:00 UTC 2024
CD304 + adipose tissue-derived mesenchymal stem cell abundance in autologous fat grafts highly correlates with improvement of localized pain syndromes.
Pain
Rezzonico Jost T,Lozito A,Mangani D,Raimondi A,Klinger F,Morone D,Klinger M,Grassi F,Vinci V
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Mon Apr 01 00:00:00 UTC 2024
Stimulated Human Umbilical Cord Mesenchymal Stem Cells Enhance the Osteogenesis and Cranial Bone Regeneration through IL-32 Mediated P38 Signaling Pathway.
Stem cells international
Zhang X,Zheng Y,Wang G,Liu Y,Wang Y,Jiang X,Liang Y,Zhao X,Li P,Zhang Y
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Fri Mar 22 00:00:00 UTC 2024
Generation of Highly Functional Hepatocyte-like Organoids from Human Adipose-Derived Mesenchymal Stem Cells Cultured with Endothelial Cells.
Cells
Chen S,Saito Y,Waki Y,Ikemoto T,Teraoku H,Yamada S,Morine Y,Shimada M
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Wed Mar 20 00:00:00 UTC 2024
Adipose-derived stem cell exosomes promote tumor characterization and immunosuppressive microenvironment in breast cancer.
Cancer immunology, immunotherapy : CII
Zhu Q,Cao Y,Yuan J,Hu Y
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Wed Jan 31 00:00:00 UTC 2024
Umbilical cord mesenchymal stem cell-derived apoptotic extracellular vesicles ameliorate cutaneous wound healing in type 2 diabetic mice via macrophage pyroptosis inhibition.
Stem cell research & therapy
Wang Y,Jing L,Lei X,Ma Z,Li B,Shi Y,Zhang W,Li Y,Zhou H,Hu K,Xue Y,Jin Y
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Tue Sep 19 00:00:00 UTC 2023
Fluorescence-activated cell sorting and phenotypic characterization of human fibro-adipogenic progenitors.
STAR protocols
Billeskov TB,Jensen JB,Jessen N,Farup J
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Fri Mar 17 00:00:00 UTC 2023
Exercise combined with administration of adipose-derived stem cells ameliorates neuropathic pain after spinal cord injury.
Neural regeneration research
Cheng X,Mao GP,Hu WJ,Yu ZR,Xu YY,Chen W,Li X,Zeng XL,Zhang WW,Chen JW,Wan Y,Wang L
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Tue Aug 01 00:00:00 UTC 2023
Engineered cardiac tissue model of restrictive cardiomyopathy for drug discovery.
Cell reports. Medicine
Wang BZ,Nash TR,Zhang X,Rao J,Abriola L,Kim Y,Zakharov S,Kim M,Luo LJ,Morsink M,Liu B,Lock RI,Fleischer S,Tamargo MA,Bohnen M,Welch CL,Chung WK,Marx SO,Surovtseva YV,Vunjak-Novakovic G,Fine BM
11-0909-42 was used in Flow cytometry/Cell sorting to demonstrate an engineered cardiac tissue model of RCM and establish the translational potential of this precision medicine approach to identify therapeutics targeting myocardial relaxation.
Tue Mar 21 00:00:00 UTC 2023
Exosomes derived from stem cells of human deciduous exfoliated teeth inhibit angiogenesis in vivo and in vitro via the transfer of miR-100-5p and miR-1246.
Stem cell research & therapy
Liu P,Zhang Q,Mi J,Wang S,Xu Q,Zhuang D,Chen W,Liu C,Zhang L,Guo J,Wu X
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Thu Mar 03 00:00:00 UTC 2022
Aberrant Expression of COX-2 and FOXG1 in Infrapatellar Fat Pad-Derived ASCs from Pre-Diabetic Donors.
Cells
O'Donnell BT,Monjure TA,Al-Ghadban S,Ives CJ,L'Ecuyer MP,Rhee C,Romero-Lopez M,Li Z,Goodman SB,Lin H,Tuan RS,Bunnell BA
11-0909-42 was used in Flow cytometry/Cell sorting to analyze the expression profiles of inflammatory and adipokine-related genes in infrapatellar fat pad-Adipose stem cells of non-diabetic (Non-T2D), pre-diabetic (Pre-T2D), and T2D donors.
Mon Aug 01 00:00:00 UTC 2022
Aberrant Expression of COX-2 and FOXG1 in Infrapatellar Fat Pad-Derived ASCs from Pre-Diabetic Donors.
Cells
O'Donnell BT,Monjure TA,Al-Ghadban S,Ives CJ,L'Ecuyer MP,Rhee C,Romero-Lopez M,Li Z,Goodman SB,Lin H,Tuan RS,Bunnell BA
11-0909-42 was used in Flow cytometry/Cell sorting to analyze the expression profiles of inflammatory and adipokine-related genes in infrapatellar fat pad-Adipose stem cells of non-diabetic (Non-T2D), pre-diabetic (Pre-T2D), and T2D donors.
Mon Aug 01 00:00:00 UTC 2022
LIM Mineralization Protein-1 Enhances the Committed Differentiation of Dental Pulp Stem Cells through the ERK1/2 and p38 MAPK Pathways and BMP Signaling.
International journal of medical sciences
Mu R,Chen B,Bi B,Yu H,Liu J,Li J,He M,Rong L,Liu B,Liu K,Zhu L,Shi X,Shuai Y,Jin L
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Mon Aug 08 00:00:00 UTC 2022
Fused Cells between Human-Adipose-Derived Mesenchymal Stem Cells and Monocytes Keep Stemness Properties and Acquire High Mobility.
International journal of molecular sciences
Montalbán-Hernández K,Casado-Sánchez C,Avendaño-Ortiz J,Casalvilla-Dueñas JC,Bonel-Pérez GC,Prado-Montero J,Valentín-Quiroga J,Lozano-Rodríguez R,Terrón-Arcos V,de la Bastida FR,Córdoba L,Laso-García F,Diekhorst L,Del Fresno C,López-Collazo E
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Fri Aug 26 00:00:00 UTC 2022
Mechanical Compression by Simulating Orthodontic Tooth Movement in an In Vitro Model Modulates Phosphorylation of AKT and MAPKs via TLR4 in Human Periodontal Ligament Cells.
International journal of molecular sciences
Roth CE,Craveiro RB,Niederau C,Malyaran H,Neuss S,Jankowski J,Wolf M
11-0909-42 was used in Flow cytometry/Cell sorting to better understand the cell signaling downstream of key molecules involved in the process of sterile inflammation via TLR4.
Fri Jul 22 00:00:00 UTC 2022
Short-Term Autophagy Preconditioning Upregulates the Expression of COX2 and PGE2 and Alters the Immune Phenotype of Human Adipose-Derived Stem Cells In Vitro.
Cells
Wise RM,Al-Ghadban S,Harrison MAA,Sullivan BN,Monaco ER,Aleman SJ,Donato UM,Bunnell BA
11-0909-42 was used in Flow cytometry/Cell sorting to suggest that a 4-h Rapa preconditioning strategy may bestow the greatest improvement to hASC expression of cytokines known to promote tissue repair and regeneration and may hold promise for augmenting the therapeutic potential of hASCs for inflammation-driven pathological conditions.
Tue Apr 19 00:00:00 UTC 2022
STK25 inhibits PKA signaling by phosphorylating PRKAR1A.
Cell reports
Zhang X,Wang BZ,Kim M,Nash TR,Liu B,Rao J,Lock R,Tamargo M,Soni RK,Belov J,Li E,Vunjak-Novakovic G,Fine B
11-0909-42 was used in Flow cytometry/Cell sorting to identify STK25 as a kinase of the type Iα regulatory subunit PRKAR1A, using human induced pluripotent stem cell cardiomyocytes (iPSC-CMs).
Tue Aug 16 00:00:00 UTC 2022
Cell-mimicking nanodecoys neutralize SARS-CoV-2 and mitigate lung injury in a non-human primate model of COVID-19.
Nature nanotechnology
Li Z,Wang Z,Dinh PC,Zhu D,Popowski KD,Lutz H,Hu S,Lewis MG,Cook A,Andersen H,Greenhouse J,Pessaint L,Lobo LJ,Cheng K
11-0909-42 was used in Immunocytochemistry to investigate if angiotensin-converting enzyme 2 nanodecoys derived from human lung spheroid cells (LSCs) can bind and neutralize SARS-CoV-2 and protect the host lung cells from infection.
Sun Aug 01 00:00:00 UTC 2021
Cell-mimicking nanodecoys neutralize SARS-CoV-2 and mitigate lung injury in a non-human primate model of COVID-19.
Nature nanotechnology
Li Z,Wang Z,Dinh PC,Zhu D,Popowski KD,Lutz H,Hu S,Lewis MG,Cook A,Andersen H,Greenhouse J,Pessaint L,Lobo LJ,Cheng K
11-0909-42 was used in Immunocytochemistry to investigate if angiotensin-converting enzyme 2 nanodecoys derived from human lung spheroid cells (LSCs) can bind and neutralize SARS-CoV-2 and protect the host lung cells from infection.
Sun Aug 01 00:00:00 UTC 2021
VAP-PLGA microspheres (VAP-PLGA) promote adipose-derived stem cells (ADSCs)-induced wound healing in chronic skin ulcers in mice via PI3K/Akt/HIF-1α pathway.
Bioengineered
Jiang W,Zhang J,Zhang X,Fan C,Huang J
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Wed Dec 01 00:00:00 UTC 2021
Exosomes Regulate Interclonal Communication on Osteogenic Differentiation Among Heterogeneous Osteogenic Single-Cell Clones Through PINK1/Parkin-Mediated Mitophagy.
Frontiers in cell and developmental biology
Fei D,Xia Y,Zhai Q,Wang Y,Zhou F,Zhao W,He X,Wang Q,Jin Y,Li B
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Tue Oct 05 00:00:00 UTC 2021
TRPA1 triggers hyperalgesia and inflammation after tooth bleaching.
Scientific reports
Chen C,Huang X,Zhu W,Ding C,Huang P,Li R
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Tue Aug 31 00:00:00 UTC 2021
In Vitro Anti-cancer Activity of Adipose-Derived Mesenchymal Stem Cells Increased after Infection with Oncolytic Reovirus.
Advanced pharmaceutical bulletin
Babaei A,Bannazadeh Baghi H,Nezhadi A,Jamalpoor Z
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2021
Therapeutic potential of human umbilical cord mesenchymal stem cells on aortic atherosclerotic plaque in a high-fat diet rabbit model.
Stem cell research & therapy
Li Y,Shi G,Han Y,Shang H,Li H,Liang W,Zhao W,Bai L,Qin C
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Thu Jul 15 00:00:00 UTC 2021
Down-Regulated Exosomal MicroRNA-221 - 3p Derived From Senescent Mesenchymal Stem Cells Impairs Heart Repair.
Frontiers in cell and developmental biology
Sun L,Zhu W,Zhao P,Zhang J,Lu Y,Zhu Y,Zhao W,Liu Y,Chen Q,Zhang F
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Mon Sep 28 00:00:00 UTC 2020
Differentiation Potential of Early- and Late-Passage Adipose-Derived Mesenchymal Stem Cells Cultured under Hypoxia and Normoxia.
Stem cells international
Zhao AG,Shah K,Freitag J,Cromer B,Sumer H
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Tue Oct 06 00:00:00 UTC 2020
Explant culture: A relevant tool for the study of telocytes.
Cell biology international
Sanches BDA,Maldarine JDS,Tamarindo GH,Da Silva ADT,Lima MLD,Rahal P,Góes RM,Taboga SR,Carvalho HF
11-0909-42 was used in Immunocytochemistry to elucidate the nature of telocytes and their functions.
Tue Dec 01 00:00:00 UTC 2020
Explant culture: A relevant tool for the study of telocytes.
Cell biology international
Sanches BDA,Maldarine JDS,Tamarindo GH,Da Silva ADT,Lima MLD,Rahal P,Góes RM,Taboga SR,Carvalho HF
11-0909-42 was used in Immunocytochemistry to elucidate the nature of telocytes and their functions.
Tue Dec 01 00:00:00 UTC 2020
Effects of CD4+ T lymphocytes from ovariectomized mice on bone marrow mesenchymal stem cell proliferation and osteogenic differentiation.
Experimental and therapeutic medicine
Shao BY,Wang L,Yu Y,Chen L,Gan N,Huang WM
11-0909 was used in Flow cytometry/Cell sorting to investigate the effects of T cells on the proliferation and osteogenic differentiation of bone marrow mesenchymal stem cells (BMMSCs).
Sun Nov 01 00:00:00 UTC 2020
Fibroblast growth factor-4 maintains cellular viability while enhancing osteogenic differentiation of stem cell spheroids in part by regulating RUNX2 and BGLAP expression.
Experimental and therapeutic medicine
Son J,Tae JY,Min SK,Ko Y,Park JB
11-0909 was used in Flow cytometry/Cell sorting to determine that the application of FGF-4 maintained cellular viability while enhancing the osteogenic differentiation of stem cell spheroids.
Tue Sep 01 00:00:00 UTC 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Fri Apr 03 00:00:00 UTC 2020
The Effect of Angiotensin II, Retinoic Acid, EGCG, and Vitamin C on the Cardiomyogenic Differentiation Induction of Human Amniotic Fluid-Derived Mesenchymal Stem Cells.
International journal of molecular sciences
Gasiūnienė M,Valatkaitė E,Navakauskaitė A,Navakauskienė R
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Thu Nov 19 00:00:00 UTC 2020
The Effect of Angiotensin II, Retinoic Acid, EGCG, and Vitamin C on the Cardiomyogenic Differentiation Induction of Human Amniotic Fluid-Derived Mesenchymal Stem Cells.
International journal of molecular sciences
Gasiūnienė M,Valatkaitė E,Navakauskaitė A,Navakauskienė R
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Thu Nov 19 00:00:00 UTC 2020
Increase in Leptin and PPAR-γ Gene Expression in Lipedema Adipocytes Differentiated in vitro from Adipose-Derived Stem Cells.
Cells
Al-Ghadban S,Diaz ZT,Singer HJ,Mert KB,Bunnell BA
11-0909-42 was used in Flow cytometry/Cell sorting to characterize the adipose-derived stem cells of healthy and lipedema patients by the expression of stemness markers and the adipogenic and osteogenic differentiation potential.
Wed Feb 12 00:00:00 UTC 2020
BNC1 regulates cell heterogeneity in human pluripotent stem cell-derived epicardium.
Development (Cambridge, England)
Gambardella L,McManus SA,Moignard V,Sebukhan D,Delaune A,Andrews S,Bernard WG,Morrison MA,Riley PR,Göttgens B,Gambardella Le Novère N,Sinha S
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Fri Dec 13 00:00:00 UTC 2019
BNC1 regulates cell heterogeneity in human pluripotent stem cell-derived epicardium.
Development (Cambridge, England)
Gambardella L,McManus SA,Moignard V,Sebukhan D,Delaune A,Andrews S,Bernard WG,Morrison MA,Riley PR,Göttgens B,Gambardella Le Novère N,Sinha S
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Fri Dec 13 00:00:00 UTC 2019
BNC1 regulates cell heterogeneity in human pluripotent stem cell-derived epicardium.
Development (Cambridge, England)
Gambardella L,McManus SA,Moignard V,Sebukhan D,Delaune A,Andrews S,Bernard WG,Morrison MA,Riley PR,Göttgens B,Gambardella Le Novère N,Sinha S
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Fri Dec 13 00:00:00 UTC 2019
LPS‑induced upregulation of the TLR4 signaling pathway inhibits osteogenic differentiation of human periodontal ligament stem cells under inflammatory conditions.
International journal of molecular medicine
Yu B,Li Q,Zhou M
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Sat Jun 01 00:00:00 UTC 2019
Spontaneous Extracellular Matrix Accumulation in a Human in vitro Model of Renal Fibrosis Is Mediated by αV Integrins.
Nephron
Bon H,Hales P,Lumb S,Holdsworth G,Johnson T,Qureshi O,Twomey BM
11-0909 was used in Flow cytometry/Cell sorting to test the hypothesis that integrins are pro-fibrotic via regulation of functional interactions between tubular epithelial cells and renal fibroblasts.
Tue Sep 01 00:00:00 UTC 2020
TGF‑β induces periodontal ligament stem cell senescence through increase of ROS production.
Molecular medicine reports
Fan C,Ji Q,Zhang C,Xu S,Sun H,Li Z
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Tue Oct 01 00:00:00 UTC 2019
High-yield isolation of menstrual blood-derived endometrial stem cells by direct red blood cell lysis treatment.
Biology open
Sun Y,Ren Y,Yang F,He Y,Liang S,Guan L,Cheng F,Liu Y,Lin J
11-0909 was used in Flow cytometry/Cell sorting to determine whether primary MenSCs exist in the sedimentation of the deciduous endometrium after DGC and further to evaluate the isolation of MenSCs by direct red blood cell lysis treatment.
Thu May 02 00:00:00 UTC 2019
High-yield isolation of menstrual blood-derived endometrial stem cells by direct red blood cell lysis treatment.
Biology open
Sun Y,Ren Y,Yang F,He Y,Liang S,Guan L,Cheng F,Liu Y,Lin J
11-0909 was used in Flow cytometry/Cell sorting to determine whether primary MenSCs exist in the sedimentation of the deciduous endometrium after DGC and further to evaluate the isolation of MenSCs by direct red blood cell lysis treatment.
Thu May 02 00:00:00 UTC 2019
Visfatin Mediates Malignant Behaviors through Adipose-Derived Stem Cells Intermediary in Breast Cancer.
Cancers
Huang JY,Wang YY,Lo S,Tseng LM,Chen DR,Wu YC,Hou MF,Yuan SF
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Fri Dec 20 00:00:00 UTC 2019
Better therapeutic potential of bone marrow-derived mesenchymal stem cells compared with chorionic villi-derived mesenchymal stem cells in airway injury model.
Regenerative medicine
Ji S,Wu C,Tong L,Wang L,Zhou J,Chen C,Song Y
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Fri Mar 01 00:00:00 UTC 2019
The suppressive effects of miR-508-5p on the odontogenic differentiation of human dental pulp stem cells by targeting glycoprotein non-metastatic melanomal protein B.
Stem cell research & therapy
Liu F,Wang X,Yang Y,Hu R,Wang W,Wang Y
11-0909-42 was used in Flow cytometry/Cell sorting to investigate the effects of miR-508-5p on the glycoprotein non-metastatic melanomal protein B expression and the odontogenic differentiation of human dental pulp stem cells.
Tue Jan 22 00:00:00 UTC 2019
Restoration of primary cilia in obese adipose-derived mesenchymal stem cells by inhibiting Aurora A or extracellular signal-regulated kinase.
Stem cell research & therapy
Ritter A,Kreis NN,Roth S,Friemel A,Jennewein L,Eichbaum C,Solbach C,Louwen F,Yuan J
11-0909-42 was used in Flow cytometry/Cell sorting to describes a novel phenomenon whereby the primary cilium of obese ASCs is rescuable by the low-dose inhibition of Aurora A or Erk1/2, restoring functional ASCs with increased stemness.
Wed Aug 14 00:00:00 UTC 2019
Subcutaneous and Visceral Adipose-Derived Mesenchymal Stem Cells: Commonality and Diversity.
Cells
Ritter A,Friemel A,Roth S,Kreis NN,Hoock SC,Safdar BK,Fischer K,Möllmann C,Solbach C,Louwen F,Yuan J
11-0909-42 was used in Flow Cytometry to analyzed the functional similarities and differences of subcutaneous and visceral adipose-derived mesenchymal stem cells.
Mon Oct 21 00:00:00 UTC 2019
Investigating the potential of the secretome of mesenchymal stem cells derived from sickle cell disease patients.
PloS one
Ribeiro TO,Silveira BM,Meira MC,Carreira ACO,Sogayar MC,Meyer R,Fortuna V
11-0909 was used in Flow cytometry/Cell sorting to find that hypoxically conditioned cells presented increased expression and secretion of several well-characterized trophic growth factors (VEGF, IL8, MCP-1, ANG) directly linked to angiogenesis and tissue repair.
Wed Mar 11 00:00:00 UTC 2020
Spatial and Single-Cell Transcriptional Profiling Identifies Functionally Distinct Human Dermal Fibroblast Subpopulations.
The Journal of investigative dermatology
Philippeos C,Telerman SB,Oulès B,Pisco AO,Shaw TJ,Elgueta R,Lombardi G,Driskell RR,Soldin M,Lynch MD,Watt FM
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Sun Apr 01 00:00:00 UTC 2018
CD90 promotes cell migration, viability and sphere‑forming ability of hepatocellular carcinoma cells.
International journal of molecular medicine
Zhang K,Che S,Su Z,Zheng S,Zhang H,Yang S,Li W,Liu J
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Thu Feb 01 00:00:00 UTC 2018
CD90 promotes cell migration, viability and sphere‑forming ability of hepatocellular carcinoma cells.
International journal of molecular medicine
Zhang K,Che S,Su Z,Zheng S,Zhang H,Yang S,Li W,Liu J
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Thu Feb 01 00:00:00 UTC 2018
In Vitro Expansion and Characterization of Mesenchymal Stromal Cells from Peritoneal Dialysis Effluent in a Human Protein Medium.
Stem cells international
Han B,Zhou L,Guan Q,da Roza G,Wang H,Du C
11-0909 was used in Flow cytometry/Cell sorting to show that upon expansion in a human protein medium, pMSCs shows a differential MSC marker expression profile from those of bone marrow or adipose tissue-derived MSCs and could maintain the multipotency.
Wed Sep 30 00:00:00 UTC 2020
Isolation and characterization of stem cells from differentially degenerated human lumbar zygapophyseal articular cartilage.
Molecular medicine reports
Xiao L,Xu S,Wang X,Jin Z,Wang J,Yang B,Xu H
11-0909 was used in Flow cytometry/Cell sorting to show stem cells with multilineage differentiation potential and clonal properties were identified in human LZAC, and these characteristics were more prominent in mildly degenerated as compared with severely degenerated articular cartilage.
Sat Dec 01 00:00:00 UTC 2018
Human umbilical cord-derived mesenchymal stem cells ameliorate the enteropathy of food allergies in mice.
Experimental and therapeutic medicine
Yan N,Xu J,Zhao C,Wu Y,Gao F,Li C,Zhou W,Xiao T,Zhou X,Shao Q,Xia S
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Sat Dec 01 00:00:00 UTC 2018
BNIP3/Bcl-2-mediated apoptosis induced by cyclic tensile stretch in human cartilage endplate-derived stem cells.
Experimental and therapeutic medicine
Yuan C,Pu L,He Z,Wang J
11-0909-42 was used in Flow Cytometry to investigate the molecular mechanisms of cyclic stretch-induced apoptosis in human intervertebral disc cartilage endplate-derived stem cells (CESCs).
Mon Jan 01 00:00:00 UTC 2018
Comprehensive characterization of chorionic villi-derived mesenchymal stromal cells from human placenta.
Stem cell research & therapy
Ventura Ferreira MS,Bienert M,Müller K,Rath B,Goecke T,Opländer C,Braunschweig T,Mela P,Brümmendorf TH,Beier F,Neuss S
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Mon Feb 05 00:00:00 UTC 2018
Immunophenotyping and transcriptional profiling of in vitro cultured human adipose tissue derived stem cells.
Scientific reports
Mieczkowska A,Schumacher A,Filipowicz N,Wardowska A,Zieliński M,Madanecki P,Nowicka E,Langa P,Deptuła M,Zieliński J,Kondej K,Renkielska A,Buckley PG,Crossman DK,Crowley MR,Czupryn A,Mucha P,Sachadyn P,Janus Ł,Skowron P,Rodziewicz-Motowidło S,Cichorek M,Pikuła M,Piotrowski A
11-0909 was used in Flow cytometry/Cell sorting to study the impact of prolonged cell culture on the adipose-derived stem cell phenotype and their differentiation potential.
Fri Jul 27 00:00:00 UTC 2018
Effect of MTA and CEM on Mineralization-Associated Gene Expression in Stem Cells Derived from Apical Papilla.
Iranian endodontic journal
Hajizadeh N,Madani ZS,Zabihi E,Golpour M,Zahedpasha A,Mohammadnia M
11-0909 was used in Flow cytometry/Cell sorting to assess the effect of mineral trioxide aggregate and calcium-enriched mixture cement on odontogenic differentiation and mineralisation of stem cells.
Thu Mar 17 00:00:00 UTC 2022
Co-culture of dendritic cells and cytokine-induced killer cells effectively suppresses liver cancer stem cell growth by inhibiting pathways in the immune system.
BMC cancer
Yang T,Zhang W,Wang L,Xiao C,Wang L,Gong Y,Huang D,Guo B,Li Q,Xiang Y,Nan Y
11-0909 was used in Flow cytometry/Cell sorting to investigate whether dendritic cells co-cultured with autologous cytokine-induced killer cells, could induce a more specific immune response against liver cancer stem cells generated from human hepatocellular carcinoma cells.
Tue Oct 16 00:00:00 UTC 2018
Loss of KLF15 accelerates chronic podocyte injury.
International journal of molecular medicine
Han SS,Yu MY,Yoo KD,Lee JP,Kim DK,Kim YS,Yang SH
11-0909-42 was used in Flow cytometry/Cell sorting to investigate podocyte injury regulation by KLF15.
Sat Sep 01 00:00:00 UTC 2018
Immunosuppressive Mesenchymal Stromal Cells Derived from Human-Induced Pluripotent Stem Cells Induce Human Regulatory T Cells In Vitro and In Vivo.
Frontiers in immunology
Roux C,Saviane G,Pini J,Belaïd N,Dhib G,Voha C,Ibáñez L,Boutin A,Mazure NM,Wakkach A,Blin-Wakkach C,Rouleau M
11-0909 was used in Flow cytometry/Cell sorting to examine the immunosuppressive activity of human induced pluripotent stem cell derived-mesenchymal stromal cells on human T-cell stimulation with a concomitant generation of human regulatory T cells in vivo.
Wed Nov 20 00:00:00 UTC 2019
Endothelial and smooth muscle cells derived from human cardiac explants demonstrate angiogenic potential and suitable for design of cell-containing vascular grafts.
Journal of translational medicine
Zakharova IS,Zhiven' MK,Saaya SB,Shevchenko AI,Smirnova AM,Strunov A,Karpenko AA,Pokushalov EA,Ivanova LN,Makarevich PI,Parfyonova YV,Aboian E,Zakian SM
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Fri Mar 03 00:00:00 UTC 2017
Human umbilical cord mesenchymal stem cells improve the reserve function of perimenopausal ovary via a paracrine mechanism.
Stem cell research & therapy
Li J,Mao Q,He J,She H,Zhang Z,Yin C
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Thu Mar 09 00:00:00 UTC 2017
Feasibility of placenta-derived mesenchymal stem cells as a tool for studying pregnancy-related disorders.
Scientific reports
Fuchi N,Miura K,Doi H,Li TS,Masuzaki H
11-0909 was used in Flow cytometry/Cell sorting to evaluate the feasibility of using placenta-derived mesenchymal stem cells as a tool to study pregnancy related disorders.
Wed Apr 12 00:00:00 UTC 2017
A Member of the Nuclear Receptor Superfamily, Designated as NR2F2, Supports the Self-Renewal Capacity and Pluripotency of Human Bone Marrow-Derived Mesenchymal Stem Cells.
Stem cells international
Zhu N,Wang H,Wang B,Wei J,Shan W,Feng J,Huang H
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Tue Jan 19 00:00:00 UTC 2016
NR2F2 regulates bone marrow-derived mesenchymal stem cell-promoted proliferation of Reh cells.
Molecular medicine reports
Zhu N,Wang H,Wei J,Wang B,Shan W,Lai X,Zhao Y,Yu J,Huang H
11-0909 was used in Flow cytometry/Cell sorting to demonstrate NR2F2 mediates bone marrow-derived mesenchymal stem cell induced proliferation of Reh cells via regulation of vascular endothelial growth factor A.
Mon Aug 01 00:00:00 UTC 2016
Influence of aging on the quantity and quality of human cardiac stem cells.
Scientific reports
Nakamura T,Hosoyama T,Kawamura D,Takeuchi Y,Tanaka Y,Samura M,Ueno K,Nishimoto A,Kurazumi H,Suzuki R,Ito H,Sakata K,Mikamo A,Li TS,Hamano K
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Mon Mar 07 00:00:00 UTC 2016
MIF Plays a Key Role in Regulating Tissue-Specific Chondro-Osteogenic Differentiation Fate of Human Cartilage Endplate Stem Cells under Hypoxia.
Stem cell reports
Yao Y,Deng Q,Song W,Zhang H,Li Y,Yang Y,Fan X,Liu M,Shang J,Sun C,Tang Y,Jin X,Liu H,Huang B,Zhou Y
11-0909 was used in Flow cytometry/Cell sorting to show MIF plays a role in regulating the differentiation fate of human cartilage endplate stem cells under hypoxia.
Tue Aug 09 00:00:00 UTC 2016
CRISPR/Cas9-based genetic correction for recessive dystrophic epidermolysis bullosa.
NPJ Regenerative medicine
Webber BR,Osborn MJ,McElroy AN,Twaroski K,Lonetree CL,DeFeo AP,Xia L,Eide C,Lees CJ,McElmurry RT,Riddle MJ,Kim CJ,Patel DD,Blazar BR,Tolar J
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Wed Sep 30 00:00:00 UTC 2020
Potency of umbilical cord blood- and Wharton's jelly-derived mesenchymal stem cells for scarless wound healing.
Scientific reports
Doi H,Kitajima Y,Luo L,Yan C,Tateishi S,Ono Y,Urata Y,Goto S,Mori R,Masuzaki H,Shimokawa I,Hirano A,Li TS
11-0909 was used in Flow cytometry/Cell sorting to determine that umbilical cord blood offers a potential source of MSCs for therapeutic scarless wound healing.
Tue Jan 05 00:00:00 UTC 2016
Stromal Activation by Tumor Cells: An in Vitro Study in Breast Cancer.
Microarrays (Basel, Switzerland)
Merlino G,Miodini P,Paolini B,Carcangiu ML,Gennaro M,Dugo M,Daidone MG,Cappelletti V
11-0909 was used in Flow cytometry/Cell sorting to suggest that stromal TGM2 may be associated with precancerous evolution at earlier stages compared to ductal carcinoma in situ.
Wed May 18 00:00:00 UTC 2016
Mesenchymal stromal cells form vascular tubes when placed in fibrin sealant and accelerate wound healing in vivo.
Biomaterials
Mendez JJ,Ghaedi M,Sivarapatna A,Dimitrievska S,Shao Z,Osuji CO,Steinbacher DM,Leffell DJ,Niklason LE
11-0909 was used in Flow cytometry/Cell sorting to show that ADSCs are able to form tubular structures within fibrin gels, and may also contribute to faster wound healing.
Sun Feb 01 00:00:00 UTC 2015
Human esophageal myofibroblasts secrete proinflammatory cytokines in response to acid and Toll-like receptor 4 ligands.
American journal of physiology. Gastrointestinal and liver physiology
Gargus M,Niu C,Vallone JG,Binkley J,Rubin DC,Shaker A
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Mon Jun 01 00:00:00 UTC 2015
Immunophenotypic comparison of heterogenous non-sorted versus sorted mononuclear cells from human umbilical cord blood: a novel cell enrichment approach.
Cytotechnology
Indumathi S,Harikrishnan R,Rajkumar JS,Dhanasekaran M
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Thu Jan 01 00:00:00 UTC 2015
Human adipose stem cell and ASC-derived cardiac progenitor cellular therapy improves outcomes in a murine model of myocardial infarction.
Stem cells and cloning : advances and applications
Davy PM,Lye KD,Mathews J,Owens JB,Chow AY,Wong L,Moisyadi S,Allsopp RC
11-0909 was used in Flow cytometry/Cell sorting to investigate the potential use of adipose-derived stem cells.
Wed Nov 25 00:00:00 UTC 2015
Efficacy of autologous stem cell-based therapy for osteonecrosis of the femoral head in sickle cell disease: a five-year follow-up study.
Stem cell research & therapy
Daltro GC,Fortuna V,de Souza ES,Salles MM,Carreira AC,Meyer R,Freire SM,Borojevic R
11-0909 was used in Flow cytometry/Cell sorting to report a significant improvement in clinical joint symptoms and pain relief in patients after bone marrow-derived mononuclear cell implantation.
Fri May 29 00:00:00 UTC 2015
Comparison of human adipose-derived stem cells isolated from subcutaneous, omental, and intrathoracic adipose tissue depots for regenerative applications.
Stem cells translational medicine
Russo V,Yu C,Belliveau P,Hamilton A,Flynn LE
11-0909 was used in Flow cytometry/Cell sorting to determine that depot selection is an important factor to consider when applying ASCs in tissue-specific cell-based regenerative therapies.
Sat Feb 01 00:00:00 UTC 2014
Epithelial cell differentiation of human mesenchymal stromal cells in decellularized lung scaffolds.
Tissue engineering. Part A
Mendez JJ,Ghaedi M,Steinbacher D,Niklason LE
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Sun Jun 01 00:00:00 UTC 2014
Hypoxia-cultured human adipose-derived mesenchymal stem cells are non-oncogenic and have enhanced viability, motility, and tropism to brain cancer.
Cell death & disease
Feng Y,Zhu M,Dangelmajer S,Lee YM,Wijesekera O,Castellanos CX,Denduluri A,Chaichana KL,Li Q,Zhang H,Levchenko A,Guerrero-Cazares H,Quiñones-Hinojosa A
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Thu Dec 11 00:00:00 UTC 2014
PDGFR-β (+) perivascular cells from infantile hemangioma display the features of mesenchymal stem cells and show stronger adipogenic potential in vitro and in vivo.
International journal of clinical and experimental pathology
Yuan SM,Guo Y,Zhou XJ,Shen WM,Chen HN
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 11-0909-42) in Flow Cytometry
Tue May 26 00:00:00 UTC 2015
Expression of intercellular adhesion molecule 1 by hepatocellular carcinoma stem cells and circulating tumor cells.
Gastroenterology
Liu S,Li N,Yu X,Xiao X,Cheng K,Hu J,Wang J,Zhang D,Cheng S,Liu S
11-0909 was used in Flow cytometry/Cell sorting to demonstrate that the stem cell factor Nanog regulates expression of ICAM-1 in hepatocellular carcinoma cells.
Wed May 01 00:00:00 UTC 2013
Downregulation of Mcl-1 synergizes the apoptotic response to combined treatment with cisplatin and a novel fiber chimeric oncolytic adenovirus.
Oncology reports
You L,Wang Y,Jin Y,Qian W
11-0909 was used in Flow cytometry/Cell sorting to determine how SG511 sensitises cancer cells to apoptosis by reducing anti-apoptotic Mcl-1 protein levels.
Sun Apr 01 00:00:00 UTC 2012
A novel mechanism involved in the pathogenesis of Graves ophthalmopathy (GO): clathrin is a possible targeting molecule for inhibiting local immune response in the orbit.
The Journal of clinical endocrinology and metabolism
Meyer zu Hörste M,Ströher E,Berchner-Pfannschmidt U,Schmitz-Spanke S,Pink M,Göthert JR,Fischer JW,Gulbins E,Eckstein AK
11-0909 was used in Flow cytometry/Cell sorting to analyse and inhibit signalling pathways in resident orbital fibroblasts that are involved in Graves opthalmopathy.
Tue Nov 01 00:00:00 UTC 2011