Product References
Osteogenic potential of apical papilla stem cells mediated by platelet-rich fibrin and low-level laser.
Odontology
Ramírez DG,Inostroza C,Rouabhia M,Rodriguez CA,Gómez LA,Losada M,Muñoz AL
12-0909-42 was used in Flow Cytometry to demonstrate for the first time that the combination of PRF and LLLT would be an excellent therapeutic tool that can be employed for dental, oral, and craniofacial repair and other tissue engineering applications.
Mon Apr 01 00:00:00 UTC 2024
Osteogenic potential of apical papilla stem cells mediated by platelet-rich fibrin and low-level laser.
Odontology
Ramírez DG,Inostroza C,Rouabhia M,Rodriguez CA,Gómez LA,Losada M,Muñoz AL
12-0909-42 was used in Flow Cytometry to demonstrate for the first time that the combination of PRF and LLLT would be an excellent therapeutic tool that can be employed for dental, oral, and craniofacial repair and other tissue engineering applications.
Mon Apr 01 00:00:00 UTC 2024
CD304 + adipose tissue-derived mesenchymal stem cell abundance in autologous fat grafts highly correlates with improvement of localized pain syndromes.
Pain
Rezzonico Jost T,Lozito A,Mangani D,Raimondi A,Klinger F,Morone D,Klinger M,Grassi F,Vinci V
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Mon Apr 01 00:00:00 UTC 2024
Stimulated Human Umbilical Cord Mesenchymal Stem Cells Enhance the Osteogenesis and Cranial Bone Regeneration through IL-32 Mediated P38 Signaling Pathway.
Stem cells international
Zhang X,Zheng Y,Wang G,Liu Y,Wang Y,Jiang X,Liang Y,Zhao X,Li P,Zhang Y
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Fri Mar 22 00:00:00 UTC 2024
Generation of Highly Functional Hepatocyte-like Organoids from Human Adipose-Derived Mesenchymal Stem Cells Cultured with Endothelial Cells.
Cells
Chen S,Saito Y,Waki Y,Ikemoto T,Teraoku H,Yamada S,Morine Y,Shimada M
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Wed Mar 20 00:00:00 UTC 2024
Adipose-derived stem cell exosomes promote tumor characterization and immunosuppressive microenvironment in breast cancer.
Cancer immunology, immunotherapy : CII
Zhu Q,Cao Y,Yuan J,Hu Y
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Wed Jan 31 00:00:00 UTC 2024
Umbilical cord mesenchymal stem cell-derived apoptotic extracellular vesicles ameliorate cutaneous wound healing in type 2 diabetic mice via macrophage pyroptosis inhibition.
Stem cell research & therapy
Wang Y,Jing L,Lei X,Ma Z,Li B,Shi Y,Zhang W,Li Y,Zhou H,Hu K,Xue Y,Jin Y
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Tue Sep 19 00:00:00 UTC 2023
Fluorescence-activated cell sorting and phenotypic characterization of human fibro-adipogenic progenitors.
STAR protocols
Billeskov TB,Jensen JB,Jessen N,Farup J
12-0909-42 was used in Flow Cytometry to describe important phenotypic traits such as time to enter the cell cycle and assessment of cell differentiation for the isolated cell populations.
Fri Mar 17 00:00:00 UTC 2023
Exercise combined with administration of adipose-derived stem cells ameliorates neuropathic pain after spinal cord injury.
Neural regeneration research
Cheng X,Mao GP,Hu WJ,Yu ZR,Xu YY,Chen W,Li X,Zeng XL,Zhang WW,Chen JW,Wan Y,Wang L
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Tue Aug 01 00:00:00 UTC 2023
Proteomics of immune cells from liver tumors reveals immunotherapy targets.
Cell genomics
Canale FP,Neumann J,von Renesse J,Loggi E,Pecoraro M,Vogel I,Zoppi G,Antonini G,Wolf T,Jin W,Zheng X,La Barba G,Birgin E,Forkel M,Nilsson T,Marone R,Mueller H,Pelletier N,Jeker LT,Civenni G,Schlapbach C,Catapano CV,Seifert L,Seifert AM,Gillessen S,De Dosso S,Cristaudi A,Rahbari NN,Ercolani G,Geiger R
12-0909-42 was used in Flow Cytometry to profile proteomes of cancer tissue as well as monocyte/macrophages, CD4+ and CD8+ T cells, and NK cells isolated from tumors, liver, and blood of 48 patients with hepatocellular carcinoma.
Wed Jun 14 00:00:00 UTC 2023
Carboxymethyl chitin or chitosan for osteoinduction effect on the human periodontal ligament stem cells.
Dental materials journal
Fan C,Li Z,Ji Q,Sun H,Liang Y,Yang P
12-0909-42 was used in Flow Cytometry to elucidate carboxymethyl chitosan (CMCTS) and carboxymethyl chitin (CMCT) effect on osteogenic differentiation.
Tue May 31 00:00:00 UTC 2022
Exosomes derived from stem cells of human deciduous exfoliated teeth inhibit angiogenesis in vivo and in vitro via the transfer of miR-100-5p and miR-1246.
Stem cell research & therapy
Liu P,Zhang Q,Mi J,Wang S,Xu Q,Zhuang D,Chen W,Liu C,Zhang L,Guo J,Wu X
12-0909-42 was used in Flow Cytometry to explore the potential role of exosomes derived from stem cells of human deciduous exfoliated teeth (SHED-Exos) in regulating angiogenesis and the underlying molecular mechanism.
Thu Mar 03 00:00:00 UTC 2022
Exosomes derived from stem cells of human deciduous exfoliated teeth inhibit angiogenesis in vivo and in vitro via the transfer of miR-100-5p and miR-1246.
Stem cell research & therapy
Liu P,Zhang Q,Mi J,Wang S,Xu Q,Zhuang D,Chen W,Liu C,Zhang L,Guo J,Wu X
12-0909-42 was used in Flow Cytometry to explore the potential role of exosomes derived from stem cells of human deciduous exfoliated teeth (SHED-Exos) in regulating angiogenesis and the underlying molecular mechanism.
Thu Mar 03 00:00:00 UTC 2022
Aberrant Expression of COX-2 and FOXG1 in Infrapatellar Fat Pad-Derived ASCs from Pre-Diabetic Donors.
Cells
O'Donnell BT,Monjure TA,Al-Ghadban S,Ives CJ,L'Ecuyer MP,Rhee C,Romero-Lopez M,Li Z,Goodman SB,Lin H,Tuan RS,Bunnell BA
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Mon Aug 01 00:00:00 UTC 2022
Fused Cells between Human-Adipose-Derived Mesenchymal Stem Cells and Monocytes Keep Stemness Properties and Acquire High Mobility.
International journal of molecular sciences
Montalbán-Hernández K,Casado-Sánchez C,Avendaño-Ortiz J,Casalvilla-Dueñas JC,Bonel-Pérez GC,Prado-Montero J,Valentín-Quiroga J,Lozano-Rodríguez R,Terrón-Arcos V,de la Bastida FR,Córdoba L,Laso-García F,Diekhorst L,Del Fresno C,López-Collazo E
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Fri Aug 26 00:00:00 UTC 2022
Mesenchymal stem cells transfer mitochondria to allogeneic Tregs in an HLA-dependent manner improving their immunosuppressive activity.
Nature communications
Piekarska K,Urban-Wójciuk Z,Kurkowiak M,Pelikant-Małecka I,Schumacher A,Sakowska J,Spodnik JH,Arcimowicz Ł,Zielińska H,Tymoniuk B,Renkielska A,Siebert J,Słomińska E,Trzonkowski P,Hupp T,Marek-Trzonkowska NM
12-0909-42 was used in Flow Cytometry to demonstrate that both direct and indirect contact with allogeneic ASCs improves therapeutic potential of Tregs (Regulatory T cells).
Mon Feb 14 00:00:00 UTC 2022
Tumor necrosis factor-α-primed mesenchymal stem cell-derived exosomes promote M2 macrophage polarization via Galectin-1 and modify intrauterine adhesion on a novel murine model.
Frontiers in immunology
Li J,Pan Y,Yang J,Wang J,Jiang Q,Dou H,Hou Y
12-0909-42 was used in Flow cytometry/Cell sorting to propose an innovative mouse model and a better MSC treatment strategy for IUA.
Wed Jan 11 00:00:00 UTC 2023
LIM Mineralization Protein-1 Enhances the Committed Differentiation of Dental Pulp Stem Cells through the ERK1/2 and p38 MAPK Pathways and BMP Signaling.
International journal of medical sciences
Mu R,Chen B,Bi B,Yu H,Liu J,Li J,He M,Rong L,Liu B,Liu K,Zhu L,Shi X,Shuai Y,Jin L
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Mon Aug 08 00:00:00 UTC 2022
RNA-seq profiling of tubulointerstitial tissue reveals a potential therapeutic role of dual anti-phosphatase 1 in glomerulonephritis.
Journal of cellular and molecular medicine
Park S,Lee H,Lee J,Lee S,Cho S,Huh H,Kim JY,Park M,Lee S,Kim Y,Choi M,Joo KW,Kim YS,Yang SH,Kim DK
12-0909-42 was used in Flow cytometry/Cell sorting to show that transcriptome profiling of tubulointerstitial tissue from patients with various glomerulonephritis revealed notable findings that may provide insights into pathophysiologic mechanisms.
Wed Jun 01 00:00:00 UTC 2022
The Composition of Adipose-Derived Regenerative Cells Isolated from Lipoaspirate Using a Point of Care System Does Not Depend on the Subject's Individual Age, Sex, Body Mass Index and Ethnicity.
Cells
Schmitz C,Alt C,Azares AR,Pearce DA,Facile TR,Furia JP,Maffulli N,Huang C,Alt EU
12-0909-42 was used in Flow cytometry/Cell sorting to show that key characteristics of unmodified, autologous, adipose-derived regenerative cells (UA-ADRCs) can be independent of the subject's age, sex, BMI and ethnicity.
Wed Dec 21 00:00:00 UTC 2022
Single-cell Transcriptomic Analysis Reveals the Cellular Heterogeneity of Mesenchymal Stem Cells.
Genomics, proteomics & bioinformatics
Zhang C,Han X,Liu J,Chen L,Lei Y,Chen K,Si J,Wang TY,Zhou H,Zhao X,Zhang X,An Y,Li Y,Wang QF
12-0909-42 was used in Flow cytometry/Cell sorting to reveal five distinct subpopulations from bone marrow and Wharton's jelly.
Tue Feb 01 00:00:00 UTC 2022
Cell-mimicking nanodecoys neutralize SARS-CoV-2 and mitigate lung injury in a non-human primate model of COVID-19.
Nature nanotechnology
Li Z,Wang Z,Dinh PC,Zhu D,Popowski KD,Lutz H,Hu S,Lewis MG,Cook A,Andersen H,Greenhouse J,Pessaint L,Lobo LJ,Cheng K
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Sun Aug 01 00:00:00 UTC 2021
VAP-PLGA microspheres (VAP-PLGA) promote adipose-derived stem cells (ADSCs)-induced wound healing in chronic skin ulcers in mice via PI3K/Akt/HIF-1α pathway.
Bioengineered
Jiang W,Zhang J,Zhang X,Fan C,Huang J
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Wed Dec 01 00:00:00 UTC 2021
Exosomes Regulate Interclonal Communication on Osteogenic Differentiation Among Heterogeneous Osteogenic Single-Cell Clones Through PINK1/Parkin-Mediated Mitophagy.
Frontiers in cell and developmental biology
Fei D,Xia Y,Zhai Q,Wang Y,Zhou F,Zhao W,He X,Wang Q,Jin Y,Li B
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Tue Oct 05 00:00:00 UTC 2021
Isolation and characterization of muscle stem cells, fibro-adipogenic progenitors, and macrophages from human skeletal muscle biopsies.
American journal of physiology. Cell physiology
Jensen JB,Møller AB,Just J,Mose M,de Paoli FV,Billeskov TB,Fred RG,Pers TH,Pedersen SB,Petersen KK,Bjerre M,Farup J,Jessen N
12-0909-42 was used in Flow cytometry/Cell sorting to demonstrate that mononuclear cell populations can be quantified and subsequently analyzed from needle biopsy material and opens the perspective for future clinical studies of cellular mechanisms in muscle wasting.
Sun Aug 01 00:00:00 UTC 2021
TRPA1 triggers hyperalgesia and inflammation after tooth bleaching.
Scientific reports
Chen C,Huang X,Zhu W,Ding C,Huang P,Li R
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Tue Aug 31 00:00:00 UTC 2021
In Vitro Anti-cancer Activity of Adipose-Derived Mesenchymal Stem Cells Increased after Infection with Oncolytic Reovirus.
Advanced pharmaceutical bulletin
Babaei A,Bannazadeh Baghi H,Nezhadi A,Jamalpoor Z
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Mon Feb 01 00:00:00 UTC 2021
Therapeutic potential of human umbilical cord mesenchymal stem cells on aortic atherosclerotic plaque in a high-fat diet rabbit model.
Stem cell research & therapy
Li Y,Shi G,Han Y,Shang H,Li H,Liang W,Zhao W,Bai L,Qin C
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Thu Jul 15 00:00:00 UTC 2021
Wip1 regulates the immunomodulatory effects of murine mesenchymal stem cells in type 1 diabetes mellitus via targeting IFN-α/BST2.
Cell death discovery
Zhou N,Liu W,Zhang W,Liu Y,Li X,Wang Y,Zheng R,Zhang Y
12-0909-42 was used in Flow Cytometry to reveal Wip1 affects MSCs immunomodulation by regulating the expression of IFN-α/BST2.
Fri Oct 29 00:00:00 UTC 2021
Low-Level Laser Irradiation Promotes Proliferation and Differentiation on Apical Papilla Stem Cells.
Journal of lasers in medical sciences
Gutiérrez D,Rouabhia M,Ortiz J,Gaviria D,Alfonso C,Muñoz A,Inostroza C
12-0909-42 was used in Flow Cytometry to conclude that LLLT promotes proliferation, osteogenic differentiation, and VEGF and TGFβ2 expression on SCAPs.
Fri Feb 18 00:00:00 UTC 2022
Mesenchymal stem cell exosome-derived miR-223 alleviates acute graft-versus-host disease via reducing the migration of donor T cells.
Stem cell research & therapy
Liu W,Zhou N,Liu Y,Zhang W,Li X,Wang Y,Zheng R,Zhang Y
12-0909-42 was used in Flow cytometry/Cell sorting to unveil a new role of MSCs-Exo containing miR-223 in the treatment of acute graft-versus-host disease (aGvHD).
Fri Feb 26 00:00:00 UTC 2021
Human skeletal muscle CD90+ fibro-adipogenic progenitors are associated with muscle degeneration in type 2 diabetic patients.
Cell metabolism
Farup J,Just J,de Paoli F,Lin L,Jensen JB,Billeskov T,Roman IS,Cömert C,Møller AB,Madaro L,Groppa E,Fred RG,Kampmann U,Gormsen LC,Pedersen SB,Bross P,Stevnsner T,Eldrup N,Pers TH,Rossi FMV,Puri PL,Jessen N
12-0909-42 was used in Flow Cytometry to identify a platelet-derived growth factor-driven conversion of a subpopulation of fibro-adipogenic progenitors as a key event in the fibrosis development in Type 2 diabetes mellitus muscle.
Tue Nov 02 00:00:00 UTC 2021
Glycemic control by umbilical cord-derived mesenchymal stem cells promotes effects of fasting-mimicking diet on type 2 diabetic mice.
Stem cell research & therapy
Zhao N,Gao YF,Bao L,Lei J,An HX,Pu FX,Cheng RP,Chen J,Ni H,Sui BD,Ji FP,Hu CH
12-0909-42 was used in Flow Cytometry to establish a strategy that combined UC-MSC infusion and FMD and was effective in treating T2D, which provided potential approaches for developing novel clinical T2D therapies.
Tue Jul 13 00:00:00 UTC 2021
Apoptotic vesicles restore liver macrophage homeostasis to counteract type 2 diabetes.
Journal of extracellular vesicles
Zheng C,Sui B,Zhang X,Hu J,Chen J,Liu J,Wu D,Ye Q,Xiang L,Qiu X,Liu S,Deng Z,Zhou J,Liu S,Shi S,Jin Y
12-0909-42 was used in Flow Cytometry to investigate if functional efferocytosis of apoptotic vesicles restore liver macrophage homeostasis and ameliorates type 2 diabetes.
Sat May 01 00:00:00 UTC 2021
Immunity-and-matrix-regulatory cells derived from human embryonic stem cells safely and effectively treat mouse lung injury and fibrosis.
Cell research
Wu J,Song D,Li Z,Guo B,Xiao Y,Liu W,Liang L,Feng C,Gao T,Chen Y,Li Y,Wang Z,Wen J,Yang S,Liu P,Wang L,Wang Y,Peng L,Stacey GN,Hu Z,Feng G,Li W,Huo Y,Jin R,Shyh-Chang N,Zhou Q,Wang L,Hu B,Dai H,Hao J
12-0909 was used in Flow cytometry/Cell sorting to investigate the use of hESC-derived immunity-regulatory and matrix-regulatory cells in treating lung injury and fibrosis in mice.
Tue Sep 01 00:00:00 UTC 2020
Down-Regulated Exosomal MicroRNA-221 - 3p Derived From Senescent Mesenchymal Stem Cells Impairs Heart Repair.
Frontiers in cell and developmental biology
Sun L,Zhu W,Zhao P,Zhang J,Lu Y,Zhu Y,Zhao W,Liu Y,Chen Q,Zhang F
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Mon Sep 28 00:00:00 UTC 2020
Differentiation Potential of Early- and Late-Passage Adipose-Derived Mesenchymal Stem Cells Cultured under Hypoxia and Normoxia.
Stem cells international
Zhao AG,Shah K,Freitag J,Cromer B,Sumer H
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Tue Oct 06 00:00:00 UTC 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Fri Apr 03 00:00:00 UTC 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Fri Apr 03 00:00:00 UTC 2020
The Effect of Angiotensin II, Retinoic Acid, EGCG, and Vitamin C on the Cardiomyogenic Differentiation Induction of Human Amniotic Fluid-Derived Mesenchymal Stem Cells.
International journal of molecular sciences
Gasiūnienė M,Valatkaitė E,Navakauskaitė A,Navakauskienė R
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Thu Nov 19 00:00:00 UTC 2020
Migration Inhibitory Factor in Conditioned Medium from Human Umbilical Cord Blood-Derived Mesenchymal Stromal Cells Stimulates Hair Growth.
Cells
Oh HA,Kwak J,Kim BJ,Jin HJ,Park WS,Choi SJ,Oh W,Um S
12-0909 was used in Flow cytometry/Cell sorting to suggest that P-CM can improve the efficacy in hair growth via a paracrine mechanism and that MIF in P-CM exerts hair growth-promoting effects via a VEGF-related β-catenin and p-GSK-3β [SER9] signaling pathway.
Thu May 28 00:00:00 UTC 2020
BNC1 regulates cell heterogeneity in human pluripotent stem cell-derived epicardium.
Development (Cambridge, England)
Gambardella L,McManus SA,Moignard V,Sebukhan D,Delaune A,Andrews S,Bernard WG,Morrison MA,Riley PR,Göttgens B,Gambardella Le Novère N,Sinha S
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Fri Dec 13 00:00:00 UTC 2019
LPS‑induced upregulation of the TLR4 signaling pathway inhibits osteogenic differentiation of human periodontal ligament stem cells under inflammatory conditions.
International journal of molecular medicine
Yu B,Li Q,Zhou M
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Sat Jun 01 00:00:00 UTC 2019
TGF‑β induces periodontal ligament stem cell senescence through increase of ROS production.
Molecular medicine reports
Fan C,Ji Q,Zhang C,Xu S,Sun H,Li Z
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Tue Oct 01 00:00:00 UTC 2019
Bone mesenchymal stromal cells exhibit functional inhibition but no chromosomal aberrations in chronic myelogenous leukemia.
Oncology letters
Xie J,Chen J,Wang B,He X,Huang H
12-0909-42 was used in Flow Cytometry to demonstrate functional inhibition of CML-BMSCs; however, no signs of chromosomal aberrations were observed, thereby providing insight into the changes occurring in the CML-BMM.
Tue Jan 01 00:00:00 UTC 2019
Isolation and Culture of Human Stem Cells from Apical Papilla under Low Oxygen Concentration Highlight Original Properties.
Cells
Rémy M,Ferraro F,Le Salver P,Rey S,Genot E,Djavaheri-Mergny M,Thébaud N,Boiziau C,Boeuf H
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Thu Nov 21 00:00:00 UTC 2019
Visfatin Mediates Malignant Behaviors through Adipose-Derived Stem Cells Intermediary in Breast Cancer.
Cancers
Huang JY,Wang YY,Lo S,Tseng LM,Chen DR,Wu YC,Hou MF,Yuan SF
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Fri Dec 20 00:00:00 UTC 2019
Better therapeutic potential of bone marrow-derived mesenchymal stem cells compared with chorionic villi-derived mesenchymal stem cells in airway injury model.
Regenerative medicine
Ji S,Wu C,Tong L,Wang L,Zhou J,Chen C,Song Y
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Fri Mar 01 00:00:00 UTC 2019
Spatial and Single-Cell Transcriptional Profiling Identifies Functionally Distinct Human Dermal Fibroblast Subpopulations.
The Journal of investigative dermatology
Philippeos C,Telerman SB,Oulès B,Pisco AO,Shaw TJ,Elgueta R,Lombardi G,Driskell RR,Soldin M,Lynch MD,Watt FM
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Sun Apr 01 00:00:00 UTC 2018
CD90 promotes cell migration, viability and sphere‑forming ability of hepatocellular carcinoma cells.
International journal of molecular medicine
Zhang K,Che S,Su Z,Zheng S,Zhang H,Yang S,Li W,Liu J
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Thu Feb 01 00:00:00 UTC 2018
Human umbilical cord-derived mesenchymal stem cells ameliorate the enteropathy of food allergies in mice.
Experimental and therapeutic medicine
Yan N,Xu J,Zhao C,Wu Y,Gao F,Li C,Zhou W,Xiao T,Zhou X,Shao Q,Xia S
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Sat Dec 01 00:00:00 UTC 2018
Comprehensive characterization of chorionic villi-derived mesenchymal stromal cells from human placenta.
Stem cell research & therapy
Ventura Ferreira MS,Bienert M,Müller K,Rath B,Goecke T,Opländer C,Braunschweig T,Mela P,Brümmendorf TH,Beier F,Neuss S
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Mon Feb 05 00:00:00 UTC 2018
Alkaline Phosphatase Controls Lineage Switching of Mesenchymal Stem Cells by Regulating the LRP6/GSK3β Complex in Hypophosphatasia.
Theranostics
Liu W,Zhang L,Xuan K,Hu C,Li L,Zhang Y,Jin F,Jin Y
12-0909 was used in Flow cytometry/Cell sorting to elucidate a novel role of tissue-nonspecific alkaline phosphatase in controlling bone marrow mesenchymal stem cell lineage selection.
Mon Sep 09 00:00:00 UTC 2019
Distinct signaling programs control human hematopoietic stem cell survival and proliferation.
Blood
Knapp DJ,Hammond CA,Aghaeepour N,Miller PH,Pellacani D,Beer PA,Sachs K,Qiao W,Wang W,Humphries RK,Sauvageau G,Zandstra PW,Bendall SC,Nolan GP,Hansen C,Eaves CJ
12-0909 was used in Flow cytometry/Cell sorting to investigate the molecular mechanisms by which distinct signalling programs control human hematopoietic stem cell survival and proliferation.
Thu Jan 19 00:00:00 UTC 2017
Endothelial and smooth muscle cells derived from human cardiac explants demonstrate angiogenic potential and suitable for design of cell-containing vascular grafts.
Journal of translational medicine
Zakharova IS,Zhiven' MK,Saaya SB,Shevchenko AI,Smirnova AM,Strunov A,Karpenko AA,Pokushalov EA,Ivanova LN,Makarevich PI,Parfyonova YV,Aboian E,Zakian SM
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Fri Mar 03 00:00:00 UTC 2017
Human umbilical cord mesenchymal stem cells improve the reserve function of perimenopausal ovary via a paracrine mechanism.
Stem cell research & therapy
Li J,Mao Q,He J,She H,Zhang Z,Yin C
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Thu Mar 09 00:00:00 UTC 2017
Nrf2 Inhibits Periodontal Ligament Stem Cell Apoptosis under Excessive Oxidative Stress.
International journal of molecular sciences
Liu Y,Yang H,Wen Y,Li B,Zhao Y,Xing J,Zhang M,Chen Y
12-0909 was used in Flow cytometry/Cell sorting to analyze novel mechanisms underlying Nrf2-mediated anti-apoptosis in periodontal ligament stem cells in the periodontitis oxidative microenvironment.
Wed May 17 00:00:00 UTC 2017
Equine Dental Pulp Connective Tissue Particles Reduced Lameness in Horses in a Controlled Clinical Trial.
Frontiers in veterinary science
Bertone AL,Reisbig NA,Kilborne AH,Kaido M,Salmanzadeh N,Lovasz R,Sizemore JL,Scheuermann L,Kopp RJ,Zekas LJ,Brokken MT
12-0909 was used in Flow cytometry/Cell sorting to assess if injection of allogeneic dental pulp tissue particles improves lameness in horses with naturally occurring osteoarthritis or soft tissue injury.
Sun Nov 12 00:00:00 UTC 2023
Effects of acute exposure to low-dose radiation on the characteristics of human bone marrow mesenchymal stromal/stem cells.
Inflammation and regeneration
Fujishiro A,Miura Y,Iwasa M,Fujii S,Sugino N,Andoh A,Hirai H,Maekawa T,Ichinohe T
12-0909 was used in Flow cytometry/Cell sorting to determine whether low dose irradiation affects the function of mesenchymal stromal/stem cells.
Fri Mar 11 00:00:00 UTC 2022
Satellite cell response to erythropoietin treatment and endurance training in healthy young men.
The Journal of physiology
Hoedt A,Christensen B,Nellemann B,Mikkelsen UR,Hansen M,Schjerling P,Farup J
12-0909 was used in Flow cytometry/Cell sorting to establish that endurance training and erythropoiesis-stimulating agent treatment may separately stimulate satellite cell commitment to the myogenic program.
Mon Feb 01 00:00:00 UTC 2016
A Member of the Nuclear Receptor Superfamily, Designated as NR2F2, Supports the Self-Renewal Capacity and Pluripotency of Human Bone Marrow-Derived Mesenchymal Stem Cells.
Stem cells international
Zhu N,Wang H,Wang B,Wei J,Shan W,Feng J,Huang H
12-0909 was used in Flow cytometry/Cell sorting to determine NR2F2 plays an important role in the stemness maintenance of bone marrow-derived mesenchymal stem cells.
Tue Jan 19 00:00:00 UTC 2016
Human cytomegalovirus infection of human adipose-derived stromal/stem cells restricts differentiation along the adipogenic lineage.
Adipocyte
Zwezdaryk KJ,Ferris MB,Strong AL,Morris CA,Bunnell BA,Dhurandhar NV,Gimble JM,Sullivan DE
12-0909 was used in Flow cytometry/Cell sorting to suggest that human cytomegalovirus (HCMV) infection during adipose-derived stromal/stem cell (ASC) therapy may have a poor outcome due to the virus interfering.
Wed May 04 00:00:00 UTC 2016
Transendothelial migration of human umbilical mesenchymal stem cells across uterine endothelial monolayers: Junctional dynamics and putative mechanisms.
Placenta
Ebrahim NA,Leach L
12-0909 was used in Flow cytometry/Cell sorting to study the transmigration of foetal stem cells across maternal endothelial barriers, and investigate whether they can subsequently influence maternal endothelial integrity.
Thu Dec 01 00:00:00 UTC 2016
MIF Plays a Key Role in Regulating Tissue-Specific Chondro-Osteogenic Differentiation Fate of Human Cartilage Endplate Stem Cells under Hypoxia.
Stem cell reports
Yao Y,Deng Q,Song W,Zhang H,Li Y,Yang Y,Fan X,Liu M,Shang J,Sun C,Tang Y,Jin X,Liu H,Huang B,Zhou Y
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Tue Aug 09 00:00:00 UTC 2016
CRISPR/Cas9-based genetic correction for recessive dystrophic epidermolysis bullosa.
NPJ Regenerative medicine
Webber BR,Osborn MJ,McElroy AN,Twaroski K,Lonetree CL,DeFeo AP,Xia L,Eide C,Lees CJ,McElmurry RT,Riddle MJ,Kim CJ,Patel DD,Blazar BR,Tolar J
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Wed Sep 30 00:00:00 UTC 2020
Vitamin D machinery and metabolism in porcine adipose-derived mesenchymal stem cells.
Stem cell research & therapy
Valle YL,Almalki SG,Agrawal DK
12-0909 was used in Flow cytometry/Cell sorting to evaluate whether porcine adipose-derived MSCs can metabolise and respond to vitamin D.
Wed Aug 17 00:00:00 UTC 2016
Human esophageal myofibroblasts secrete proinflammatory cytokines in response to acid and Toll-like receptor 4 ligands.
American journal of physiology. Gastrointestinal and liver physiology
Gargus M,Niu C,Vallone JG,Binkley J,Rubin DC,Shaker A
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Mon Jun 01 00:00:00 UTC 2015
Immunophenotypic comparison of heterogenous non-sorted versus sorted mononuclear cells from human umbilical cord blood: a novel cell enrichment approach.
Cytotechnology
Indumathi S,Harikrishnan R,Rajkumar JS,Dhanasekaran M
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Thu Jan 01 00:00:00 UTC 2015
Impaired function of bone marrow stromal cells in systemic mastocytosis.
Stem cell research
Nemeth K,Wilson TM,Ren JJ,Sabatino M,Stroncek DM,Krepuska M,Bai Y,Robey PG,Metcalfe DD,Mezey E
12-0909 was used in Flow cytometry/Cell sorting to implicate epigenetic change in mesenchymal stem cells as a driver of some of the symptoms associated with systemic mastocytosis.
Wed Jul 01 00:00:00 UTC 2015
Epithelial cell differentiation of human mesenchymal stromal cells in decellularized lung scaffolds.
Tissue engineering. Part A
Mendez JJ,Ghaedi M,Steinbacher D,Niklason LE
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Sun Jun 01 00:00:00 UTC 2014
Hypoxia-cultured human adipose-derived mesenchymal stem cells are non-oncogenic and have enhanced viability, motility, and tropism to brain cancer.
Cell death & disease
Feng Y,Zhu M,Dangelmajer S,Lee YM,Wijesekera O,Castellanos CX,Denduluri A,Chaichana KL,Li Q,Zhang H,Levchenko A,Guerrero-Cazares H,Quiñones-Hinojosa A
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Thu Dec 11 00:00:00 UTC 2014
PDGFR-β (+) perivascular cells from infantile hemangioma display the features of mesenchymal stem cells and show stronger adipogenic potential in vitro and in vivo.
International journal of clinical and experimental pathology
Yuan SM,Guo Y,Zhou XJ,Shen WM,Chen HN
Published figure using CD90 (Thy-1) monoclonal antibody (Product # 12-0909-42) in Flow Cytometry
Tue May 26 00:00:00 UTC 2015