Product References
The histone H2B ubiquitination regulator Wac is essential for plasma cell differentiation.
FEBS letters
Li Y,Ruan GX,Chen W,Huang H,Zhang R,Wang J,Ouyang Y,Zhu Z,Meng L,Wang R,Huo J,Xu S,Ou X
17-0951-82 was used in Flow Cytometry to demonstrate an essential role of Wac-mediated ubH2B in PC differentiation and shed light on the epigenetic mechanisms underlying this process.
Sat Jul 01 00:00:00 UTC 2023
CD4+ CTLs Act as a Key Effector Population for Allograft Rejection of MSCs in a Donor MHC-II Dependent Manner in Injured Liver.
Aging and disease
Shen S,Li Y,Jin M,Fan D,Pan R,Lin A,Chen Y,Xiang L,Zhao RC,Shao J
Published figure using CD95 (APO-1/Fas) monoclonal antibody (Product # 17-0951-82) in Flow Cytometry
Thu Dec 01 00:00:00 UTC 2022
Elimination of receptor binding by influenza hemagglutinin improves vaccine-induced immunity.
NPJ vaccines
Hendin HE,Lavoie PO,Gravett JM,Pillet S,Saxena P,Landry N,D'Aoust MA,Ward BJ
Published figure using CD95 (APO-1/Fas) monoclonal antibody (Product # 17-0951-82) in Flow Cytometry
Mon Apr 11 00:00:00 UTC 2022
Characterization of the spectrum of trivalent VAV1-mutation-driven tumours using a gene-edited mouse model.
Molecular oncology
Robles-Valero J,Fernández-Nevado L,Cuadrado M,Lorenzo-Martín LF,Fernández-Pisonero I,Abad A,Redín E,Montuenga L,Martín-Zanca D,Bigas A,Mallo M,Dosil M,Bustelo XR
17-0951-82 was used in Flow cytometry/Cell sorting to indicate that VAV1 mutations play critical, although highly cell-type-specific, roles in tumourigenesis.
Sat Oct 01 00:00:00 UTC 2022
The ubiquitin ligase Peli1 inhibits ICOS and thereby Tfh-mediated immunity.
Cellular & molecular immunology
Huang X,Hao S,Liu J,Huang Y,Liu M,Xiao C,Wang Y,Pei S,Yu T,Xu J,Wang H,Dai D,Su X,Xiao Y
Published figure using CD95 (APO-1/Fas) monoclonal antibody (Product # 17-0951-82) in Flow Cytometry
Thu Apr 01 00:00:00 UTC 2021
The ubiquitin ligase Peli1 inhibits ICOS and thereby Tfh-mediated immunity.
Cellular & molecular immunology
Huang X,Hao S,Liu J,Huang Y,Liu M,Xiao C,Wang Y,Pei S,Yu T,Xu J,Wang H,Dai D,Su X,Xiao Y
Published figure using CD95 (APO-1/Fas) monoclonal antibody (Product # 17-0951-82) in Flow Cytometry
Thu Apr 01 00:00:00 UTC 2021
Berberine modulates hyper-inflammation in mouse macrophages stimulated with polyinosinic-polycytidylic acid via calcium-CHOP/STAT pathway.
Scientific reports
Kim HJ,Kim YJ,Park W
Published figure using CD95 (APO-1/Fas) monoclonal antibody (Product # 17-0951-82) in Flow Cytometry
Fri May 28 00:00:00 UTC 2021
CCL9/CCR1 induces myeloid‑derived suppressor cell recruitment to the spleen in a murine H22 orthotopic hepatoma model.
Oncology reports
Li B,Zhang S,Huang N,Chen H,Wang P,Yang J,Li Z
17-0951-82 was used in Flow Cytometry to indicate that CCL9 secreted by splenic macrophages induces a C-C motif chemokine receptor 1-dependent accumulation of myeloid-derived suppressor cells in the spleen in a murine H22 hepatoma model.
Tue Jan 01 00:00:00 UTC 2019
Initiation of Antiviral B Cell Immunity Relies on Innate Signals from Spatially Positioned NKT Cells.
Cell
Gaya M,Barral P,Burbage M,Aggarwal S,Montaner B,Warren Navia A,Aid M,Tsui C,Maldonado P,Nair U,Ghneim K,Fallon PG,Sekaly RP,Barouch DH,Shalek AK,Bruckbauer A,Strid J,Batista FD
Published figure using CD95 (APO-1/Fas) monoclonal antibody (Product # 17-0951-82) in Flow Cytometry
Thu Jan 25 00:00:00 UTC 2018
Transforming Growth Factor-β and Interleukin-10 Synergistically Regulate Humoral Immunity via Modulating Metabolic Signals.
Frontiers in immunology
Komai T,Inoue M,Okamura T,Morita K,Iwasaki Y,Sumitomo S,Shoda H,Yamamoto K,Fujio K
17-0951 was used in Flow cytometry/Cell sorting to study the requirement of a combination of TGF-Β and IL-10 to suppress B cell activation induced by toll-like receptor stimulation.
Wed Nov 20 00:00:00 UTC 2019
Peli1 negatively regulates noncanonical NF-κB signaling to restrain systemic lupus erythematosus.
Nature communications
Liu J,Huang X,Hao S,Wang Y,Liu M,Xu J,Zhang X,Yu T,Gan S,Dai D,Luo X,Lu Q,Mao C,Zhang Y,Shen N,Li B,Huang M,Zhu X,Jin J,Cheng X,Sun SC,Xiao Y
Published figure using CD95 (APO-1/Fas) monoclonal antibody (Product # 17-0951-82) in Flow Cytometry
Mon Mar 19 00:00:00 UTC 2018
γδ T cells control humoral immune response by inducing T follicular helper cell differentiation.
Nature communications
Rezende RM,Lanser AJ,Rubino S,Kuhn C,Skillin N,Moreira TG,Liu S,Gabriely G,David BA,Menezes GB,Weiner HL
17-0951 was used in Flow cytometry/Cell sorting to demonstrate that complete Freund's adjuvant immunisation induces a subpopulation of CXCR5-expressing γδ T cells in the draining lymph nodes.
Wed Aug 08 00:00:00 UTC 2018
Vaccine-induced Th17 cells are established as resident memory cells in the lung and promote local IgA responses.
Mucosal immunology
Christensen D,Mortensen R,Rosenkrands I,Dietrich J,Andersen P
17-0951 was used in Flow cytometry/Cell sorting to demonstrate that subcutaneous priming with a recombinant antigen in a T helper (Th)17-inducing adjuvant followed by airway boosting promotes high and sustained levels of IgA in the lungs.
Sun Jan 01 00:00:00 UTC 2017
Armored CAR T cells enhance antitumor efficacy and overcome the tumor microenvironment.
Scientific reports
Yeku OO,Purdon TJ,Koneru M,Spriggs D,Brentjens RJ
Published figure using CD95 (APO-1/Fas) monoclonal antibody (Product # 17-0951-82) in Flow Cytometry
Tue Sep 05 00:00:00 UTC 2017
Immuno-engineered organoids for regulating the kinetics of B-cell development and antibody production.
Nature protocols
Purwada A,Singh A
17-0951-82 was used in Flow Cytometry to develop immune organoids that mimic the B-cell zone of a lymphoid tissue to efficiently generate B cells with a germinal center (GC)-like phenotype from naive murine B cells. The organoid is created using gelatin transformed into a network with biocompatible silicate nanoparticles (SiNPs).
Sun Jan 01 00:00:00 UTC 2017
YY1 plays an essential role at all stages of B-cell differentiation.
Proceedings of the National Academy of Sciences of the United States of America
Kleiman E,Jia H,Loguercio S,Su AI,Feeney AJ
Published figure using CD95 (APO-1/Fas) monoclonal antibody (Product # 17-0951-82) in Flow Cytometry
Tue Jul 05 00:00:00 UTC 2016
YY1 Regulates the Germinal Center Reaction by Inhibiting Apoptosis.
Journal of immunology (Baltimore, Md. : 1950)
Trabucco SE,Gerstein RM,Zhang H
17-0951 was used in Flow cytometry/Cell sorting to understand the role of transcription factor YY1 in the regulation of the germinal centre reaction in mice.
Thu Sep 01 00:00:00 UTC 2016
Helios Controls a Limited Subset of Regulatory T Cell Functions.
Journal of immunology (Baltimore, Md. : 1950)
Sebastian M,Lopez-Ocasio M,Metidji A,Rieder SA,Shevach EM,Thornton AM
17-0951 was used in Flow cytometry/Cell sorting to determine the beneficial effects of the transcription factor, Helios, for Treg maintenance, differentiation, and survival.
Fri Jan 01 00:00:00 UTC 2016
The histone lysine methyltransferase KMT2D sustains a gene expression program that represses B cell lymphoma development.
Nature medicine
Ortega-Molina A,Boss IW,Canela A,Pan H,Jiang Y,Zhao C,Jiang M,Hu D,Agirre X,Niesvizky I,Lee JE,Chen HT,Ennishi D,Scott DW,Mottok A,Hother C,Liu S,Cao XJ,Tam W,Shaknovich R,Garcia BA,Gascoyne RD,Ge K,Shilatifard A,Elemento O,Nussenzweig A,Melnick AM,Wendel HG
Published figure using CD95 (APO-1/Fas) monoclonal antibody (Product # 17-0951-82) in Flow Cytometry
Thu Oct 01 00:00:00 UTC 2015
Suppression of Th2 and Tfh immune reactions by Nr4a receptors in mature T reg cells.
The Journal of experimental medicine
Sekiya T,Kondo T,Shichita T,Morita R,Ichinose H,Yoshimura A
17-0951 was used in Flow cytometry/Cell sorting to demonstrate a crucial role for Nr4a factors in controlling a genetic program that is indispensable for Treg cell maintenance and function.
Mon Sep 21 00:00:00 UTC 2015
Substituting threonine 187 with alanine in p27Kip1 prevents pituitary tumorigenesis by two-hit loss of Rb1 and enhances humoral immunity in old age.
The Journal of biological chemistry
Zhao H,Bauzon F,Bi E,Yu JJ,Fu H,Lu Z,Cui J,Jeon H,Zang X,Ye BH,Zhu L
Published figure using CD95 (APO-1/Fas) monoclonal antibody (Product # 17-0951-82) in Flow Cytometry
Fri Feb 27 00:00:00 UTC 2015
Reactive oxygen species deficiency induces autoimmunity with type 1 interferon signature.
Antioxidants & redox signaling
Kelkka T,Kienhöfer D,Hoffmann M,Linja M,Wing K,Sareila O,Hultqvist M,Laajala E,Chen Z,Vasconcelos J,Neves E,Guedes M,Marques L,Krönke G,Helminen M,Kainulainen L,Olofsson P,Jalkanen S,Lahesmaa R,Souto-Carneiro MM,Holmdahl R
17-0951 was used in Flow cytometry/Cell sorting to explore the connection between reactive oxygen species deficiencies and increased susceptibility to autoimmunity.
Mon Dec 01 00:00:00 UTC 2014
Up-regulation of Fas (CD95) expression in tumour cells in vivo.
Immunology
Peshes-Yaloz N,Rosen D,Sondel PM,Krammer PH,Berke G
Published figure using CD95 (APO-1/Fas) monoclonal antibody (Product # 17-0951-82) in Flow Cytometry
Sun Apr 01 00:00:00 UTC 2007