Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: QDSRGCARWC PQNSSCVNAT ACRCNPGFSS FSEIITTPTE TCDDINECAT PSKVSCGKFS DCWNTEGSYD CVCSPGYEPV SGAKTFKNES ENTCQDVDEC QQNPRLCKSY GTCVNTLGSY TCQCLPGFKF IPEDPKVCTD VNECTSGQNP CHSSTHCLNN VGSYQCRCRP GWQPIPGSPN GPNNTVCEDV DECSSGQHQC DSSTVCFNTV GSYSCRCRPG WKPRHGIPNN QKDTVCEDMT FSTWTPPPGV HSQTLSRFFD KVQDLGRDSK TSSAEVTIQN VIKLVDELME APGDVEALAP PVRHLIATQL LSNLEDIMRI LAKSLPKGPF TYISPSNTEL TLMIQERGDK NVTMGQSSAR MKLNWAVAAG AEDPGPAV
CD97 is a G-protein-coupled seven-span transmembrane adhesive receptor that is constitutively expressed on granulocytes and monocytes and rapidly upregulated on T and B cells upon activation. CD97 is produced in alternatively spliced forms and its cellular ligand is CD55 (DAF), which protects various cell types from complement-mediated damage. Interaction of CD97 on leukocytes and CD55 on vessel cells probably facilitate leukocyte activation and migration into the tissues, similarly, CD97 seems to play a role in tumour migration and invasiveness. CD97 is involved in T cell regulation and peripheral granulocyte homeostasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adhesion G protein-coupled receptor E5; CD97; CD97 molecule; Leukocyte antigen CD97; seven transmembrane helix receptor; seven-span transmembrane protein; seven-transmembrane, heterodimeric receptor associated with inflammation
Gene Aliases: ADGRE5; CD97; TM7LN1
UniProt ID: (Human) P48960
Entrez Gene ID: (Human) 976
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support