Search Thermo Fisher Scientific
FIGURE: 1 / 6
MA5-12954 targets CD99 in WB, IF, FACS and IHC (P) applications and shows reactivity with Human and Rat samples. This antibody is not suitable for Mouse spleen cells in Western blot analysis.
The MA5-12954 immunogen is purified E-rosette forming cells from human peripheral blood lymphocytes.
CD99 (E2, MIC2) is a transmembrane glycoprotein that is involved in regulation of T cell addhesive properties and programmed cell death distinct from typical apoptosis course. CD99 roles are specific to certain stages of T cell differentiation such as corticothymocytes. CD99R isoform expression is restricted in the haematopoietic system to T, NK and myeloid cells.