Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MLPEDKEADS LRGNISVKAV KKEVEKKLRC LLADLPLPPE LPGGDDLSKS PEEKKTATQL HSKRRPKICG PRYGETKEKD IDWGKRCVDK
CDK13/cyclin K serine/threonine kinase is dimeric protein complex. CDK13 is a member of the cyclin-dependent serine/threonine protein kinase family. Members of this family are well known for their essential roles as master switches in cell cycle control. The exact function of this protein has not yet been determined, but it may play a role in mRNA processing and may be involved in regulation of hematopoiesis. Cyclin K is a member of the transcription cyclin family. These cyclins may regulate transcription through their association with and activation of cyclin-dependent kinases (CDK) that phosphorylate the C-terminal domain of the large subunit of RNA polymerase II.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC2-related protein kinase 5; Cell division cycle 2-like protein kinase 5; Cell division protein kinase 13; Cholinesterase-related cell division controller; Cyclin-dependent kinase 13; hCDK13
Gene Aliases: CDC2L; CDC2L5; CDK13; CHED; hCDK13; KIAA1791
UniProt ID: (Human) Q14004
Entrez Gene ID: (Human) 8621
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support