Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MEQPWPPPGP WSLPRAEGEA EEESDFDVFP SSPRCPQLPG GGAQMYSHGI ELACQKQKEF VKSSVACKWN LAEAQQKLGS LALHNSESLD QEHAKAQTAV SELRQREEEW RQKEEALVQR EKMCLWSTDA ISKDVFNKSF INQDKRKDTE DEDKSESFMQ KYEQKIRHFG MLSRWDDSQR FLSDHPYLVC EETAKYLILW CFHLEAEKKG ALMEQIAHQA VVMQFIMEMA KNCNVDPRGC FRLFFQKAKA EEEGYFEAFK NELEAFKSRV RLYSQSQSFQ PMTVQNHVPH SGVGSIGLLE SLPQNPDYLQ YSISTALCSL NSVVHKEDDE PKMMDTV
CDC37L1 is a cytoplasmic phosphoprotein that exists in complex with HSP90 (HSPCA; MIM 140571) as well as several other proteins involved in HSP90-mediated protein folding (Scholz et al., 2001 [PubMed 11413142]).[supplied by OMIM, Mar 2008]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC37 cell division cycle 37 homolog-like 1; cell division cycle 37 homolog-like 1; cell division cycle 37-like 1; Hsp90 co-chaperone Cdc37-like 1; Hsp90-associating relative of Cdc37; RP11-6J24.5
Gene Aliases: CDC37B; CDC37L1; HARC
UniProt ID: (Human) Q7L3B6
Entrez Gene ID: (Human) 55664
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support