Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QDLKKELKGF KTIMLNCMSL RTAQAVFPAI AQEICQEEVS RPAGKDMMRK LEKHMTAEKG PMIVLVLDEM DQLDSKGQDV LYTLFEWPWL SNSHLVLIGI ANTLDLTDRI LPRLQAREKC KPQLLNFPPY TRNQIVTILQ DRLNQVSRDQ VLDNAAVQFC ARKVSAVSGD VRKALDVCRR AIEIVESDVK SQTILKPLSE CKSPSEPLIP KRVGLIHISQ VISEIDGNRM TLSQEGAQDS FPLQQKILVC SLMLLIRQLK IKEVTLGKLY EAYSKVCRKQ QVAAVDQSEC LSLSGLLEAR GILGLKRNKE TRLTKVFFKI EEKEIEHALK DKALIGNILA TGLP
The protein encoded by this gene is highly similar to Saccharomyces cerevisiae Cdc6, a protein essential for the initiation of DNA replication. This protein functions as a regulator at the early steps of DNA replication. It localizes in cell nucleus during cell cyle G1, but translocates to the cytoplasm at the start of S phase. The subcellular translocation of this protein during cell cyle is regulated through its phosphorylation by Cdks. Transcription of this protein was reported to be regulated in response to mitogenic signals through transcriptional control mechanism involving E2F proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC18(S.pombe); Cdc18-related protein; CDC6 cell division cycle 6 homolog; CDC6-related protein; Cell division control protein 6 homolog; cell division cycle 6 homolog; HsCdc18; HsCDC6; p62(cdc6)
Gene Aliases: CDC18L; CDC6; HsCDC18; HsCDC6
UniProt ID: (Human) Q99741, (Mouse) O89033
Entrez Gene ID: (Human) 990, (Mouse) 23834, (Rat) 360621
Molecular Function:
replication origin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support