Product References
CDK1 inhibition reduces osteogenesis in endothelial cells in vascular calcification.
JCI insight
Zhao Y,Yang Y,Wu X,Zhang L,Cai X,Ji J,Chen S,Vera A,Boström KI,Yao Y
Published figure using CDK1 monoclonal antibody (Product # MA5-11472) in Western Blot
Fri Mar 08 00:00:00 UTC 2024
Deubiquitylase YOD1 regulates CDK1 stability and drives triple-negative breast cancer tumorigenesis.
Journal of experimental & clinical cancer research : CR
Han Z,Jia Q,Zhang J,Chen M,Wang L,Tong K,He W,Zhang Y,Zhu W,Qin J,Wang T,Liu T,Ma Y,Chen Y,Zha S,Zhang C
Published figure using CDK1 monoclonal antibody (Product # MA5-11472) in Immunohistochemistry
Mon Sep 04 00:00:00 UTC 2023
Deubiquitylase YOD1 regulates CDK1 stability and drives triple-negative breast cancer tumorigenesis.
Journal of experimental & clinical cancer research : CR
Han Z,Jia Q,Zhang J,Chen M,Wang L,Tong K,He W,Zhang Y,Zhu W,Qin J,Wang T,Liu T,Ma Y,Chen Y,Zha S,Zhang C
Published figure using CDK1 monoclonal antibody (Product # MA5-11472) in Immunohistochemistry
Mon Sep 04 00:00:00 UTC 2023
Prognostic value of nuclear morphometry in myxoid liposarcoma.
Cancer science
Kawaguchi K,Kohashi K,Iwasaki T,Yamamoto T,Ishihara S,Toda Y,Yamamoto H,Nakashima Y,Oda Y
MA5-11472 was used in Immunohistochemistry to reveal that the high-nuclear-grade group according to the modified Fuhrman grading system exhibited a significantly poor disease-free survival (hazard ratio: 4.43; 95% confidence interval: 0.9-22.6; p = 0.047).
Mon May 01 00:00:00 UTC 2023
The Oligomeric Assemblies of Cytomegalovirus Core Nuclear Egress Proteins Are Associated with Host Kinases and Show Sensitivity to Antiviral Kinase Inhibitors.
Viruses
Kicuntod J,Häge S,Hahn F,Sticht H,Marschall M
MA5-11472 was used in PCR to underline the functional importance of the oligomerization of the HCMV-specific NEC that is both phosphorylation-dependent and sensitive to antiviral kinase inhibitors.
Wed May 11 00:00:00 UTC 2022
Temporal Proteomic Analysis of BK Polyomavirus Infection Reveals Virus-Induced G2 Arrest and Highly Effective Evasion of Innate Immune Sensing.
Journal of virology
Caller LG,Davies CTR,Antrobus R,Lehner PJ,Weekes MP,Crump CM
Published figure using CDK1 monoclonal antibody (Product # MA5-11472) in Western Blot
Thu Aug 15 00:00:00 UTC 2019
Pharmaceutically inhibiting polo-like kinase 1 exerts a broad anti-tumour activity in retinoblastoma cell lines.
Clinical & experimental ophthalmology
Schwermer M,Dreesmann S,Eggert A,Althoff K,Steenpass L,Schramm A,Schulte JH,Temming P
Published figure using CDK1 monoclonal antibody (Product # MA5-11472) in Western Blot
Sat Apr 01 00:00:00 UTC 2017
Pharmaceutically inhibiting polo-like kinase 1 exerts a broad anti-tumour activity in retinoblastoma cell lines.
Clinical & experimental ophthalmology
Schwermer M,Dreesmann S,Eggert A,Althoff K,Steenpass L,Schramm A,Schulte JH,Temming P
Published figure using CDK1 monoclonal antibody (Product # MA5-11472) in Western Blot
Sat Apr 01 00:00:00 UTC 2017
IFI27, a novel epidermal growth factor-stabilized protein, is functionally involved in proliferation and cell cycling of human epidermal keratinocytes.
Cell proliferation
Hsieh WL,Huang YH,Wang TM,Ming YC,Tsai CN,Pang JH
MA5-11472 was used in western blot to identify the role of IFI27 in the cell proliferation of human epidermal keratinocytes
Wed Apr 01 00:00:00 UTC 2015
Genomic binding and regulation of gene expression by the thyroid carcinoma-associated PAX8-PPARG fusion protein.
Oncotarget
Zhang Y,Yu J,Lee C,Xu B,Sartor MA,Koenig RJ
MA5-11472 was used in western blot to characterize the thyroid carcinoma-associated PAX8-PPARG fusion protein and genomic binding and regulation of gene expression
Tue Dec 01 00:00:00 UTC 2015
Cyclin C is a haploinsufficient tumour suppressor.
Nature cell biology
Li N,Fassl A,Chick J,Inuzuka H,Li X,Mansour MR,Liu L,Wang H,King B,Shaik S,Gutierrez A,Ordureau A,Otto T,Kreslavsky T,Baitsch L,Bury L,Meyer CA,Ke N,Mulry KA,Kluk MJ,Roy M,Kim S,Zhang X,Geng Y,Zagozdzon A,Jenkinson S,Gale RE,Linch DC,Zhao JJ,Mullighan CG,Harper JW,Aster JC,Aifantis I,von Boehmer H,Gygi SP,Wei W,Look AT,Sicinski P
MA5-11472 was used in immunoprecipitation and western blot to elucidate the regulation of Notch by Cyclin C.
Sat Nov 01 00:00:00 UTC 2014
Cyclin C is a haploinsufficient tumour suppressor.
Nature cell biology
Li N,Fassl A,Chick J,Inuzuka H,Li X,Mansour MR,Liu L,Wang H,King B,Shaik S,Gutierrez A,Ordureau A,Otto T,Kreslavsky T,Baitsch L,Bury L,Meyer CA,Ke N,Mulry KA,Kluk MJ,Roy M,Kim S,Zhang X,Geng Y,Zagozdzon A,Jenkinson S,Gale RE,Linch DC,Zhao JJ,Mullighan CG,Harper JW,Aster JC,Aifantis I,von Boehmer H,Gygi SP,Wei W,Look AT,Sicinski P
MA5-11472 was used in immunoprecipitation and western blot to elucidate the regulation of Notch by Cyclin C.
Sat Nov 01 00:00:00 UTC 2014
The cyclin D1-CDK4 oncogenic interactome enables identification of potential novel oncogenes and clinical prognosis.
Cell cycle (Georgetown, Tex.)
Jirawatnotai S,Sharma S,Michowski W,Suktitipat B,Geng Y,Quackenbush J,Elias JE,Gygi SP,Wang YE,Sicinski P
MA5-11472 was used in western blot to identify CDK4 binding partners.
Thu Aug 20 00:00:00 UTC 2015
Repair of oxidative DNA damage, cell-cycle regulation and neuronal death may influence the clinical manifestation of Alzheimer's disease.
PloS one
Silva AR,Santos AC,Farfel JM,Grinberg LT,Ferretti RE,Campos AH,Cunha IW,Begnami MD,Rocha RM,Carraro DM,de Bragança Pereira CA,Jacob-Filho W,Brentani H
Published figure using CDK1 monoclonal antibody (Product # MA5-11472) in Immunohistochemistry
Thu Feb 12 00:00:00 UTC 2015
Repair of oxidative DNA damage, cell-cycle regulation and neuronal death may influence the clinical manifestation of Alzheimer's disease.
PloS one
Silva AR,Santos AC,Farfel JM,Grinberg LT,Ferretti RE,Campos AH,Cunha IW,Begnami MD,Rocha RM,Carraro DM,de Bragança Pereira CA,Jacob-Filho W,Brentani H
Published figure using CDK1 monoclonal antibody (Product # MA5-11472) in Immunohistochemistry
Thu Feb 12 00:00:00 UTC 2015
Activated Akt as an indicator of prognosis in gastric cancer.
Virchows Archiv : an international journal of pathology
Cinti C,Vindigni C,Zamparelli A,La Sala D,Epistolato MC,Marrelli D,Cevenini G,Tosi P
MA5-11472 was used in immunohistochemistry to examine the prognostic significance of activated AKT in gastric cancer
Sat Nov 01 00:00:00 UTC 2008
Ixocarpalactone A isolated from the Mexican tomatillo shows potent antiproliferative and apoptotic activity in colon cancer cells.
The FEBS journal
Choi JK,Murillo G,Su BN,Pezzuto JM,Kinghorn AD,Mehta RG
MA5-11472 was used in western blot to study the antiproliferative and apoptotic effects of ixocarpalactone A on colon cancer cells
Fri Dec 01 00:00:00 UTC 2006
In vitro and in vivo pharmacokinetic-pharmacodynamic relationships for the trisubstituted aminopurine cyclin-dependent kinase inhibitors olomoucine, bohemine and CYC202.
Clinical cancer research : an official journal of the American Association for Cancer Research
Raynaud FI,Whittaker SR,Fischer PM,McClue S,Walton MI,Barrie SE,Garrett MD,Rogers P,Clarke SJ,Kelland LR,Valenti M,Brunton L,Eccles S,Lane DP,Workman P
MA5-11472 was used in western blot to study the pharmacokinetics and pharmacodynamics of three tri-substituted aminopurine Cdk inhibitors in a model of colon carcinoma
Fri Jul 01 00:00:00 UTC 2005
Prognostic significance of expressions of cell-cycle regulatory proteins in gastrointestinal stromal tumor and the relevance of the risk grade.
Human pathology
Nakamura N,Yamamoto H,Yao T,Oda Y,Nishiyama K,Imamura M,Yamada T,Nawata H,Tsuneyoshi M
MA5-11472 was used in immunohistochemistry to study the prognostic value of the expression of a range of cell cycle regulatory proteins in gastrointestinal stromal tumors
Fri Jul 01 00:00:00 UTC 2005
Mitotic cell cycle proteins increase in podocytes despite lack of proliferation.
Kidney international
Petermann AT,Pippin J,Hiromura K,Monkawa T,Durvasula R,Couser WG,Kopp J,Shankland SJ
MA5-11472 was used in immunocytochemistry to study the increase in mitotic cell cycle protein in podocyes in the absence of increased proliferation
Wed Jan 01 00:00:00 UTC 2003
c-Myc alters the DNA damage-induced G2/M arrest in human mammary epithelial cells.
British journal of cancer
Sheen JH,Woo JK,Dickson RB
MA5-11472 was used in western blot to study the effect of c-Myc on DNA damage-induced G2/M arrest.
Mon Oct 20 00:00:00 UTC 2003
A novel, extraneuronal role for cyclin-dependent protein kinase 5 (CDK5): modulation of cAMP-induced apoptosis in rat leukemia cells.
The Journal of biological chemistry
Sandal T,Stapnes C,Kleivdal H,Hedin L,Døskeland SO
MA5-11472 was used in western blot to study the role cyclin-dependent protein kinase 5 in modulating cAMP-induced apoptosis in rat leukemia cells
Fri Jun 07 00:00:00 UTC 2002
Expression of the ALK protein by anaplastic large-cell lymphomas correlates with high proliferative activity.
International journal of cancer
Leoncini L,Lazzi S,Scano D,Mura A,Onida A,Massarelli G,Tosi P,Barbini P,Cevenini G,Massai MR,Pileri S,Falini B,Giordano A,Kraft R,Laissue JA,Cottier H
MA5-11472 was used in immunohistochemistry to study the correlation between ALK protein expression and the proliferative capacity of anaplastic large-cell lymphomas
Thu Jun 15 00:00:00 UTC 2000
Retinoblastoma-related p107 and pRb2/p130 proteins in malignant lymphomas: distinct mechanisms of cell growth control.
Clinical cancer research : an official journal of the American Association for Cancer Research
Leoncini L,Bellan C,Cossu A,Claudio PP,Lazzi S,Cinti C,Cevenini G,Megha T,Laurini L,Luzi P,Orcioni GF,Piccioli M,Pileri S,Giardino C,Tosi P,Giordano A
MA5-11472 was used in immunohistochemistry to study the expression and role of retinoblastoma protein/p107 and retinoblastoma protein2/p130 in non-Hodgkin's lymphoma
Wed Dec 01 00:00:00 UTC 1999
Expression of p34(cdc2) and cyclins A and B compared to other proliferative features of non-Hodgkin's lymphomas: a multivariate cluster analysis.
International journal of cancer
Leoncini L,Cossu A,Megha T,Bellan C,Lazzi S,Luzi P,Tosi P,Barbini P,Cevenini G,Pileri S,Giordano A,Kraft R,Laissue JA,Cottier H
MA5-11472 was used in immunohistochemistry to perform a multivariate cluster analysis of p34(cdc2), cyclin A and cyclin B expresssion in non-Hodgkin's lymphoma
Fri Oct 08 00:00:00 UTC 1999
Dissociation between cell cycle arrest and apoptosis can occur in Li-Fraumeni cells heterozygous for p53 gene mutations.
Oncogene
Delia D,Goi K,Mizutani S,Yamada T,Aiello A,Fontanella E,Lamorte G,Iwata S,Ishioka C,Krajewski S,Reed JC,Pierotti MA
MA5-11472 was used in Western Blotting to show that, in cells harbouring heterozygous mutations of p53, the G1 checkpoint is not necessarily disrupted, and this may be related to the endogenous p53 heterocomplexes having lost or not the capacity to bind DNA.
Thu May 08 00:00:00 UTC 1997