Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NIWPGFSKLP LVGQYSLRKQ PYNNLKHKFP WLSEAGLRLL HFLFMATAGD CLESSYFKEK PLPCEPELMP TFPHHRNKRA APATSEGQSK RCKP
The protein encoded by this gene belongs to the CDK subfamily of the Ser/Thr protein kinase family. The CDK subfamily members are highly similar to the gene products of S. cerevisiae cdc28, and S. pombe cdc2, and are known to be essential for cell cycle progression. This kinase has been shown to play a role in cellular proliferation and its function is limited to cell cycle G2-M phase. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC2-related protein kinase; Cell division protein kinase 10; cyclin dependent kinase 10; cyclin-dependent kinase (CDC2-like) 10; Cyclin-dependent kinase 10; cyclin-dependent kinase related protein; serine/threonine protein kinase PISSLRE; Serine/threonine-protein kinase PISSLRE
Gene Aliases: BC017131; CDK10; PISSLRE
UniProt ID: (Human) Q15131, (Mouse) Q3UMM4
Entrez Gene ID: (Human) 8558, (Mouse) 234854
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support