Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 6
Immunogen sequence: MGDEKDSWKV KTLDEILQEK KRRKEQEEKA EIKRLKNSDD RDSKRDSLEE GELRDHRMEI TIRNSPYRRE DSMEDRGEED DSLAIKPPQQ MSRKEKAHHR KDEKRKEKRR HRSHSAEGGK HARVKEKERE; Positive Samples: HeLa, Mouse brain; Cellular Location: Cytoplasm, Nucleus
The PITSLRE beta1 protein, a distantly related member of the Cdk family of protein kinases, induces apoptosis after low levels of ectopic expression. Apoptosis, or programmed cell death, is similarly induced by ectopic expression of an amino terminal deletion mutant retaining the catalytic and carboxyterminal domains of PITSLRE beta1, but not by other mutants lacking Histone H1 kinase activity or by other Cdk family members. The terminology for the ten isoforms of the PITSLRE subfamily of proteins is based on the conserved PSTAIRE box region of Cdc2 p34. Depending on which of the PITSLRE genes produce the protein, the cDNA and protein are designated alpha, beta or gamma (i.e., PITSLRE A gene, alpha; PITSLRE B gene, beta and PITSLRE C gene, gamma). Some of the isoforms such as PITSLRE alpha1 (T cells) and PITSLRE beta1 (B cells and brain), are expressed in specific cell types, while others are expressed ubiquitously.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC-related protein kinase p58; CDC2L3; CDK11 p110; CDK11 p46; CDK11 p58; CDK11A; cell division cycle 2 homolog-like 1; cell division cycle 2 homolog-like 2; cell division cycle 2-like 1; cell division cycle 2-like 1 (PITSLRE proteins); cell division cycle 2-like 2; Cell division cycle 2-like protein kinase 1; Cell division protein kinase 11; Cell division protein kinase 11B; CLK-1; Cyclin-dependent kinase 11; Cyclin-dependent kinase 11B; Galactosyltransferase-associated protein kinase p58/GTA; p58 CLK-1; PITSLRE proteins; PITSLRE serine/threonine-protein kinase CDC2L1
Gene Aliases: AA989746; Cdc11b; CDC2L1; Cdc2l2; CDK11; CDK11-p110; CDK11-p46; CDK11-p58; CDK11B; CLK-1; p58; p58CDC2L1; p58CLK-1; PITSLREA; PK58
UniProt ID: (Human) Q9NZS5, (Mouse) P24788
Entrez Gene ID: (Human) 984, (Mouse) 12537
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support