Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
This antibody is predicted to react with rat based on sequence homology.
Plays important functions in early cell signaling. Binds the cAMP response element (CRE) (consensus: 5'-GTGACGT[AG][AG]-3'), a sequence present in many viral and cellular promoters. Activator of the NF-ELAM1/delta-A site of the E-selectin promoter. Has no intrinsic transcriptional activity, but activates transcription on formation of JUN or FOS heterodimers. Also can bind TRE promoter sequences when heterodimerized with members of the JUN family. Isoform 4/ATF-A0acts as a dominant repressor of the E-selectin/NF-ELAM1/delta-A promoter. Isoform 5/ATF-4acts as a negative regulator, inhibiting both ATF2 and ATF7 transcriptional activities. It may exert these effects by sequestrating in the cytoplasm the Thr-53 phosphorylating kinase, preventing activation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC2-related kinase 7; CDC2-related kinase, arginine/serine-rich; CDC2-related protein kinase 7; Cell division cycle 2-related protein kinase 7; Cell division protein kinase 12; CrkRS; Cyclin-dependent kinase 12; H KIAA0904; hCDK12; protein kinase for splicing component
Gene Aliases: 1810022J16Rik; AI646528; CDK12; CRK7; CRKR; CRKRS; D11Ertd752e; KIAA0904; Pksc
UniProt ID: (Human) Q9NYV4, (Mouse) Q6ZQ27
Entrez Gene ID: (Human) 51755, (Mouse) 69131
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support