Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene contains a kinase domain most closely related to the cyclin-dependent protein kinases. The encoded kinase may activate cyclin-dependent kinase 2 and is involved in cell growth. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAK-kinase p42; CDK-activating kinase p42; Cell cycle-related kinase; Cell division protein kinase 20; Cyclin-dependent kinase 20; Cyclin-dependent protein kinase H; Cyclin-kinase-activating kinase p42
Gene Aliases: CCRK; CDCH; CDK20; P42; PNQALRE
UniProt ID: (Human) Q8IZL9
Entrez Gene ID: (Human) 23552
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support