Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSYKPIAPA PSSTPGSSTP GPGTPVPTGS VPSPSGSVPG AGAPFRPLFN DFGPPSMGYV QAMKPPGAQG SQSTYTDLLS VIEEMGKEIR PTYAGSKSAM ERLKRGIIHA RALVRECLAE TERNART (1-126 aa encoded by BC002850)
This gene encodes a protein that interacts with cyclin-dependent kinase 2 associated protein 1. Pseudogenes associated with this gene are located on chromosomes 7 and 9. Alternatively spliced transcript variants have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDK2-associated protein 2; cyclin-dependent kinase 2 associated protein 2; Cyclin-dependent kinase 2-associated protein 2; DOC-1-related protein; DOC-1R; tumor suppressor deleted in oral cancer related 1
Gene Aliases: CDK2AP2; DOC-1R; DOC1R; p14
UniProt ID: (Human) O75956
Entrez Gene ID: (Human) 10263
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support