Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: MTELRQRVAH EPVAPPEDKE SESEAKVDGE TASDSESRAE SAPLPVSADD TPEVLNRALS NLSSRWK
Provides CDP-diacylglycerol, an important precursor for the synthesis of phosphatidylinositol, phosphatidylglycerol, and cardiolipin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDP-DAG synthase 2; CDP-DG synthase 2; CDP-DG synthetase 2; CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 2; CDP-diacylglycerol synthase 2; CDP-diglyceride diphosphorylase 2; CDP-diglyceride pyrophosphorylase 2; CDP-diglyceride synthase 2; CDP-diglyceride synthetase 2; CDS 2; CTP:phosphatidate cytidylyltransferase 2; Phosphatidate cytidylyltransferase 2
Gene Aliases: CDS2
UniProt ID: (Human) O95674
Entrez Gene ID: (Human) 8760
Molecular Function: transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support