Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YLVPGMTYSK GKIYPVGYFT KENPVKGSPG VPSFAAGPPI SEGKYFSSNP
This gene encodes a protein containing KELCH-1 like domains, as well as a BTB/POZ domain. Kelch-like ECH-associated protein 1 interacts with NF-E2-related factor 2 in a redox-sensitive manner and the dissociation of the proteins in the cytoplasm is followed by transportation of NF-E2-related factor 2 to the nucleus. This interaction results in the expression of the catalytic subunit of gamma-glutamylcysteine synthetase. Two alternatively spliced transcript variants encoding the same isoform have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Corneodesmosin; D6S586E; differentiated keratinocyte S protein; S protein
Gene Aliases: CDSN; HTSS; HTSS1; HYPT2; PSS; PSS1; S
UniProt ID: (Human) Q15517
Entrez Gene ID: (Human) 1041
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support