Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
For maximum recovery of product, centrifuge the vial prior to removing the cap.
CDY1 and CDY2 (Chromodomain Y-linked 1 and 2) are genes located on the Y chromosome that encode proteins involved in chromatin remodeling and spermatogenesis. These proteins contain a chromodomain, which is important for binding to specific regions of chromatin and regulating gene expression during the development of sperm cells. Mutations or deletions in these genes can lead to male infertility due to defects in sperm development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chromodomain protein, Y chromosome, 1; chromodomain protein, Y chromosome, 2; testis-specific chromodomain protein on Y; Testis-specific chromodomain protein Y 1; Testis-specific chromodomain protein Y 2; Y chromosome chromodomain protein 2A
Gene Aliases: CDY; CDY1; CDY1A; CDY1B; CDY2; CDY2A; CDY2B
UniProt ID: (Human) Q9Y6F8, (Human) Q9Y6F7
Entrez Gene ID: (Human) 9085, (Human) 9426
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support