Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Peptide sequence: FPRTWWQSAE DVHREKIQLD LEAEFYFTHL IVMFKSPRPA AMVLDRSQDF
Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 93%; Horse: 85%; Human: 100%; Mouse: 86%; Rat: 93%
CDY1 containing a chromodomain and a histone acetyltransferase catalytic domain. Chromodomain proteins are components of heterochromatin-like complexes and can act as gene repressors. Histone hyperacetylation is thought to facilitate the transition in which protamines replace histones as the major DNA-packaging protein. This gene encodes a protein containing a chromodomain and a histone acetyltransferase catalytic domain. Chromodomain proteins are components of heterochromatin-like complexes and can act as gene repressors. This protein is localized to the nucleus of late spermatids where histone hyperacetylation takes place. Histone hyperacetylation is thought to facilitate the transition in which protamines replace histones as the major DNA-packaging protein. The human chromosome Y has two identical copies of this gene within a palindromic region; this record represents the more telomeric copy. Chromosome Y also contains a pair of closely related genes in another more telomeric palindrome as well as several related pseudogenes. Two protein isoforms are encoded by transcript variants of this gene. Additional transcript variants have been described, but their full-length nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chromodomain protein, Y chromosome, 1; testis-specific chromodomain protein on Y; Testis-specific chromodomain protein Y 1
Gene Aliases: CDY; CDY1; CDY1A; CDY1B
UniProt ID: (Human) Q9Y6F8
Entrez Gene ID: (Human) 9085
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support