Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MQRLVAWDP ACLPLPPPPP AFKSMEVANF YYEADCLAAA YGGKAAPAAP PAARPGPRPP AGELGSIGDH ERAIDFSPYL EPLGAPQAPA PATATDTFEA APPAPAPAPA SSGQHHDFLS DLFSDDYGGK NCKKPAEYGY VSLGRLGAAK GALHPGCFAP LHPPPPPPPP PAELKAEPGF EPADCKRKEE AGAPGGGAGM AAGFPYALRA YLGYQAVPSG SSGSLSTSSS SSPPGTPSPA DAKAPPTACY AGAAPAPSQV KSKAKKT (1-266 aa encoded by BC007538)
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AGP/EBP; C EBP; C/EBP BETA; C/EBP-related protein 2; CCAAT/enhancer binding protein (C/EBP), beta; CCAAT/enhancer-binding protein beta; IL-6DBP; interleukin 6-dependent DNA-binding protein; Interleukin-6-dependent-binding protein; LAP; LIP; Liver activator protein; Liver-enriched inhibitory protein; Liver-enriched transcriptional activator; Nuclear factor NF-IL6; nuclear factor of interleukin 6; nuclear factor-IL6; nuclear protein Il6; OTTMUSP00000017503; SF-B; Silencer factor B; TCF-5; Transcription factor 5; transcription factor C/EBP beta
Gene Aliases: C/EBP-beta; C/EBPbeta; CEBPB; CRP2; IL-6DBP; IL6DBP; LAP; LIP; NF-IL6; NF-M; Nfil6; PP9092; Sfb; TCF5
UniProt ID: (Human) P17676, (Mouse) P28033, (Rat) P21272
Entrez Gene ID: (Human) 1051, (Mouse) 12608, (Rat) 24253
Molecular Function: basic leucine zipper transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support