Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Immunogen sequence: SEFTITKGL RGAKTTFDVY TESWAQDPSQ ENKKKTVVDF ETDVLFLVPT EIALAQHRAN AKSAKTYAYL FSHPSRMPVY PKWVGADHAD DIQYVFGKPF ATPTGYRPQD RTVSKAMIAY WTNFAKTGDP NMGDSAVPTH WEPYTTENSG YLEITKKMGS SSMKRSLRTN FLRYWTLTYL ALPTVTDQEA TPVPPTGDSE ATPVPPTGDS ETAPVPPTGD SGAPPVPPTG DSGAPPVPPT GDSGAPPVPP TGDSGAPPVP PTGDSGAPPV PPTGDAGPPP VPPTGDSGAP PVPPTGDSGA PPVTPTGDSE TAPVPPTGDS GAPPVPPTGD SEAAPVPPTD DSKEAQMPAV IRF (371-722 aa encoded by BC042510)
Caboxyl Ester Lipase (CEL), also known as Bile Salt-Activated Lipase, catalyzes the hydrolysis of a wide range of substrates including cholesteryl esters, phospholipids, lysophospholipids, di- and tri-acylglycerols, and fatty acid esters of hydroxy fatty acids (FAHFAs). Preferentially hydrolyzes FAHFAs with the ester bond further away from the carboxylate. Unsaturated FAHFAs are hydrolyzed more quickly than saturated FAHFAs. It also has an essential role in the complete digestion of dietary lipids and their intestinal absorption, along with the absorption of fat-soluble vitamins. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BAL; Bile salt-activated lipase; bile salt-dependent lipase, oncofetal isoform; Bile salt-stimulated lipase; BSSL; bucelipase; carboxyl ester hydrolase; Carboxyl ester lipase; carboxyl ester lipase (bile salt-stimulated lipase); Cholesterol esterase; fetoacinar pancreatic protein; lysophospholipase, pancreatic; Pancreatic lysophospholipase; Sterol esterase
Gene Aliases: 1810036E18Rik; BAL; BSDL; BSSL; CEase; Cel; CELL; FAP; FAPP; Lip1; LIPA; MODY8
UniProt ID: (Mouse) Q64285
Entrez Gene ID: (Human) 1056, (Mouse) 12613, (Rat) 24254
Molecular Function:
cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support