Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CELSR3 is a member of the flamingo subfamily, part of the cadherin superfamily. The flamingo subfamily consists of nonclassic-type cadherins; a subpopulation that does not interact with catenins. The flamingo cadherins are located at the plasma membrane and have nine cadherin domains, seven epidermal growth factor-like repeats and two laminin A G-type repeats in their ectodomain. They also have seven transmembrane domains, a characteristic unique to this subfamily. It is postulated that these proteins are receptors involved in contact-mediated communication, with cadherin domains acting as homophilic binding regions and the EGF-like domains involved in cell adhesion and receptor-ligand interactions. The specific function of this particular member has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adhesion G protein-coupled receptor C3; anchor protein; Cadherin EGF LAG seven-pass G-type receptor 3; Cadherin family member 11; cadherin, EGF LAG seven-pass G-type receptor 3 (flamingo homolog, Drosophila); EGF-like protein 1; EGF-like-domain, multiple 1; Epidermal growth factor-like protein 1; Flamingo homolog 1; hFmi1; Megf3; Multiple EGF-like domains protein 2; multiple epidermal growth factor-like domains 2; Multiple epidermal growth factor-like domains protein 2
Gene Aliases: ADGRC3; CDHF11; CELSR3; EGFL1; flamingo; FMI1; HFMI1; KIAA0812; MEGF2; mKIAA0812; RESDA1
UniProt ID: (Human) Q9NYQ7, (Mouse) Q91ZI0, (Rat) O88278
Entrez Gene ID: (Human) 1951, (Mouse) 107934, (Rat) 83466
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support