Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a protein that functions in the microtubule-dependent coupling of the nucleus and the centrosome. A similar protein in mouse plays a role in both interkinetic nuclear migration, which is a characteristic pattern of nuclear movement in neural progenitors, and in neural progenitor self-renewal. Mutations in this gene are predicted to result in neurogenic defects. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: centrosomal protein 120kDa; Centrosomal protein of 120 kDa; Cep120; coiled-coil domain containing 100; Coiled-coil domain-containing protein 100
Gene Aliases: A230075C01; AU016693; CCDC100; CEP120; RGD1565619; SRTD13
UniProt ID: (Human) Q8N960, (Mouse) Q7TSG1
Entrez Gene ID: (Human) 153241, (Rat) 307302, (Mouse) 225523
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support