Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
AZI1, also named as CEP131 and KIAA1118, may play a role in spermatogenesis. It is located at the ciliary base and required for cilium formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5-azacytidine induced 1; 5-azacytidine-induced protein 1; centrosomal protein 131 kDa; centrosomal protein 131kDa; Centrosomal protein of 131 kDa; Pre-acrosome localization protein 1
Gene Aliases: AZ1; AZI1; CEP131; KIAA1118; ZA1
UniProt ID: (Human) Q9UPN4
Entrez Gene ID: (Human) 22994
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support