Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: RKLKNSQEA QQRQATLVRK LQAKVLQYRS WCQELEKRLE ATGGPIPQRW ENVEEPNLDE LLVRLEEEQQ RCESLAEVNT QLRLHMEKAD VVNKALREDV EKLTVDWSRA RDELMRKESQ WQMEQEFFKG YLKGEHGRLL SLWREVVTFR RHFLEMKSAT DRDLMELKAE HVRLSGSLLT CCLRLTVGAQ SREPNGSGRM DGREPAQLLL LLAKTQELEK EAHERSQELI QLKSQGDLEK AELQDRVTEL SALLT (40-293 aa encoded by B C001433)
Probably plays an important role in centrosome cohesion during interphase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 250 kDa centrosomal protein; C-Nap1; Centrosomal Nek2-associated protein 1; Centrosomal protein 2; centrosomal protein 250kDa; centrosome associated protein; Centrosome-associated protein CEP250; Cep250
Gene Aliases: C-NAP1; CEP2; CEP250; CNAP1
UniProt ID: (Human) Q9BV73
Entrez Gene ID: (Human) 11190
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support