Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: VISVPSWKSE RPTAISEHWD QGQRLKLSPN KYQPIQPIQT SKLEQDHFQV ARQNHFPQRQ VETTETLRAS DILTNQALES QEHLRQFSQT ETQQRDYKLV P
Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 50%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: centriolar protein CEP295; centrosomal protein 295kDa; Centrosomal protein of 295 kDa
Gene Aliases: CEP295; KIAA1731
UniProt ID: (Human) Q9C0D2
Entrez Gene ID: (Human) 85459
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support