Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Plays a role in mitotic exit and cytokinesis (PubMed:16198290, PubMed:17853893). Recruits PDCD6IP and TSG101 to midbody during cytokinesis. Required for successful completion of cytokinesis (PubMed:17853893). Not required for microtubule nucleation (PubMed:16198290). Plays a role in the development of the brain and kidney (PubMed:28264986). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen 111; centrosomal protein 55kDa; Centrosomal protein of 55 kDa; Cep55; Up-regulated in colon cancer 6
Gene Aliases: 1200008O12Rik; 2700032M20Rik; C10orf3; CEP55; CT111; RGD1305340; URCC6
UniProt ID: (Human) Q53EZ4, (Mouse) Q8BT07, (Rat) Q4V7C8
Entrez Gene ID: (Human) 55165, (Mouse) 74107, (Rat) 294074
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support