Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSGHRSTRK RCGDSHPESP VGFGHMSTTG CVLNKLFQLP TPPLSRHQLK RLEEHRYQSA GRSLLEPLMQ GYWEWLVRRV PSWIAPNLIT II (1-91 aa encoded by BC032610)
Choline phosphotransferase catalyzes the final step in the synthesis of phosphatidylcholine by the transfer of phosphocholine from CDP-choline to diacylglycerol. The synthesis of phosphatidyl ethanolamine by ethanolamine phosphotransferase occurs using an analogous reaction. This gene codes for a choline/ethanolamine phosphotransferase. The protein can synthesize either choline- or ethanolamine- containing phospholipids. Two alternatively spliced transcripts encoding the same isoform have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-alkenyl-2-acylglycerol choline phosphotransferase; choline/ethanolaminephosphotransferase; Choline/ethanolaminephosphotransferase 1; hCEPT1; mCEPT1
Gene Aliases: 9930118K05Rik; CEPT1; mCEPT1; PRO1101
UniProt ID: (Human) Q9Y6K0, (Rat) Q6AXM5, (Mouse) Q8BGS7
Entrez Gene ID: (Human) 10390, (Rat) 310773, (Mouse) 99712
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support