Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: GL261, C8D30, NIH-3T3, mouse brain.
Predicted reactivity: Mouse (83%), Rat (86%), Bovine (91%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Probable cell adhesion protein involved in leukocyte transmigration across the blood-brain barrier. Does not express any beta-galactosyltransferase activity in vitro. [UniProt]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cerebral cell adhesion molecule; Cerebral endothelial cell adhesion molecule; cerebral endothelial cell adhesion molecule 1; glycosyltransferase 25 domain containing 3; glycosyltransferase 25 family member 3; Inactive glycosyltransferase 25 family member 3; Probable inactive glycosyltransferase 25 family member 3
Gene Aliases: 2600006K01Rik; AL024097; CEECAM1; CERCAM; GLT25D3; KIAA1502
UniProt ID: (Human) Q5T4B2, (Rat) Q5U309, (Mouse) A3KGW5
Entrez Gene ID: (Human) 51148, (Rat) 296616, (Mouse) 99151
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support