Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Immunogen sequence: CRRSPRGLSP AAHLGDGSSD LILIRKCSRF NFLRFLIRHT NQQDQFDFTF VEVYRVKKFQ FTSKHMEDED SDLKEGGKKR FGHICSSHPS CCCTVSNSSW NCDGEVLHSP AIEVRVHCQL VRLFARGIEE NPKPDSHS
CERK converts ceramide to ceramide 1-phosphate (C1P), a sphingolipid metabolite. Both CERK and C1P have been implicated in various cellular processes, including proliferation, apoptosis, phagocytosis, and inflammation (Kim et al., 2006 ).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acylsphingosine kinase; Ceramide kinase; CERK antibody; hCERK; Lipid kinase 4; lipid kinase LK4; LK4; mCERK
Gene Aliases: AI848610; CERK; D330016D08Rik; dA59H18.2; dA59H18.3; hCERK; KIAA1646; LK4; mKIAA1646
UniProt ID: (Human) Q8TCT0, (Mouse) Q8K4Q7
Entrez Gene ID: (Human) 64781, (Mouse) 223753, (Rat) 300129
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support