Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: RQKTEEELLE TTLKMKFLSL DLQGDPRESQ PLLGTVIDGM LLLKTPEELQ AERNFHTVPY MVGINKQEFG WLIPMLMSYP LSEGQLDQKT AMSLLWKSYP LVCIAKELIP EATEKYLGGT DDTVKKKDLF LDLIADVMFG VPSVIVARNH RDAGAPTYMY EFQYRPSFSS DMKPKTVIGD HGDELFSVFG APFLKEGASE EEIRLSKMVM KFWANFARNG NPNGEGLPHW PEYNQKEGYL QIGANTQAAQ KLKDKEVAFW TNLFAKKAVE KPPQTEHIEL; Positive Samples: Mouse liver, A-549, U-937; Cellular Location: Endoplasmic reticulum lumen
This gene encodes a member of the carboxylesterase large family. The family members are responsible for the hydrolysis or transesterification of various xenobiotics, such as cocaine and heroin, and endogenous substrates with ester, thioester, or amide bonds. They may participate in fatty acyl and cholesterol ester metabolism, and may play a role in the blood-brain barrier system. This enzyme is the major liver enzyme and functions in liver drug clearance. Mutations of this gene cause carboxylesterase 1 deficiency. Three transcript variants encoding three different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACAT; acyl coenzyme A:cholesterol acyltransferase; Acyl-coenzyme A:cholesterol acyltransferase; Brain carboxylesterase hBr1; Carboxylesterase 1; carboxylesterase 1 (monocyte/macrophage serine esterase 1); carboxylesterase 1-like; Carboxylesterase 1C; Carboxylesterase 1E; Carboxylesterase 1G; carboxylesterase 2 (liver); CE-1; CEH; Cholesteryl ester hydrolase; Cocaine carboxylesterase; Egasyn; Es-22; ES-x; esterase 1; esterase 22; esterase-22; HMSE; human monocyte/macrophage serine esterase 1; Liver carboxylesterase 1; Liver carboxylesterase 22; Liver carboxylesterase N; Lung surfactant convertase; Methylumbelliferyl-acetate deacetylase 1; MGC117365; Monocyte/macrophage serine esterase; PES-N; REH; Retinyl ester hydrolase; Serine esterase 1; TGH; triacylglycero; Triacylglycerol hydrolase
Gene Aliases: ACAT; CE-1; CEH; Ces-1; Ces-N; CES1; Ces1c; Ces1e; Ces1g; Ces1l; CES2; Ee-1; Ee1; Eg; EG244595; egasyn; Es-22; Es-4; Es-N; Es1; Es22; Es4; EsN; Gm4976; hCE-1; HMSE; HMSE1; PCE-1; PESN; REH; Ses-1; SES1; TGH
UniProt ID: (Human) P23141, (Mouse) P23953, (Mouse) Q64176, (Mouse) Q8VCC2
Entrez Gene ID: (Human) 1066, (Mouse) 244595, (Mouse) 13884, (Mouse) 13897, (Mouse) 12623
Molecular Function:
esterase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support