Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total CES3.
CES3 major lipase in white adipose tissue. Involved in the metabolism of xenobiotics and of natural substrates. Hydrolyzes triacylglycerols and monoacylglycerols, with a preference for monoacylglycerols. The susceptibility of the substrate increases with decreasing acyl chain length of the fatty acid moiety. Catalyzes the synthesis of fatty acid ethyl esters.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carboxylesterase 1D; Carboxylesterase 3; carboxylesterase 3 (brain); Carboxylesterase 3A; EC 3.1.1; EC 3.1.1.1; ES-male; Es-male carboxylesterase; ES31FLJ21736; esterase 31; Esterase-31; FAEE synthase; Fatty acid ethyl ester synthase; Liver carboxylesterase 31; Liver carboxylesterase 31 homolog; TGH; Triacylglycerol hydrolase
Gene Aliases: Ces1; Ces1d; CES3; Ces31; Ces3a; ES31; TGH; UNQ869/PRO1887
UniProt ID: (Human) B2Z3W9, (Mouse) Q8VCT4, (Mouse) Q63880
Entrez Gene ID: (Human) 23491, (Mouse) 104158, (Mouse) 382053
Molecular Function: esterase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support