Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
This product can be used in direct ELISA.
CES7 is involved in the detoxification of xenobiotics and in the activation of ester and amide prodrugs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carboxylesterase 5A; carboxylesterase 7; Carboxylesterase-like urinary excreted protein homolog; Cauxin
Gene Aliases: 1700081L16Rik; 1700122C07Rik; BB081581; cauxin; Ces5a; Ces7; Gm503
UniProt ID: (Mouse) Q6AW46
Entrez Gene ID: (Mouse) 67935
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support