Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TQLTCDSGRV RTDAPDCYLS FHKLLLHLQG EREPGWIKQL FTNFISFTLK LVLKGQICKE INVISNIMAD FVQTRAASIL SDGDIGVDIS LTGDPVITAS YLESHHKGHF IYKNVSEDLP LPTFSPTLLG DSRMLYFWFS ERVFHSLAKV AFQDGRLMLS LMGDEFKAVL ETWGFNTNQE IFQEVVGGFP SQAQVTVHCL KMPKISCQNK GVVVNSSVMV KFLFPRPDQQ HSVAYTFEED IVT
It has been proposed that high density lipoproteins (HDLs) function jointly with lecithin:cholesterol acyltransferase and CETP to facilitate cholesterol transport from tissues to the liver. This mechanism, referred to as reverse cholesterol transport, is physiologically important because it maintains systemic cholesterol levels. CETP is responsible for neutral lipid transfer activity in plasma in numerous species. Since CETP is able to accelerate specifically the exchange of lipid components between pro- and anti-atherogenic lipoprotein fractions, it may be a key determinant of the global atherogenicity of the plasma lipoprotein profile and arises as a possible target in atherosclerosis prevention. CETP has an important role in reverse cholesterol transport and shaping and affecting the composition of plasma lipoproteins. In general elevated levels of CETP have been associated with increased risk of coronary heart disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BPI fold containing family F; Cholesteryl ester transfer protein; cholesteryl ester transfer protein plasma; cholesteryl ester transfer protein, plasma; Lipid transfer protein I
Gene Aliases: BPIFF; CETP; HDLCQ10
UniProt ID: (Human) P11597
Entrez Gene ID: (Human) 1071
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support