Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTWRHHVRLL FTVSLALQII NLGNSYQREK HNGGREEVTK VATQKHRQSP LNWTSSHFGE VTGSAEGWGP EEPLPYSRAF GEGASARPRC CRNGGTCVLG SFCVCPAHFT GRYCEHDQRR SECGALEHGA WTLRACHLCR CIFGALHCLP LQTPDRCDPK DFLASHAHGP SAGGAPSLLL LLPCALLHRL LRPDAPAHPR SLVPSVLQRE RRPCGRPGLG HRL
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CFC-1; cripto; cryptic family 1; Cryptic family protein 1; Cryptic protein; CRYPTICFLJ77897; heterotaxy 2 (autosomal dominant); HTX2MGC133213
Gene Aliases: CFC1; CFC1B; CRYPTIC; DTGA2; HTX2
UniProt ID: (Human) P0CG37
Entrez Gene ID: (Human) 55997
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support