Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MWLLVSVILI SRISSVGGEA MFCDFPKINH GILYDEEKYK PFSQVPTGEV FYYSCEYNFV SPSKSFWTRI TCAEEGWSPT PKCLRLCFFP FVENGHSESS GQTHLEGDTV QIICNTGYRL QNNENNISCV ERGWSTPPKC RSTISAEKCG PPPPIDNGDI TSFLLSVYAP GSSVEYQCQN LYQLEGNNQI TCRNGQWSEP PKCLDPCVIS QEIMEKYNIK LKWTNQQKLY SRTGDIVEFV CKSGYHPTKS HSFRAMCQNG KLVYPSCEEK
Might be involved in complement regulation. Can associate with lipoproteins and may play a role in lipid metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: complement factor H-related 2; Complement factor H-related protein 2; DDESK59; factor H-related gene 2; FHR-2; H factor (complement)-like 3; H factor-like 3; H factor-like protein 2
Gene Aliases: CFHL2; CFHR2; FHR2; HFL3
UniProt ID: (Human) P36980
Entrez Gene ID: (Human) 3080
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support