Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: WHLLHHKVPW LYRTFHKVHH QNSSSFALAT QYMSVWELFS LGFFDMMNVT LLGCHPLTTL TFHVVNIWLS VEDHSGYNFP WSTHRLVPFG WYGGVVHHDL HHSHFN
Catalyzes the formation of 25-hydroxycholesterol from cholesterol, leading to repress cholesterol biosynthetic enzymes (PubMed:9852097). Plays a key role in cell positioning and movement in lymphoid tissues: 25-hydroxycholesterol is an intermediate in biosynthesis of 7-alpha,25-dihydroxycholesterol (7-alpha,25-OHC), an oxysterol that acts as a ligand for the G protein-coupled receptor GPR183/EBI2, a chemotactic receptor for a number of lymphoid cells. May play an important role in regulating lipid metabolism by synthesizing a corepressor that blocks sterol regulatory element binding protein (SREBP) processing. In testis, production of 25-hydroxycholesterol by macrophages may play a role in Leydig cell differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1.14.99.38; Cholesterol 25-hydroxylase; Cholesterol 25-monooxygenase; h25OH
Gene Aliases: C25H; CH25H
UniProt ID: (Human) O95992
Entrez Gene ID: (Human) 9023
Molecular Function:
oxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support