Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Recommended positive controls: HeLa(4ug/ml TM_12hr, 4ug/ml TM_24hr)(30ug).
Predicted reactivity: Mouse (94%), Rat (96%), Bovine (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Pro-apoptotic component of the unfolded protein response pathway. May mediate the pro-apoptotic effects of the ATF4-ATF3-DDIT3/CHOP cascade.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Blocks Notch protein; Botch; Cation transport regulator-like protein 1; ChaC glutathione-specific gamma-glutamylcyclotransferase 1; ChaC, cation transport regulator homolog 1; ChaC, cation transport regulator-like 1; Gamma-GCG 1; gamma-GCT acting on glutathione homolog 1; Glutathione-specific gamma-glutamylcyclotransferase 1
Gene Aliases: BOTCH; CHAC1
UniProt ID: (Human) Q9BUX1
Entrez Gene ID: (Human) 79094
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support