Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
Immunogen sequence: DENENITVVK GIRLSENVID RMKESSPSGS KSQRYSGAYG ASVSDEELKR RVAEELALEQ AKKESEDQKR LKQAK
Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
Component of the MICOS complex, a large protein complex of the mitochondrial inner membrane that plays crucial roles in the maintenance of crista junctions, inner membrane architecture, and formation of contact sites to the outer membrane. Has also been shown to function as a transcription factor which binds to the BAG1 promoter and represses BAG1 transcription. Plays an important role in the maintenance of the MICOS complex stability and the mitochondrial cristae morphology.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil-helix-coiled-coil-helix domain-containing protein 3; coiled-coil-helix-coiled-coil-helix domain-containing protein 3, mitochondrial; FLJ20420; MICOS complex subunit MIC19; mitochondrial inner membrane organizing system 3; protein phosphatase 1, regulatory subunit 22
Gene Aliases: 0610041L09Rik; 1700039J09Rik; AW558177; CHCHD3; MIC19; MINOS3; PPP1R22
UniProt ID: (Human) Q9NX63, (Mouse) Q9CRB9
Entrez Gene ID: (Human) 54927, (Rat) 296966, (Mouse) 66075
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support