Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: HepG2.
Immunogen sequence: GEESSAELDY QDPDATSLKY VSGDVTHPQA GAEDALIVHC VDDSGHWGRG GLFTALEKRS AEPRKIYELA GKMKDLSLGG VLLFPVDDKE SRNKGQDLLA L
In response to DNA strand breaks, chromatin adopts a relaxed structure due to the addition of poly(ADP-ribose) (PAR) to chromatin proteins by PARP enzymes (see PARP1; MIM 173870), and this relaxation facilitates the repair of DNA damage. CHD1L interacts with PAR and has a role in chromatin relaxation following DNA damage (Ahel et al., 2009 [PubMed 19661379]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: amplified in liver cancer 1; Amplified in liver cancer protein 1; chromodomain helicase DNA binding protein 1-like; Chromodomain-helicase-DNA-binding protein 1-like
Gene Aliases: ALC1; CHD1L; CHDL
UniProt ID: (Human) Q86WJ1
Entrez Gene ID: (Human) 9557
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support